Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 33575651-33576050 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BAK1
Entrez Gene ID
578
Gene Name
BCL2 antagonist/killer 1
Gene Aliases
BAK, BAK-LIKE, BCL2L7, CDN1
UniGene
Hs.485139
Gene Chromosome Location
Chr.6: 33572546-33580293 on Build GRCh38
Transcripts
NM_001188.3 XM_011514779.2 XM_011514780.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6609847 c.104delT p.F35fs*113 Chr.6: 33575895-33575895 on Build GRCh38
6562206 c.152G>C p.G51A Chr.6: 33575847-33575847 on Build GRCh38
6428651 c.158C>T p.A53V Chr.6: 33575841-33575841 on Build GRCh38
4823382 c.125G>A p.R42H Chr.6: 33575874-33575874 on Build GRCh38
6380927 c.166G>T p.A56S Chr.6: 33575833-33575833 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs778549210 Chr.6: 33575830-33575830 on Build GRCh38
rs4987115 Chr.6: 33575916-33575916 on Build GRCh38
rs866268758 Chr.6: 33575781-33575781 on Build GRCh38
rs149960684 Chr.6: 33576013-33576013 on Build GRCh38
rs747731940 Chr.6: 33575953-33575953 on Build GRCh38
rs368815273 Chr.6: 33575875-33575875 on Build GRCh38
rs757842945 Chr.6: 33575780-33575780 on Build GRCh38
rs935374957 Chr.6: 33575748-33575748 on Build GRCh38
rs958359222 Chr.6: 33575992-33575992 on Build GRCh38
rs748774801 Chr.6: 33575820-33575820 on Build GRCh38
rs574614209 Chr.6: 33575932-33575932 on Build GRCh38
rs1051912 Chr.6: 33575844-33575844 on Build GRCh38
rs375549037 Chr.6: 33575891-33575891 on Build GRCh38
rs778260832 Chr.6: 33575945-33575945 on Build GRCh38
rs377347576 Chr.6: 33575807-33575807 on Build GRCh38
rs915224343 Chr.6: 33575779-33575779 on Build GRCh38
rs764068634 Chr.6: 33575884-33575884 on Build GRCh38
rs756559288 Chr.6: 33575903-33575903 on Build GRCh38
rs760488257 Chr.6: 33575848-33575848 on Build GRCh38
rs768052277 Chr.6: 33575825-33575825 on Build GRCh38
rs762958558 Chr.6: 33575771-33575771 on Build GRCh38
rs1051911 Chr.6: 33575874-33575874 on Build GRCh38
rs994566351 Chr.6: 33575981-33575981 on Build GRCh38
rs371989238 Chr.6: 33575796-33575796 on Build GRCh38
rs539913308 Chr.6: 33575699-33575699 on Build GRCh38
rs139218453 Chr.6: 33575700-33575700 on Build GRCh38
rs754713518 Chr.6: 33575943-33575943 on Build GRCh38
rs775583296 Chr.6: 33575857-33575857 on Build GRCh38
rs750857077 Chr.6: 33575791-33575791 on Build GRCh38
rs772953650 Chr.6: 33575847-33575847 on Build GRCh38
rs201643389 Chr.6: 33575892-33575892 on Build GRCh38
rs1027751269 Chr.6: 33575989-33575989 on Build GRCh38
rs375013861 Chr.6: 33575841-33575841 on Build GRCh38
rs752006972 Chr.6: 33575774-33575774 on Build GRCh38
rs781674780 Chr.6: 33575784-33575784 on Build GRCh38
rs959610391 Chr.6: 33575863-33575863 on Build GRCh38
rs765477406 Chr.6: 33575753-33575753 on Build GRCh38
rs772389399 Chr.6: 33575680-33575681 on Build GRCh38
rs768325698 Chr.6: 33575839-33575842 on Build GRCh38
rs770713054 Chr.6: 33575971-33575971 on Build GRCh38
rs773075870 Chr.6: 33575951-33575952 on Build GRCh38
rs368144891 Chr.6: 33575744-33575744 on Build GRCh38
rs755786933 Chr.6: 33575793-33575793 on Build GRCh38
rs543095051 Chr.6: 33575762-33575762 on Build GRCh38
rs143727127 Chr.6: 33575831-33575831 on Build GRCh38
rs752727667 Chr.6: 33575768-33575768 on Build GRCh38
rs747643368 Chr.6: 33575903-33575904 on Build GRCh38
rs376881326 Chr.6: 33575741-33575741 on Build GRCh38
rs1054756095 Chr.6: 33575697-33575697 on Build GRCh38
rs770964772 Chr.6: 33575725-33575725 on Build GRCh38
rs932439028 Chr.6: 33575736-33575736 on Build GRCh38
rs971169411 Chr.6: 33575683-33575683 on Build GRCh38
rs777705035 Chr.6: 33575964-33575964 on Build GRCh38
rs372257626 Chr.6: 33575885-33575885 on Build GRCh38
rs778587427 Chr.6: 33575877-33575878 on Build GRCh38
rs754663151 Chr.6: 33575956-33575956 on Build GRCh38
rs750765569 Chr.6: 33575893-33575893 on Build GRCh38
rs375153092 Chr.6: 33575743-33575743 on Build GRCh38
rs776547478 Chr.6: 33575973-33575973 on Build GRCh38
rs746743686 Chr.6: 33575965-33575965 on Build GRCh38
rs764595945 Chr.6: 33575773-33575773 on Build GRCh38
rs771637336 Chr.6: 33575949-33575950 on Build GRCh38
rs1012494393 Chr.6: 33575886-33575886 on Build GRCh38
rs769950150 Chr.6: 33575855-33575855 on Build GRCh38

View Full Product Details