Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 35312539-35313038 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DEF6
Entrez Gene ID
50619
Gene Name
DEF6, guanine nucleotide exchange factor
Gene Aliases
IBP, SLAT, SWAP70L
UniGene
Hs.15476
Gene Chromosome Location
Chr.6: 35297818-35321771 on Build GRCh38
Transcripts
NM_022047.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5459259 c.676C>T p.R226* Chr.6: 35312641-35312641 on Build GRCh38
742227 c.671G>T p.W224L Chr.6: 35312636-35312636 on Build GRCh38
6036799 c.756G>C p.E252D Chr.6: 35312721-35312721 on Build GRCh38
5554961 c.680G>A p.G227E Chr.6: 35312645-35312645 on Build GRCh38
1203309 c.686T>C p.L229P Chr.6: 35312651-35312651 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1041036610 Chr.6: 35312939-35312939 on Build GRCh38
rs759341284 Chr.6: 35312590-35312590 on Build GRCh38
rs190936359 Chr.6: 35312608-35312608 on Build GRCh38
rs9469989 Chr.6: 35313008-35313008 on Build GRCh38
rs374929186 Chr.6: 35312812-35312812 on Build GRCh38
rs760818846 Chr.6: 35312720-35312720 on Build GRCh38
rs374279802 Chr.6: 35312810-35312810 on Build GRCh38
rs774536963 Chr.6: 35312673-35312673 on Build GRCh38
rs187676919 Chr.6: 35312830-35312830 on Build GRCh38
rs761923772 Chr.6: 35312597-35312597 on Build GRCh38
rs755415003 Chr.6: 35312615-35312615 on Build GRCh38
rs531031369 Chr.6: 35312586-35312586 on Build GRCh38
rs980266246 Chr.6: 35312898-35312898 on Build GRCh38
rs375239867 Chr.6: 35312712-35312712 on Build GRCh38
rs777215908 Chr.6: 35312645-35312645 on Build GRCh38
rs745626187 Chr.6: 35312668-35312668 on Build GRCh38
rs922624764 Chr.6: 35313006-35313006 on Build GRCh38
rs1037290052 Chr.6: 35312930-35312930 on Build GRCh38
rs778668461 Chr.6: 35312616-35312616 on Build GRCh38
rs762001805 Chr.6: 35312682-35312682 on Build GRCh38
rs766994017 Chr.6: 35312609-35312609 on Build GRCh38
rs750925849 Chr.6: 35312598-35312598 on Build GRCh38
rs774592064 Chr.6: 35312595-35312595 on Build GRCh38
rs6938946 Chr.6: 35312920-35312920 on Build GRCh38
rs918560524 Chr.6: 35312947-35312947 on Build GRCh38
rs772086122 Chr.6: 35312872-35312872 on Build GRCh38
rs888650389 Chr.6: 35312647-35312647 on Build GRCh38
rs781081700 Chr.6: 35312818-35312818 on Build GRCh38
rs985967803 Chr.6: 35312909-35312909 on Build GRCh38
rs757924293 Chr.6: 35312805-35312805 on Build GRCh38
rs1000077318 Chr.6: 35312734-35312734 on Build GRCh38
rs747794235 Chr.6: 35312618-35312618 on Build GRCh38
rs764255696 Chr.6: 35312591-35312591 on Build GRCh38
rs754608321 Chr.6: 35312573-35312573 on Build GRCh38
rs770715951 Chr.6: 35312661-35312661 on Build GRCh38
rs776572938 Chr.6: 35312667-35312667 on Build GRCh38
rs969816391 Chr.6: 35312877-35312877 on Build GRCh38
rs377414352 Chr.6: 35312748-35312748 on Build GRCh38
rs201064463 Chr.6: 35312582-35312582 on Build GRCh38
rs756795165 Chr.6: 35312814-35312814 on Build GRCh38
rs183880571 Chr.6: 35312788-35312788 on Build GRCh38
rs777495649 Chr.6: 35312807-35312807 on Build GRCh38
rs771511140 Chr.6: 35312583-35312583 on Build GRCh38
rs776341457 Chr.6: 35312587-35312587 on Build GRCh38
rs889849912 Chr.6: 35313012-35313012 on Build GRCh38
rs981772543 Chr.6: 35312766-35312766 on Build GRCh38
rs1007334490 Chr.6: 35312640-35312640 on Build GRCh38
rs759947283 Chr.6: 35312756-35312756 on Build GRCh38
rs141509916 Chr.6: 35312929-35312929 on Build GRCh38
rs934021142 Chr.6: 35312998-35312998 on Build GRCh38
rs564635696 Chr.6: 35312646-35312646 on Build GRCh38
rs778721451 Chr.6: 35312574-35312574 on Build GRCh38
rs34676589 Chr.6: 35312603-35312604 on Build GRCh38
rs757902283 Chr.6: 35312622-35312622 on Build GRCh38
rs915074471 Chr.6: 35312840-35312840 on Build GRCh38
rs769622016 Chr.6: 35312671-35312671 on Build GRCh38
rs752391780 Chr.6: 35312802-35312802 on Build GRCh38
rs878890228 Chr.6: 35312863-35312863 on Build GRCh38
rs528442740 Chr.6: 35312747-35312747 on Build GRCh38

View Full Product Details