Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 150466592-150467104 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GIMAP8
Entrez Gene ID
155038
Gene Name
GTPase, IMAP family member 8
Gene Aliases
IAN-9, IAN6, IAN9, IANT
UniGene
Hs.647121
Gene Chromosome Location
Chr.7: 150450630-150479395 on Build GRCh38
Transcripts
NM_175571.3 XM_005249950.4 XM_005249951.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4004369 c.51A>G p.G17G Chr.7: 150466749-150466749 on Build GRCh38
364766 c.198C>G p.T66T Chr.7: 150466896-150466896 on Build GRCh38
7369459 c.219A>G p.I73M Chr.7: 150466917-150466917 on Build GRCh38
3635931 c.152G>A p.R51K Chr.7: 150466850-150466850 on Build GRCh38
1699554 c.172G>A p.E58K Chr.7: 150466870-150466870 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759062965 Chr.7: 150466648-150466648 on Build GRCh38
rs756698192 Chr.7: 150466903-150466903 on Build GRCh38
rs909925348 Chr.7: 150466994-150466994 on Build GRCh38
rs370191374 Chr.7: 150466725-150466725 on Build GRCh38
rs772996358 Chr.7: 150466940-150466940 on Build GRCh38
rs776514273 Chr.7: 150466965-150466965 on Build GRCh38
rs757525666 Chr.7: 150466781-150466781 on Build GRCh38
rs750836899 Chr.7: 150467003-150467003 on Build GRCh38
rs376916445 Chr.7: 150467029-150467029 on Build GRCh38
rs143529569 Chr.7: 150466726-150466726 on Build GRCh38
rs749342845 Chr.7: 150466847-150466848 on Build GRCh38
rs147182412 Chr.7: 150466699-150466699 on Build GRCh38
rs996767188 Chr.7: 150466718-150466718 on Build GRCh38
rs201906357 Chr.7: 150466901-150466901 on Build GRCh38
rs192459526 Chr.7: 150466800-150466800 on Build GRCh38
rs201081436 Chr.7: 150466756-150466756 on Build GRCh38
rs141075334 Chr.7: 150466764-150466764 on Build GRCh38
rs768376920 Chr.7: 150466821-150466821 on Build GRCh38
rs758956378 Chr.7: 150466850-150466850 on Build GRCh38
rs1009493171 Chr.7: 150466956-150466956 on Build GRCh38
rs146845234 Chr.7: 150466811-150466811 on Build GRCh38
rs771938080 Chr.7: 150466899-150466899 on Build GRCh38
rs372553409 Chr.7: 150466753-150466753 on Build GRCh38
rs780157150 Chr.7: 150466847-150466850 on Build GRCh38
rs746636931 Chr.7: 150466723-150466723 on Build GRCh38
rs773835781 Chr.7: 150466825-150466825 on Build GRCh38
rs959171508 Chr.7: 150466998-150466998 on Build GRCh38
rs866532158 Chr.7: 150466827-150466827 on Build GRCh38
rs767299597 Chr.7: 150466683-150466683 on Build GRCh38
rs768954714 Chr.7: 150466640-150466640 on Build GRCh38
rs762887213 Chr.7: 150466668-150466668 on Build GRCh38
rs78247321 Chr.7: 150466803-150466803 on Build GRCh38
rs775508082 Chr.7: 150466957-150466957 on Build GRCh38
rs575697809 Chr.7: 150466882-150466882 on Build GRCh38
rs745785913 Chr.7: 150467045-150467045 on Build GRCh38
rs149849472 Chr.7: 150467023-150467023 on Build GRCh38
rs752378751 Chr.7: 150466799-150466800 on Build GRCh38
rs771741644 Chr.7: 150466851-150466851 on Build GRCh38
rs753973467 Chr.7: 150466685-150466685 on Build GRCh38
rs774306095 Chr.7: 150467072-150467072 on Build GRCh38
rs780657422 Chr.7: 150466913-150466913 on Build GRCh38
rs745405463 Chr.7: 150466729-150466729 on Build GRCh38
rs991050528 Chr.7: 150466691-150466691 on Build GRCh38
rs879621875 Chr.7: 150466792-150466792 on Build GRCh38
rs372720546 Chr.7: 150466878-150466878 on Build GRCh38
rs76135975 Chr.7: 150467021-150467021 on Build GRCh38
rs983550663 Chr.7: 150466808-150466808 on Build GRCh38
rs111489735 Chr.7: 150466711-150466711 on Build GRCh38
rs61748320 Chr.7: 150467067-150467067 on Build GRCh38
rs992857235 Chr.7: 150466658-150466658 on Build GRCh38
rs2140595 Chr.7: 150466749-150466749 on Build GRCh38
rs762686337 Chr.7: 150466733-150466733 on Build GRCh38
rs764062527 Chr.7: 150466992-150466992 on Build GRCh38
rs770684528 Chr.7: 150467043-150467043 on Build GRCh38
rs918140429 Chr.7: 150466776-150466776 on Build GRCh38
rs925171820 Chr.7: 150466644-150466644 on Build GRCh38
rs758062268 Chr.7: 150467025-150467025 on Build GRCh38
rs746840063 Chr.7: 150467040-150467040 on Build GRCh38
rs370655027 Chr.7: 150466897-150466897 on Build GRCh38
rs763137255 Chr.7: 150466986-150466986 on Build GRCh38
rs140677657 Chr.7: 150466964-150466964 on Build GRCh38
rs377094452 Chr.7: 150466854-150466854 on Build GRCh38
rs150937690 Chr.7: 150466762-150466762 on Build GRCh38
rs750327624 Chr.7: 150466757-150466757 on Build GRCh38
rs541547816 Chr.7: 150466898-150466898 on Build GRCh38
rs983706109 Chr.7: 150467046-150467046 on Build GRCh38
rs147806007 Chr.7: 150466745-150466745 on Build GRCh38
rs375220398 Chr.7: 150466916-150466916 on Build GRCh38
rs774605560 Chr.7: 150466677-150466677 on Build GRCh38
rs766238778 Chr.7: 150466856-150466856 on Build GRCh38
rs918080787 Chr.7: 150466667-150466667 on Build GRCh38
rs754277049 Chr.7: 150467012-150467012 on Build GRCh38
rs761942193 Chr.7: 150466997-150466997 on Build GRCh38
rs201085075 Chr.7: 150466679-150466679 on Build GRCh38
rs771971319 Chr.7: 150466939-150466939 on Build GRCh38
rs751703860 Chr.7: 150466888-150466888 on Build GRCh38
rs779712212 Chr.7: 150466809-150466809 on Build GRCh38
rs781744259 Chr.7: 150466791-150466791 on Build GRCh38
rs372078848 Chr.7: 150466722-150466722 on Build GRCh38
rs779145567 Chr.7: 150466700-150466700 on Build GRCh38
rs145682936 Chr.7: 150467007-150467007 on Build GRCh38
rs746749799 Chr.7: 150466950-150466950 on Build GRCh38
rs747743467 Chr.7: 150466935-150466935 on Build GRCh38
rs752895303 Chr.7: 150466701-150466701 on Build GRCh38
rs751513571 Chr.7: 150466678-150466678 on Build GRCh38
rs777803867 Chr.7: 150466770-150466770 on Build GRCh38
rs775245961 Chr.7: 150466654-150466654 on Build GRCh38
rs144303536 Chr.7: 150466732-150466732 on Build GRCh38
rs375974414 Chr.7: 150466895-150466895 on Build GRCh38
rs1039562727 Chr.7: 150466896-150466896 on Build GRCh38
rs866044980 Chr.7: 150467065-150467065 on Build GRCh38
rs754574138 Chr.7: 150467017-150467017 on Build GRCh38
rs369209909 Chr.7: 150466872-150466872 on Build GRCh38
rs930093322 Chr.7: 150466968-150466968 on Build GRCh38
rs767787407 Chr.7: 150467000-150467000 on Build GRCh38
rs778674333 Chr.7: 150466932-150466932 on Build GRCh38
rs148032111 Chr.7: 150466820-150466820 on Build GRCh38
rs769594752 Chr.7: 150466652-150466652 on Build GRCh38
rs754413716 Chr.7: 150466917-150466917 on Build GRCh38
rs751981283 Chr.7: 150466775-150466775 on Build GRCh38
rs903724377 Chr.7: 150466972-150466972 on Build GRCh38
rs770460277 Chr.7: 150466761-150466761 on Build GRCh38
rs767031054 Chr.7: 150467008-150467008 on Build GRCh38
rs763932988 Chr.7: 150466669-150466669 on Build GRCh38

View Full Product Details