Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 150466978-150467521 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GIMAP8
Entrez Gene ID
155038
Gene Name
GTPase, IMAP family member 8
Gene Aliases
IAN-9, IAN6, IAN9, IANT
UniGene
Hs.647121
Gene Chromosome Location
Chr.7: 150450630-150479395 on Build GRCh38
Transcripts
NM_175571.3 XM_005249950.4 XM_005249951.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4924874 c.481T>A p.Y161N Chr.7: 150467179-150467179 on Build GRCh38
1699556 c.526G>A p.D176N Chr.7: 150467224-150467224 on Build GRCh38
7369457 c.634C>A p.Q212K Chr.7: 150467332-150467332 on Build GRCh38
3635935 c.537C>T p.I179I Chr.7: 150467235-150467235 on Build GRCh38
3635937 c.589G>A p.G197R Chr.7: 150467287-150467287 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759897976 Chr.7: 150467207-150467207 on Build GRCh38
rs1005078563 Chr.7: 150467452-150467452 on Build GRCh38
rs749750785 Chr.7: 150467263-150467263 on Build GRCh38
rs747118904 Chr.7: 150467357-150467357 on Build GRCh38
rs201302071 Chr.7: 150467220-150467220 on Build GRCh38
rs750836899 Chr.7: 150467003-150467003 on Build GRCh38
rs376916445 Chr.7: 150467029-150467029 on Build GRCh38
rs752172592 Chr.7: 150467130-150467130 on Build GRCh38
rs780846275 Chr.7: 150467241-150467241 on Build GRCh38
rs756123094 Chr.7: 150467189-150467189 on Build GRCh38
rs764523607 Chr.7: 150467290-150467290 on Build GRCh38
rs778005273 Chr.7: 150467273-150467273 on Build GRCh38
rs765695742 Chr.7: 150467208-150467208 on Build GRCh38
rs1016493894 Chr.7: 150467460-150467460 on Build GRCh38
rs746261806 Chr.7: 150467284-150467284 on Build GRCh38
rs770174770 Chr.7: 150467286-150467286 on Build GRCh38
rs766836722 Chr.7: 150467105-150467105 on Build GRCh38
rs1012337307 Chr.7: 150467128-150467128 on Build GRCh38
rs746092742 Chr.7: 150467379-150467379 on Build GRCh38
rs767618404 Chr.7: 150467137-150467137 on Build GRCh38
rs138233370 Chr.7: 150467350-150467350 on Build GRCh38
rs540607407 Chr.7: 150467257-150467257 on Build GRCh38
rs148301701 Chr.7: 150467146-150467146 on Build GRCh38
rs755917939 Chr.7: 150467317-150467317 on Build GRCh38
rs761270896 Chr.7: 150467295-150467295 on Build GRCh38
rs756911801 Chr.7: 150467232-150467232 on Build GRCh38
rs1041905726 Chr.7: 150467438-150467442 on Build GRCh38
rs761170315 Chr.7: 150467102-150467102 on Build GRCh38
rs1025057650 Chr.7: 150467395-150467395 on Build GRCh38
rs745785913 Chr.7: 150467045-150467045 on Build GRCh38
rs149849472 Chr.7: 150467023-150467023 on Build GRCh38
rs147231393 Chr.7: 150467287-150467287 on Build GRCh38
rs757868006 Chr.7: 150467131-150467131 on Build GRCh38
rs368973504 Chr.7: 150467369-150467369 on Build GRCh38
rs774306095 Chr.7: 150467072-150467072 on Build GRCh38
rs886139450 Chr.7: 150467413-150467413 on Build GRCh38
rs76135975 Chr.7: 150467021-150467021 on Build GRCh38
rs759966278 Chr.7: 150467113-150467113 on Build GRCh38
rs963614892 Chr.7: 150467470-150467470 on Build GRCh38
rs61748320 Chr.7: 150467067-150467067 on Build GRCh38
rs770684528 Chr.7: 150467043-150467043 on Build GRCh38
rs987842228 Chr.7: 150467166-150467166 on Build GRCh38
rs975284865 Chr.7: 150467490-150467490 on Build GRCh38
rs758062268 Chr.7: 150467025-150467025 on Build GRCh38
rs758616514 Chr.7: 150467262-150467262 on Build GRCh38
rs746840063 Chr.7: 150467040-150467040 on Build GRCh38
rs202178130 Chr.7: 150467218-150467218 on Build GRCh38
rs200592029 Chr.7: 150467258-150467258 on Build GRCh38
rs763584946 Chr.7: 150467227-150467227 on Build GRCh38
rs750066764 Chr.7: 150467300-150467300 on Build GRCh38
rs547768101 Chr.7: 150467443-150467443 on Build GRCh38
rs776560216 Chr.7: 150467245-150467245 on Build GRCh38
rs753219773 Chr.7: 150467215-150467215 on Build GRCh38
rs765719798 Chr.7: 150467124-150467124 on Build GRCh38
rs983706109 Chr.7: 150467046-150467046 on Build GRCh38
rs777445544 Chr.7: 150467132-150467132 on Build GRCh38
rs760973666 Chr.7: 150467188-150467188 on Build GRCh38
rs753620580 Chr.7: 150467344-150467344 on Build GRCh38
rs1029169432 Chr.7: 150467495-150467495 on Build GRCh38
rs776873502 Chr.7: 150467282-150467282 on Build GRCh38
rs143212982 Chr.7: 150467101-150467101 on Build GRCh38
rs747028717 Chr.7: 150467275-150467275 on Build GRCh38
rs766191571 Chr.7: 150467332-150467332 on Build GRCh38
rs776872577 Chr.7: 150467205-150467205 on Build GRCh38
rs763450247 Chr.7: 150467289-150467289 on Build GRCh38
rs773869620 Chr.7: 150467293-150467293 on Build GRCh38
rs758169110 Chr.7: 150467202-150467204 on Build GRCh38
rs556709763 Chr.7: 150467380-150467380 on Build GRCh38
rs754277049 Chr.7: 150467012-150467012 on Build GRCh38
rs754981356 Chr.7: 150467347-150467347 on Build GRCh38
rs375738160 Chr.7: 150467363-150467363 on Build GRCh38
rs565268033 Chr.7: 150467476-150467476 on Build GRCh38
rs777702188 Chr.7: 150467134-150467134 on Build GRCh38
rs751000509 Chr.7: 150467229-150467229 on Build GRCh38
rs779998996 Chr.7: 150467156-150467156 on Build GRCh38
rs771211629 Chr.7: 150467191-150467191 on Build GRCh38
rs527724948 Chr.7: 150467398-150467398 on Build GRCh38
rs936159090 Chr.7: 150467448-150467448 on Build GRCh38
rs866765845 Chr.7: 150467224-150467224 on Build GRCh38
rs780849961 Chr.7: 150467141-150467141 on Build GRCh38
rs145682936 Chr.7: 150467007-150467007 on Build GRCh38
rs145618535 Chr.7: 150467180-150467180 on Build GRCh38
rs771081911 Chr.7: 150467276-150467276 on Build GRCh38
rs772254030 Chr.7: 150467169-150467169 on Build GRCh38
rs903310796 Chr.7: 150467441-150467441 on Build GRCh38
rs762501861 Chr.7: 150467217-150467217 on Build GRCh38
rs116316852 Chr.7: 150467254-150467254 on Build GRCh38
rs767113572 Chr.7: 150467299-150467299 on Build GRCh38
rs757105707 Chr.7: 150467140-150467140 on Build GRCh38
rs769638956 Chr.7: 150467150-150467150 on Build GRCh38
rs866044980 Chr.7: 150467065-150467065 on Build GRCh38
rs753025993 Chr.7: 150467279-150467279 on Build GRCh38
rs754574138 Chr.7: 150467017-150467017 on Build GRCh38
rs746795576 Chr.7: 150467143-150467144 on Build GRCh38
rs149007858 Chr.7: 150467157-150467157 on Build GRCh38
rs754436007 Chr.7: 150467110-150467110 on Build GRCh38
rs767031054 Chr.7: 150467008-150467008 on Build GRCh38

View Full Product Details