Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 29069911-29070401 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KIF13B
Entrez Gene ID
23303
Gene Name
kinesin family member 13B
Gene Aliases
GAKIN
UniGene
Hs.444767
Gene Chromosome Location
Chr.8: 29067278-29263384 on Build GRCh38
Transcripts
NM_015254.3 XM_005273458.1 XM_011544457.1 XM_011544458.1 XM_011544459.2 XM_011544460.2 XM_011544461.1

SNP Detail

SNP ID Chromosome Location
rs374620807 Chr.8: 29070301-29070301 on Build GRCh38
rs952060403 Chr.8: 29070359-29070359 on Build GRCh38
rs927463584 Chr.8: 29070093-29070094 on Build GRCh38
rs966595970 Chr.8: 29070376-29070376 on Build GRCh38
rs756160986 Chr.8: 29070195-29070195 on Build GRCh38
rs899136617 Chr.8: 29070196-29070196 on Build GRCh38
rs745967397 Chr.8: 29070135-29070135 on Build GRCh38
rs930563274 Chr.8: 29070218-29070218 on Build GRCh38
rs907012104 Chr.8: 29070263-29070263 on Build GRCh38
rs1054085988 Chr.8: 29070139-29070139 on Build GRCh38
rs113089403 Chr.8: 29070099-29070099 on Build GRCh38
rs746537561 Chr.8: 29070166-29070169 on Build GRCh38
rs1022731963 Chr.8: 29070262-29070262 on Build GRCh38
rs956449467 Chr.8: 29070017-29070017 on Build GRCh38
rs961314461 Chr.8: 29070321-29070321 on Build GRCh38
rs981742045 Chr.8: 29069961-29069961 on Build GRCh38
rs143602672 Chr.8: 29070148-29070148 on Build GRCh38
rs1023188005 Chr.8: 29069933-29069933 on Build GRCh38
rs986595184 Chr.8: 29070368-29070368 on Build GRCh38
rs1029255254 Chr.8: 29069935-29069935 on Build GRCh38
rs1040086389 Chr.8: 29070069-29070069 on Build GRCh38
rs1050321895 Chr.8: 29070232-29070232 on Build GRCh38
rs1036787278 Chr.8: 29070182-29070182 on Build GRCh38
rs903462888 Chr.8: 29070099-29070099 on Build GRCh38
rs892240392 Chr.8: 29070311-29070311 on Build GRCh38
rs1021943878 Chr.8: 29070046-29070046 on Build GRCh38
rs1015534942 Chr.8: 29069992-29069992 on Build GRCh38
rs888915127 Chr.8: 29069959-29069959 on Build GRCh38
rs910659329 Chr.8: 29070375-29070375 on Build GRCh38
rs566359715 Chr.8: 29070095-29070095 on Build GRCh38
rs145704831 Chr.8: 29070356-29070356 on Build GRCh38
rs892186552 Chr.8: 29070073-29070073 on Build GRCh38
rs986186199 Chr.8: 29070027-29070027 on Build GRCh38
rs934313818 Chr.8: 29070128-29070128 on Build GRCh38
rs780428752 Chr.8: 29070253-29070253 on Build GRCh38
rs962005797 Chr.8: 29070007-29070007 on Build GRCh38
rs537457397 Chr.8: 29070320-29070320 on Build GRCh38
rs920675525 Chr.8: 29070023-29070023 on Build GRCh38
rs930688986 Chr.8: 29070025-29070025 on Build GRCh38
rs1041006669 Chr.8: 29070096-29070096 on Build GRCh38
rs546375896 Chr.8: 29070011-29070011 on Build GRCh38
rs969394436 Chr.8: 29070074-29070074 on Build GRCh38
rs973963127 Chr.8: 29070345-29070345 on Build GRCh38
rs11775056 Chr.8: 29070288-29070288 on Build GRCh38
rs972074084 Chr.8: 29070014-29070014 on Build GRCh38
rs971271070 Chr.8: 29069952-29069952 on Build GRCh38
rs538545523 Chr.8: 29070144-29070144 on Build GRCh38
rs1008603723 Chr.8: 29069980-29069980 on Build GRCh38
rs910682979 Chr.8: 29070033-29070033 on Build GRCh38

View Full Product Details