Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 33591650-33592158 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DUSP26
Entrez Gene ID
78986
Gene Name
dual specificity phosphatase 26 (putative)
Gene Aliases
DSP-4, DUSP24, LDP-4, MKP-8, MKP8, NATA1, NEAP, SKRP3
UniGene
Hs.8719
Gene Chromosome Location
Chr.8: 33591330-33600106 on Build GRCh38
Transcripts
NM_001305115.1 NM_001305116.1 NM_024025.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3648582 c.499C>T p.L167F Chr.8: 33592150-33592150 on Build GRCh38
5409496 c.522C>T p.T174T Chr.8: 33592127-33592127 on Build GRCh38
7199764 c.618G>A p.R206R Chr.8: 33592031-33592031 on Build GRCh38
1099354 c.561C>T p.G187G Chr.8: 33592088-33592088 on Build GRCh38
3648581 c.562A>C p.I188L Chr.8: 33592087-33592087 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs886656525 Chr.8: 33591903-33591903 on Build GRCh38
rs761258145 Chr.8: 33592033-33592033 on Build GRCh38
rs773834151 Chr.8: 33592016-33592016 on Build GRCh38
rs1016840220 Chr.8: 33591959-33591959 on Build GRCh38
rs189026194 Chr.8: 33591676-33591676 on Build GRCh38
rs548892457 Chr.8: 33592001-33592002 on Build GRCh38
rs369149153 Chr.8: 33592026-33592026 on Build GRCh38
rs559939925 Chr.8: 33592124-33592124 on Build GRCh38
rs753478213 Chr.8: 33591848-33591848 on Build GRCh38
rs780839636 Chr.8: 33592115-33592115 on Build GRCh38
rs556228411 Chr.8: 33591950-33591950 on Build GRCh38
rs545078102 Chr.8: 33592123-33592123 on Build GRCh38
rs149055106 Chr.8: 33592103-33592103 on Build GRCh38
rs147901900 Chr.8: 33591884-33591884 on Build GRCh38
rs909678237 Chr.8: 33591901-33591902 on Build GRCh38
rs753947633 Chr.8: 33592042-33592042 on Build GRCh38
rs895474980 Chr.8: 33591885-33591885 on Build GRCh38
rs937084147 Chr.8: 33591838-33591838 on Build GRCh38
rs769353438 Chr.8: 33592003-33592003 on Build GRCh38
rs761936757 Chr.8: 33592046-33592046 on Build GRCh38
rs539030581 Chr.8: 33591749-33591749 on Build GRCh38
rs1056110378 Chr.8: 33591971-33591971 on Build GRCh38
rs945866282 Chr.8: 33591779-33591779 on Build GRCh38
rs780047531 Chr.8: 33591983-33591983 on Build GRCh38
rs913127905 Chr.8: 33591768-33591768 on Build GRCh38
rs748987864 Chr.8: 33592007-33592007 on Build GRCh38
rs1056563721 Chr.8: 33591680-33591680 on Build GRCh38
rs369870564 Chr.8: 33591974-33591974 on Build GRCh38
rs904030526 Chr.8: 33591818-33591818 on Build GRCh38
rs954639380 Chr.8: 33591743-33591743 on Build GRCh38
rs754177344 Chr.8: 33591978-33591978 on Build GRCh38
rs962517244 Chr.8: 33591901-33591901 on Build GRCh38
rs576727362 Chr.8: 33591932-33591932 on Build GRCh38
rs751396402 Chr.8: 33591985-33591985 on Build GRCh38
rs754693833 Chr.8: 33591988-33591988 on Build GRCh38
rs953932918 Chr.8: 33591877-33591877 on Build GRCh38
rs765528913 Chr.8: 33592061-33592061 on Build GRCh38
rs1020802665 Chr.8: 33591914-33591914 on Build GRCh38
rs757523529 Chr.8: 33591982-33591982 on Build GRCh38
rs76195424 Chr.8: 33591729-33591729 on Build GRCh38
rs974808448 Chr.8: 33591732-33591732 on Build GRCh38
rs777262383 Chr.8: 33592006-33592006 on Build GRCh38
rs777630542 Chr.8: 33592126-33592126 on Build GRCh38
rs537615053 Chr.8: 33591944-33591944 on Build GRCh38
rs925760738 Chr.8: 33591806-33591806 on Build GRCh38
rs750549145 Chr.8: 33592093-33592093 on Build GRCh38
rs899268002 Chr.8: 33591960-33591960 on Build GRCh38
rs1046650608 Chr.8: 33591894-33591894 on Build GRCh38
rs1016355772 Chr.8: 33591706-33591706 on Build GRCh38
rs547511557 Chr.8: 33591840-33591840 on Build GRCh38
rs764307383 Chr.8: 33592041-33592041 on Build GRCh38
rs1004966896 Chr.8: 33591699-33591699 on Build GRCh38
rs747978858 Chr.8: 33592000-33592000 on Build GRCh38
rs966554761 Chr.8: 33591842-33591842 on Build GRCh38
rs144747809 Chr.8: 33591786-33591786 on Build GRCh38
rs973013943 Chr.8: 33591912-33591912 on Build GRCh38
rs999572115 Chr.8: 33591827-33591827 on Build GRCh38
rs978968671 Chr.8: 33591791-33591791 on Build GRCh38
rs566060029 Chr.8: 33591855-33591855 on Build GRCh38
rs949694520 Chr.8: 33591886-33591886 on Build GRCh38
rs1043889564 Chr.8: 33591750-33591750 on Build GRCh38
rs939124442 Chr.8: 33591957-33591957 on Build GRCh38
rs1029426533 Chr.8: 33591868-33591868 on Build GRCh38
rs78301676 Chr.8: 33591742-33591742 on Build GRCh38
rs144929688 Chr.8: 33592032-33592032 on Build GRCh38
rs755611051 Chr.8: 33591778-33591778 on Build GRCh38
rs555289628 Chr.8: 33591895-33591895 on Build GRCh38
rs796092759 Chr.8: 33591967-33591967 on Build GRCh38
rs996650979 Chr.8: 33591963-33591963 on Build GRCh38
rs781194345 Chr.8: 33591991-33591991 on Build GRCh38
rs1053057106 Chr.8: 33592070-33592070 on Build GRCh38
rs770590852 Chr.8: 33592012-33592012 on Build GRCh38

View Full Product Details