Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 83872606-83873144 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYLC1
Entrez Gene ID
1538
Gene Name
cylicin 1
Gene Aliases
CYCL1
UniGene
Hs.444230
Gene Chromosome Location
Chr.X: 83861126-83886700 on Build GRCh38
Transcripts
NM_001271680.1 NM_021118.2 XM_005262086.3

SNP Detail

SNP ID Chromosome Location
rs920156943 Chr.X: 83872803-83872803 on Build GRCh38
rs779359410 Chr.X: 83873040-83873040 on Build GRCh38
rs756963807 Chr.X: 83873053-83873053 on Build GRCh38
rs769298767 Chr.X: 83873061-83873061 on Build GRCh38
rs943762609 Chr.X: 83872923-83872923 on Build GRCh38
rs774130639 Chr.X: 83873085-83873085 on Build GRCh38
rs750576655 Chr.X: 83872840-83872840 on Build GRCh38
rs61531587 Chr.X: 83872754-83872755 on Build GRCh38
rs762011110 Chr.X: 83872973-83872973 on Build GRCh38
rs760842314 Chr.X: 83873116-83873116 on Build GRCh38
rs763818449 Chr.X: 83872723-83872724 on Build GRCh38
rs748842258 Chr.X: 83872718-83872718 on Build GRCh38
rs113417633 Chr.X: 83873034-83873034 on Build GRCh38
rs746589643 Chr.X: 83872897-83872897 on Build GRCh38
rs759700592 Chr.X: 83872719-83872724 on Build GRCh38
rs766396664 Chr.X: 83873006-83873006 on Build GRCh38
rs866305578 Chr.X: 83873112-83873112 on Build GRCh38
rs779448267 Chr.X: 83872891-83872891 on Build GRCh38
rs904438661 Chr.X: 83873037-83873037 on Build GRCh38
rs369082755 Chr.X: 83872719-83872720 on Build GRCh38
rs1016820405 Chr.X: 83872831-83872831 on Build GRCh38
rs759151705 Chr.X: 83872918-83872918 on Build GRCh38
rs763495580 Chr.X: 83872717-83872720 on Build GRCh38
rs775009305 Chr.X: 83872718-83872719 on Build GRCh38
rs767796942 Chr.X: 83872982-83872982 on Build GRCh38
rs755426814 Chr.X: 83873011-83873011 on Build GRCh38
rs918054587 Chr.X: 83872750-83872750 on Build GRCh38
rs762829687 Chr.X: 83872941-83872941 on Build GRCh38
rs868294207 Chr.X: 83873073-83873073 on Build GRCh38
rs968069209 Chr.X: 83872632-83872632 on Build GRCh38
rs1000903494 Chr.X: 83873038-83873038 on Build GRCh38
rs1000851132 Chr.X: 83872662-83872662 on Build GRCh38
rs762183157 Chr.X: 83872906-83872908 on Build GRCh38
rs983110874 Chr.X: 83872789-83872789 on Build GRCh38
rs769586166 Chr.X: 83872928-83872928 on Build GRCh38
rs985133569 Chr.X: 83872838-83872838 on Build GRCh38
rs781192642 Chr.X: 83872839-83872839 on Build GRCh38
rs951371643 Chr.X: 83872755-83872755 on Build GRCh38
rs762793282 Chr.X: 83872718-83872723 on Build GRCh38
rs149146924 Chr.X: 83873095-83873095 on Build GRCh38
rs190670756 Chr.X: 83872873-83872873 on Build GRCh38
rs369247752 Chr.X: 83873056-83873056 on Build GRCh38
rs775989724 Chr.X: 83872908-83872908 on Build GRCh38
rs867080166 Chr.X: 83872932-83872932 on Build GRCh38
rs748578403 Chr.X: 83873044-83873044 on Build GRCh38
rs779485878 Chr.X: 83872716-83872719 on Build GRCh38
rs374942195 Chr.X: 83872966-83872966 on Build GRCh38
rs756576595 Chr.X: 83872857-83872857 on Build GRCh38
rs61098675 Chr.X: 83872755-83872756 on Build GRCh38
rs745347208 Chr.X: 83873059-83873059 on Build GRCh38
rs750441560 Chr.X: 83872991-83872991 on Build GRCh38
rs145159196 Chr.X: 83873005-83873005 on Build GRCh38
rs769089249 Chr.X: 83872876-83872876 on Build GRCh38
rs959478014 Chr.X: 83872700-83872700 on Build GRCh38
rs113628077 Chr.X: 83872756-83872756 on Build GRCh38
rs768640100 Chr.X: 83873068-83873068 on Build GRCh38
rs77167754 Chr.X: 83872698-83872698 on Build GRCh38
rs149962384 Chr.X: 83872939-83872939 on Build GRCh38
rs200104709 Chr.X: 83872834-83872834 on Build GRCh38
rs774147166 Chr.X: 83872952-83872952 on Build GRCh38
rs755542659 Chr.X: 83873019-83873019 on Build GRCh38
rs775805617 Chr.X: 83872942-83872942 on Build GRCh38
rs773438086 Chr.X: 83873115-83873115 on Build GRCh38
rs376662712 Chr.X: 83872807-83872807 on Build GRCh38
rs770718761 Chr.X: 83872903-83872903 on Build GRCh38
rs763098413 Chr.X: 83873062-83873062 on Build GRCh38
rs774080713 Chr.X: 83872746-83872746 on Build GRCh38
rs961625787 Chr.X: 83872754-83872754 on Build GRCh38
rs780654163 Chr.X: 83872858-83872858 on Build GRCh38
rs774631294 Chr.X: 83872903-83872905 on Build GRCh38
rs761268913 Chr.X: 83872716-83872721 on Build GRCh38
rs767980804 Chr.X: 83872746-83872747 on Build GRCh38
rs764041980 Chr.X: 83873075-83873075 on Build GRCh38

View Full Product Details