Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 101487115-101487620 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARMCX4
Entrez Gene ID
100131755
Gene Name
armadillo repeat containing, X-linked 4
Gene Aliases
CXorf35, GASP4
UniGene
Hs.729062
Gene Chromosome Location
Chr.X: 101418263-101535988 on Build GRCh38
Transcripts
NR_028407.2 NR_045861.1 NR_045862.1 NR_045863.1 NR_045864.1

SNP Detail

SNP ID Chromosome Location
rs2253062 Chr.X: 101487285-101487285 on Build GRCh38
rs782130416 Chr.X: 101487290-101487290 on Build GRCh38
rs1056100161 Chr.X: 101487336-101487336 on Build GRCh38
rs782072080 Chr.X: 101487228-101487228 on Build GRCh38
rs190428887 Chr.X: 101487242-101487242 on Build GRCh38
rs900469803 Chr.X: 101487574-101487574 on Build GRCh38
rs781909563 Chr.X: 101487180-101487180 on Build GRCh38
rs1044694246 Chr.X: 101487311-101487311 on Build GRCh38
rs976156266 Chr.X: 101487151-101487151 on Build GRCh38
rs937688529 Chr.X: 101487318-101487318 on Build GRCh38
rs782443596 Chr.X: 101487352-101487352 on Build GRCh38
rs782175680 Chr.X: 101487534-101487534 on Build GRCh38
rs144789797 Chr.X: 101487296-101487296 on Build GRCh38
rs782700116 Chr.X: 101487479-101487479 on Build GRCh38
rs926286182 Chr.X: 101487314-101487314 on Build GRCh38
rs782581816 Chr.X: 101487516-101487516 on Build GRCh38
rs781918955 Chr.X: 101487149-101487149 on Build GRCh38
rs1001187898 Chr.X: 101487533-101487533 on Build GRCh38
rs945198603 Chr.X: 101487401-101487401 on Build GRCh38
rs781834790 Chr.X: 101487334-101487334 on Build GRCh38
rs182646508 Chr.X: 101487593-101487593 on Build GRCh38
rs2361298 Chr.X: 101487564-101487564 on Build GRCh38
rs896257519 Chr.X: 101487367-101487367 on Build GRCh38
rs903708300 Chr.X: 101487527-101487527 on Build GRCh38
rs950838328 Chr.X: 101487247-101487247 on Build GRCh38
rs942257670 Chr.X: 101487310-101487310 on Build GRCh38

View Full Product Details