Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 16057795-16058347 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM131C
Entrez Gene ID
348487
Gene Name
family with sequence similarity 131 member C
Gene Aliases
C1orf117
UniGene
Hs.126825
Gene Chromosome Location
Chr.1: 16057769-16073632 on Build GRCh38
Transcripts
NM_182623.2

SNP Detail

SNP ID Chromosome Location
rs529434088 Chr.1: 16057850-16057850 on Build GRCh38
rs540363999 Chr.1: 16058231-16058231 on Build GRCh38
rs3978472 Chr.1: 16058083-16058083 on Build GRCh38
rs374809250 Chr.1: 16058059-16058059 on Build GRCh38
rs577938171 Chr.1: 16058183-16058183 on Build GRCh38
rs1055081 Chr.1: 16058004-16058004 on Build GRCh38
rs956927529 Chr.1: 16057924-16057924 on Build GRCh38
rs573945567 Chr.1: 16058284-16058284 on Build GRCh38
rs144487379 Chr.1: 16058263-16058263 on Build GRCh38
rs142829093 Chr.1: 16057886-16057886 on Build GRCh38
rs368860706 Chr.1: 16057947-16057947 on Build GRCh38
rs867395228 Chr.1: 16057890-16057890 on Build GRCh38
rs531913455 Chr.1: 16058327-16058327 on Build GRCh38
rs533671488 Chr.1: 16057916-16057916 on Build GRCh38
rs547479769 Chr.1: 16057951-16057951 on Build GRCh38
rs563034635 Chr.1: 16057859-16057859 on Build GRCh38
rs576267163 Chr.1: 16058018-16058018 on Build GRCh38
rs2019490 Chr.1: 16058289-16058289 on Build GRCh38
rs6656097 Chr.1: 16058306-16058306 on Build GRCh38
rs188065560 Chr.1: 16057857-16057857 on Build GRCh38
rs146657664 Chr.1: 16057838-16057838 on Build GRCh38
rs374982892 Chr.1: 16057874-16057874 on Build GRCh38
rs2863442 Chr.1: 16057858-16057858 on Build GRCh38
rs2863443 Chr.1: 16058161-16058161 on Build GRCh38
rs902447056 Chr.1: 16058154-16058154 on Build GRCh38
rs536428970 Chr.1: 16057972-16057972 on Build GRCh38
rs567502948 Chr.1: 16057952-16057952 on Build GRCh38
rs538880637 Chr.1: 16058075-16058075 on Build GRCh38
rs112143172 Chr.1: 16057986-16057986 on Build GRCh38
rs542800921 Chr.1: 16058288-16058288 on Build GRCh38
rs572783347 Chr.1: 16058048-16058048 on Build GRCh38
rs7418808 Chr.1: 16058063-16058063 on Build GRCh38
rs571173493 Chr.1: 16057895-16057895 on Build GRCh38

View Full Product Details