Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 21986584-21987127 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CELA3B
Entrez Gene ID
23436
Gene Name
chymotrypsin like elastase family member 3B
Gene Aliases
CBPP, ELA3B
UniGene
Hs.728752
Gene Chromosome Location
Chr.1: 21976894-21989354 on Build GRCh38
Transcripts
NM_007352.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3746809 c.722C>G p.A241G Chr.1: 21986610-21986610 on Build GRCh38
6560699 c.773C>A p.A258D Chr.1: 21986661-21986661 on Build GRCh38
7216100 c.780T>C p.I260I Chr.1: 21986668-21986668 on Build GRCh38
6312557 c.702C>A p.G234G Chr.1: 21986590-21986590 on Build GRCh38
5507248 c.744G>T p.R248S Chr.1: 21986632-21986632 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs751546095 Chr.1: 21986659-21986659 on Build GRCh38
rs71638814 Chr.1: 21987028-21987028 on Build GRCh38
rs562421786 Chr.1: 21987021-21987021 on Build GRCh38
rs569841282 Chr.1: 21986647-21986647 on Build GRCh38
rs34262568 Chr.1: 21986878-21986878 on Build GRCh38
rs939935965 Chr.1: 21987008-21987008 on Build GRCh38
rs746537803 Chr.1: 21986699-21986699 on Build GRCh38
rs375487399 Chr.1: 21986671-21986671 on Build GRCh38
rs10917100 Chr.1: 21986705-21986705 on Build GRCh38
rs1045620711 Chr.1: 21986895-21986895 on Build GRCh38
rs372943878 Chr.1: 21986651-21986651 on Build GRCh38
rs547056533 Chr.1: 21986603-21986603 on Build GRCh38
rs546407308 Chr.1: 21986867-21986867 on Build GRCh38
rs760680777 Chr.1: 21986627-21986627 on Build GRCh38
rs141334622 Chr.1: 21986608-21986608 on Build GRCh38
rs776611897 Chr.1: 21986701-21986701 on Build GRCh38
rs114895362 Chr.1: 21986610-21986610 on Build GRCh38
rs3806296 Chr.1: 21986976-21986976 on Build GRCh38
rs535884453 Chr.1: 21986756-21986756 on Build GRCh38
rs755070869 Chr.1: 21986720-21986720 on Build GRCh38
rs751775191 Chr.1: 21986716-21986716 on Build GRCh38
rs766559000 Chr.1: 21986714-21986714 on Build GRCh38
rs140718049 Chr.1: 21986652-21986652 on Build GRCh38
rs932376174 Chr.1: 21987062-21987062 on Build GRCh38
rs755064029 Chr.1: 21986660-21986660 on Build GRCh38
rs991313880 Chr.1: 21986773-21986773 on Build GRCh38
rs1803530 Chr.1: 21986668-21986668 on Build GRCh38
rs768203437 Chr.1: 21986700-21986700 on Build GRCh38
rs576585295 Chr.1: 21986946-21986946 on Build GRCh38
rs775183395 Chr.1: 21986697-21986697 on Build GRCh38
rs531045738 Chr.1: 21986834-21986834 on Build GRCh38
rs34963341 Chr.1: 21986704-21986704 on Build GRCh38
rs572617109 Chr.1: 21986852-21986852 on Build GRCh38
rs979378317 Chr.1: 21986739-21986739 on Build GRCh38
rs763186531 Chr.1: 21986708-21986708 on Build GRCh38
rs866395563 Chr.1: 21986866-21986866 on Build GRCh38
rs554285160 Chr.1: 21986802-21986802 on Build GRCh38
rs188438439 Chr.1: 21986640-21986640 on Build GRCh38
rs778214872 Chr.1: 21986684-21986684 on Build GRCh38
rs1038010153 Chr.1: 21987031-21987031 on Build GRCh38
rs767575374 Chr.1: 21986728-21986728 on Build GRCh38
rs770160337 Chr.1: 21986711-21986712 on Build GRCh38
rs763983026 Chr.1: 21986638-21986638 on Build GRCh38
rs748651866 Chr.1: 21986669-21986669 on Build GRCh38
rs890907492 Chr.1: 21987100-21987100 on Build GRCh38
rs925290542 Chr.1: 21986755-21986755 on Build GRCh38
rs771572488 Chr.1: 21986694-21986694 on Build GRCh38
rs755632831 Chr.1: 21987083-21987083 on Build GRCh38
rs774820124 Chr.1: 21986626-21986626 on Build GRCh38
rs369016783 Chr.1: 21986641-21986641 on Build GRCh38
rs553137741 Chr.1: 21987051-21987051 on Build GRCh38
rs1038465465 Chr.1: 21986642-21986642 on Build GRCh38
rs887057466 Chr.1: 21986628-21986628 on Build GRCh38
rs540655667 Chr.1: 21987032-21987032 on Build GRCh38

View Full Product Details