Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 24428073-24428628 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GPLD1
Entrez Gene ID
2822
Gene Name
glycosylphosphatidylinositol specific phospholipase D1
Gene Aliases
GPIPLD, GPIPLDM, PIGPLD, PIGPLD1, PLD
UniGene
Hs.533291
Gene Chromosome Location
Chr.6: 24425832-24495163 on Build GRCh38
Transcripts
NM_001503.3 XM_017010753.1 XM_017010754.1

SNP Detail

SNP ID Chromosome Location
rs548562851 Chr.6: 24428236-24428236 on Build GRCh38
rs950728708 Chr.6: 24428307-24428307 on Build GRCh38
rs1017415041 Chr.6: 24428562-24428562 on Build GRCh38
rs900347022 Chr.6: 24428583-24428583 on Build GRCh38
rs890252281 Chr.6: 24428224-24428224 on Build GRCh38
rs1053544402 Chr.6: 24428434-24428434 on Build GRCh38
rs562790387 Chr.6: 24428197-24428197 on Build GRCh38
rs1016679162 Chr.6: 24428099-24428099 on Build GRCh38
rs74293231 Chr.6: 24428599-24428599 on Build GRCh38
rs937786407 Chr.6: 24428151-24428154 on Build GRCh38
rs969434211 Chr.6: 24428556-24428556 on Build GRCh38
rs1029301771 Chr.6: 24428120-24428120 on Build GRCh38
rs755231194 Chr.6: 24428160-24428161 on Build GRCh38
rs964808290 Chr.6: 24428292-24428295 on Build GRCh38
rs538783981 Chr.6: 24428505-24428505 on Build GRCh38
rs1029727642 Chr.6: 24428448-24428448 on Build GRCh38
rs560382217 Chr.6: 24428273-24428273 on Build GRCh38
rs893492901 Chr.6: 24428165-24428165 on Build GRCh38
rs527997645 Chr.6: 24428294-24428294 on Build GRCh38
rs899553481 Chr.6: 24428408-24428408 on Build GRCh38
rs535846321 Chr.6: 24428323-24428323 on Build GRCh38
rs56354968 Chr.6: 24428174-24428175 on Build GRCh38
rs989554001 Chr.6: 24428268-24428268 on Build GRCh38
rs950384327 Chr.6: 24428364-24428364 on Build GRCh38
rs939588613 Chr.6: 24428332-24428332 on Build GRCh38
rs558687300 Chr.6: 24428511-24428511 on Build GRCh38
rs955439645 Chr.6: 24428134-24428134 on Build GRCh38
rs915294104 Chr.6: 24428272-24428272 on Build GRCh38
rs980746731 Chr.6: 24428275-24428275 on Build GRCh38
rs751884037 Chr.6: 24428111-24428111 on Build GRCh38
rs757355029 Chr.6: 24428262-24428262 on Build GRCh38
rs1047336041 Chr.6: 24428355-24428355 on Build GRCh38
rs869258178 Chr.6: 24428171-24428174 on Build GRCh38
rs777899549 Chr.6: 24428159-24428161 on Build GRCh38
rs927475220 Chr.6: 24428390-24428390 on Build GRCh38
rs750344401 Chr.6: 24428541-24428541 on Build GRCh38
rs9467140 Chr.6: 24428238-24428238 on Build GRCh38
rs928001709 Chr.6: 24428295-24428295 on Build GRCh38
rs1053436898 Chr.6: 24428155-24428160 on Build GRCh38
rs1042112602 Chr.6: 24428539-24428539 on Build GRCh38
rs957279363 Chr.6: 24428515-24428515 on Build GRCh38
rs546816678 Chr.6: 24428460-24428460 on Build GRCh38
rs917497384 Chr.6: 24428344-24428344 on Build GRCh38
rs977394319 Chr.6: 24428210-24428210 on Build GRCh38
rs531937630 Chr.6: 24428235-24428235 on Build GRCh38
rs1038511666 Chr.6: 24428404-24428404 on Build GRCh38
rs957264286 Chr.6: 24428267-24428267 on Build GRCh38
rs941565991 Chr.6: 24428396-24428396 on Build GRCh38
rs541120776 Chr.6: 24428121-24428122 on Build GRCh38
rs994899086 Chr.6: 24428590-24428590 on Build GRCh38
rs989941351 Chr.6: 24428525-24428525 on Build GRCh38
rs755404732 Chr.6: 24428336-24428336 on Build GRCh38
rs1126621 Chr.6: 24428476-24428476 on Build GRCh38
rs544696916 Chr.6: 24428173-24428174 on Build GRCh38
rs57986398 Chr.6: 24428159-24428160 on Build GRCh38
rs767709504 Chr.6: 24428395-24428395 on Build GRCh38
rs1009457025 Chr.6: 24428173-24428173 on Build GRCh38
rs143899630 Chr.6: 24428433-24428433 on Build GRCh38
rs999175218 Chr.6: 24428488-24428488 on Build GRCh38
rs1027584393 Chr.6: 24428298-24428298 on Build GRCh38
rs963675218 Chr.6: 24428106-24428106 on Build GRCh38
rs980280776 Chr.6: 24428378-24428378 on Build GRCh38
rs747537266 Chr.6: 24428425-24428436 on Build GRCh38

View Full Product Details