Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 98849694-98850097 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GALNT12
Entrez Gene ID
79695
Gene Name
polypeptide N-acetylgalactosaminyltransferase 12
Gene Aliases
CRCS1, GalNAc-T12
UniGene
Hs.47099
Gene Chromosome Location
Chr.9: 98807699-98850081 on Build GRCh38
Transcripts
NM_024642.4 XM_006717287.1 XM_011519018.2 XM_011519020.1 XM_017015133.1

SNP Detail

SNP ID Chromosome Location
rs563124146 Chr.9: 98849800-98849800 on Build GRCh38
rs532122685 Chr.9: 98850048-98850048 on Build GRCh38
rs943692783 Chr.9: 98849917-98849917 on Build GRCh38
rs979477840 Chr.9: 98849993-98849993 on Build GRCh38
rs550688174 Chr.9: 98849939-98849939 on Build GRCh38
rs532965637 Chr.9: 98849971-98849971 on Build GRCh38
rs977111337 Chr.9: 98849857-98849857 on Build GRCh38
rs991639776 Chr.9: 98849782-98849782 on Build GRCh38
rs570330805 Chr.9: 98849944-98849944 on Build GRCh38
rs1027593348 Chr.9: 98849803-98849803 on Build GRCh38
rs751891554 Chr.9: 98849969-98849969 on Build GRCh38
rs921201676 Chr.9: 98849929-98849929 on Build GRCh38
rs543249021 Chr.9: 98849789-98849789 on Build GRCh38
rs952028579 Chr.9: 98849811-98849813 on Build GRCh38
rs1007409269 Chr.9: 98849811-98849811 on Build GRCh38
rs144829717 Chr.9: 98849774-98849774 on Build GRCh38
rs965858499 Chr.9: 98849840-98849840 on Build GRCh38
rs968906221 Chr.9: 98849827-98849827 on Build GRCh38
rs766815191 Chr.9: 98849957-98849957 on Build GRCh38
rs1037070477 Chr.9: 98849785-98849785 on Build GRCh38
rs942150107 Chr.9: 98850074-98850074 on Build GRCh38
rs1048734996 Chr.9: 98849946-98849946 on Build GRCh38
rs1017412771 Chr.9: 98849814-98849814 on Build GRCh38
rs1035913343 Chr.9: 98849769-98849769 on Build GRCh38
rs76666042 Chr.9: 98849919-98849919 on Build GRCh38
rs924232768 Chr.9: 98849898-98849898 on Build GRCh38
rs1055186209 Chr.9: 98849725-98849725 on Build GRCh38
rs896577548 Chr.9: 98849761-98849761 on Build GRCh38

View Full Product Details