Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 32129411-32129921 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FKBPL
Entrez Gene ID
63943
Gene Name
FK506 binding protein like
Gene Aliases
DIR1, FKBP4, NG7, WISP39
UniGene
Hs.520042
Gene Chromosome Location
Chr.6: 32128707-32130290 on Build GRCh38
Transcripts
NM_022110.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6664335 c.301G>A p.D101N Chr.6: 32129480-32129480 on Build GRCh38
5443554 c.8C>T p.T3M Chr.6: 32129773-32129773 on Build GRCh38
1077826 c.226G>T p.D76Y Chr.6: 32129555-32129555 on Build GRCh38
4495265 c.44C>T p.S15F Chr.6: 32129737-32129737 on Build GRCh38
451248 c.331C>G p.R111G Chr.6: 32129450-32129450 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs376123905 Chr.6: 32129455-32129455 on Build GRCh38
rs764433288 Chr.6: 32129600-32129600 on Build GRCh38
rs772594941 Chr.6: 32129735-32129735 on Build GRCh38
rs375189513 Chr.6: 32129643-32129643 on Build GRCh38
rs780338252 Chr.6: 32129831-32129831 on Build GRCh38
rs181626499 Chr.6: 32129688-32129688 on Build GRCh38
rs28732176 Chr.6: 32129513-32129513 on Build GRCh38
rs778362706 Chr.6: 32129555-32129584 on Build GRCh38
rs148821071 Chr.6: 32129730-32129730 on Build GRCh38
rs753518230 Chr.6: 32129772-32129772 on Build GRCh38
rs776870873 Chr.6: 32129557-32129557 on Build GRCh38
rs769049613 Chr.6: 32129718-32129718 on Build GRCh38
rs748063531 Chr.6: 32129671-32129671 on Build GRCh38
rs767006912 Chr.6: 32129623-32129623 on Build GRCh38
rs202133549 Chr.6: 32129666-32129666 on Build GRCh38
rs781604045 Chr.6: 32129823-32129823 on Build GRCh38
rs763363499 Chr.6: 32129573-32129573 on Build GRCh38
rs751587851 Chr.6: 32129815-32129815 on Build GRCh38
rs749969439 Chr.6: 32129633-32129633 on Build GRCh38
rs765018084 Chr.6: 32129783-32129783 on Build GRCh38
rs895923695 Chr.6: 32129649-32129649 on Build GRCh38
rs754783682 Chr.6: 32129547-32129547 on Build GRCh38
rs1000814463 Chr.6: 32129672-32129672 on Build GRCh38
rs543078519 Chr.6: 32129529-32129529 on Build GRCh38
rs748072964 Chr.6: 32129553-32129553 on Build GRCh38
rs370982277 Chr.6: 32129564-32129564 on Build GRCh38
rs377381015 Chr.6: 32129443-32129443 on Build GRCh38
rs577722330 Chr.6: 32129704-32129704 on Build GRCh38
rs1054087646 Chr.6: 32129700-32129700 on Build GRCh38
rs1029445798 Chr.6: 32129456-32129456 on Build GRCh38
rs762277748 Chr.6: 32129621-32129621 on Build GRCh38
rs749539238 Chr.6: 32129703-32129703 on Build GRCh38
rs765645856 Chr.6: 32129453-32129453 on Build GRCh38
rs1009941406 Chr.6: 32129806-32129806 on Build GRCh38
rs41268905 Chr.6: 32129834-32129834 on Build GRCh38
rs778901246 Chr.6: 32129548-32129548 on Build GRCh38
rs752432219 Chr.6: 32129501-32129501 on Build GRCh38
rs770043228 Chr.6: 32129696-32129696 on Build GRCh38
rs765944141 Chr.6: 32129766-32129766 on Build GRCh38
rs373328430 Chr.6: 32129540-32129541 on Build GRCh38
rs756258378 Chr.6: 32129830-32129830 on Build GRCh38
rs756787721 Chr.6: 32129495-32129495 on Build GRCh38
rs558245493 Chr.6: 32129774-32129774 on Build GRCh38
rs1045038881 Chr.6: 32129544-32129544 on Build GRCh38
rs1029810723 Chr.6: 32129821-32129821 on Build GRCh38
rs752064442 Chr.6: 32129487-32129487 on Build GRCh38
rs770144471 Chr.6: 32129568-32129568 on Build GRCh38
rs140876468 Chr.6: 32129608-32129608 on Build GRCh38
rs146458519 Chr.6: 32129556-32129556 on Build GRCh38
rs781382732 Chr.6: 32129680-32129680 on Build GRCh38
rs983189783 Chr.6: 32129883-32129883 on Build GRCh38
rs777831492 Chr.6: 32129802-32129802 on Build GRCh38
rs753551814 Chr.6: 32129663-32129663 on Build GRCh38
rs755789304 Chr.6: 32129522-32129522 on Build GRCh38
rs762311429 Chr.6: 32129731-32129731 on Build GRCh38
rs370458106 Chr.6: 32129476-32129476 on Build GRCh38
rs949911347 Chr.6: 32129658-32129658 on Build GRCh38
rs759127595 Chr.6: 32129773-32129773 on Build GRCh38
rs766978668 Chr.6: 32129503-32129503 on Build GRCh38
rs117160266 Chr.6: 32129698-32129698 on Build GRCh38
rs760151044 Chr.6: 32129755-32129755 on Build GRCh38
rs758307356 Chr.6: 32129797-32129797 on Build GRCh38
rs752467062 Chr.6: 32129794-32129794 on Build GRCh38
rs763485877 Chr.6: 32129472-32129472 on Build GRCh38
rs756512778 Chr.6: 32129549-32129550 on Build GRCh38
rs975725419 Chr.6: 32129740-32129740 on Build GRCh38
rs143627051 Chr.6: 32129745-32129745 on Build GRCh38
rs779787697 Chr.6: 32129525-32129525 on Build GRCh38
rs770979514 Chr.6: 32129437-32129437 on Build GRCh38
rs943639049 Chr.6: 32129545-32129545 on Build GRCh38
rs747095122 Chr.6: 32129434-32129434 on Build GRCh38
rs759884157 Chr.6: 32129450-32129450 on Build GRCh38
rs368388615 Chr.6: 32129691-32129691 on Build GRCh38
rs747865039 Chr.6: 32129735-32129736 on Build GRCh38
rs35580488 Chr.6: 32129644-32129644 on Build GRCh38
rs779948407 Chr.6: 32129852-32129852 on Build GRCh38
rs775939173 Chr.6: 32129570-32129570 on Build GRCh38
rs758274167 Chr.6: 32129676-32129676 on Build GRCh38
rs778618620 Chr.6: 32129667-32129667 on Build GRCh38

View Full Product Details