Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 32181195-32181687 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AGER
Entrez Gene ID
177
Gene Name
advanced glycosylation end product-specific receptor
Gene Aliases
RAGE, SCARJ1
UniGene
Hs.534342
Gene Chromosome Location
Chr.6: 32180968-32185247 on Build GRCh38
Transcripts
NM_001136.4 NM_001206929.1 NM_001206932.1 NM_001206934.1 NM_001206936.1 NM_001206940.1 NM_001206954.1 NM_001206966.1 NM_172197.2 NR_038190.1 XM_017010328.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4778665 c.956G>A p.R319K Chr.6: 32181358-32181358 on Build GRCh38
6596213 c.1161A>G p.E387E Chr.6: 32181197-32181197 on Build GRCh38
4817581 c.1129G>A p.E377K Chr.6: 32181229-32181229 on Build GRCh38
6354189 c.1085-6G>A p.? Chr.6: 32181483-32181483 on Build GRCh38
4816998 c.1147G>C p.E383Q Chr.6: 32181211-32181211 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs979874998 Chr.6: 32181550-32181550 on Build GRCh38
rs529787297 Chr.6: 32181373-32181373 on Build GRCh38
rs779678781 Chr.6: 32181224-32181224 on Build GRCh38
rs367834402 Chr.6: 32181367-32181367 on Build GRCh38
rs748393668 Chr.6: 32181555-32181555 on Build GRCh38
rs759732183 Chr.6: 32181360-32181360 on Build GRCh38
rs757188869 Chr.6: 32181525-32181525 on Build GRCh38
rs746986964 Chr.6: 32181508-32181508 on Build GRCh38
rs944708312 Chr.6: 32181350-32181350 on Build GRCh38
rs775408609 Chr.6: 32181249-32181249 on Build GRCh38
rs749778750 Chr.6: 32181576-32181583 on Build GRCh38
rs749432309 Chr.6: 32181545-32181545 on Build GRCh38
rs764515549 Chr.6: 32181276-32181276 on Build GRCh38
rs546564455 Chr.6: 32181356-32181356 on Build GRCh38
rs777827096 Chr.6: 32181398-32181398 on Build GRCh38
rs373730068 Chr.6: 32181366-32181366 on Build GRCh38
rs752401874 Chr.6: 32181616-32181616 on Build GRCh38
rs986387307 Chr.6: 32181628-32181628 on Build GRCh38
rs752530013 Chr.6: 32181491-32181491 on Build GRCh38
rs773772892 Chr.6: 32181459-32181459 on Build GRCh38
rs757416925 Chr.6: 32181283-32181283 on Build GRCh38
rs767852320 Chr.6: 32181450-32181450 on Build GRCh38
rs559420109 Chr.6: 32181372-32181372 on Build GRCh38
rs753000568 Chr.6: 32181265-32181265 on Build GRCh38
rs763261367 Chr.6: 32181365-32181365 on Build GRCh38
rs775637420 Chr.6: 32181407-32181407 on Build GRCh38
rs1012469882 Chr.6: 32181439-32181439 on Build GRCh38
rs768578267 Chr.6: 32181330-32181330 on Build GRCh38
rs759655645 Chr.6: 32181263-32181263 on Build GRCh38
rs758229436 Chr.6: 32181492-32181492 on Build GRCh38
rs956777325 Chr.6: 32181567-32181567 on Build GRCh38
rs768698847 Chr.6: 32181250-32181250 on Build GRCh38
rs752089701 Chr.6: 32181277-32181277 on Build GRCh38
rs758267863 Chr.6: 32181396-32181396 on Build GRCh38
rs778829903 Chr.6: 32181392-32181392 on Build GRCh38
rs749517277 Chr.6: 32181498-32181498 on Build GRCh38
rs768852655 Chr.6: 32181547-32181547 on Build GRCh38
rs3176931 Chr.6: 32181363-32181363 on Build GRCh38
rs374700378 Chr.6: 32181252-32181252 on Build GRCh38
rs368755329 Chr.6: 32181253-32181253 on Build GRCh38
rs755716881 Chr.6: 32181307-32181307 on Build GRCh38
rs781164158 Chr.6: 32181662-32181662 on Build GRCh38
rs765537169 Chr.6: 32181264-32181264 on Build GRCh38
rs749487080 Chr.6: 32181427-32181427 on Build GRCh38
rs575685813 Chr.6: 32181299-32181299 on Build GRCh38
rs749159581 Chr.6: 32181235-32181235 on Build GRCh38
rs780541384 Chr.6: 32181286-32181286 on Build GRCh38
rs552065901 Chr.6: 32181624-32181624 on Build GRCh38
rs200581867 Chr.6: 32181334-32181334 on Build GRCh38
rs753856019 Chr.6: 32181664-32181664 on Build GRCh38
rs766453544 Chr.6: 32181490-32181490 on Build GRCh38
rs762908978 Chr.6: 32181658-32181660 on Build GRCh38
rs762604177 Chr.6: 32181625-32181625 on Build GRCh38
rs143357175 Chr.6: 32181442-32181442 on Build GRCh38
rs777507435 Chr.6: 32181496-32181496 on Build GRCh38
rs768135894 Chr.6: 32181369-32181369 on Build GRCh38
rs745893963 Chr.6: 32181390-32181391 on Build GRCh38
rs775021980 Chr.6: 32181282-32181283 on Build GRCh38
rs748120205 Chr.6: 32181344-32181344 on Build GRCh38
rs772516480 Chr.6: 32181558-32181558 on Build GRCh38
rs1036107113 Chr.6: 32181351-32181351 on Build GRCh38
rs953602242 Chr.6: 32181294-32181294 on Build GRCh38
rs767034856 Chr.6: 32181585-32181585 on Build GRCh38
rs765053494 Chr.6: 32181484-32181484 on Build GRCh38
rs764928579 Chr.6: 32181605-32181605 on Build GRCh38
rs770950798 Chr.6: 32181256-32181256 on Build GRCh38
rs149093508 Chr.6: 32181432-32181432 on Build GRCh38
rs766011184 Chr.6: 32181474-32181474 on Build GRCh38
rs560762988 Chr.6: 32181266-32181266 on Build GRCh38
rs145090083 Chr.6: 32181406-32181406 on Build GRCh38
rs780073787 Chr.6: 32181527-32181527 on Build GRCh38
rs747997506 Chr.6: 32181393-32181393 on Build GRCh38
rs181811810 Chr.6: 32181288-32181288 on Build GRCh38
rs901719559 Chr.6: 32181255-32181255 on Build GRCh38
rs760066381 Chr.6: 32181466-32181466 on Build GRCh38
rs769870878 Chr.6: 32181430-32181430 on Build GRCh38
rs781455467 Chr.6: 32181283-32181284 on Build GRCh38
rs753577972 Chr.6: 32181382-32181382 on Build GRCh38
rs773545620 Chr.6: 32181573-32181573 on Build GRCh38
rs757177565 Chr.6: 32181655-32181655 on Build GRCh38
rs754772182 Chr.6: 32181387-32181387 on Build GRCh38
rs749031938 Chr.6: 32181322-32181322 on Build GRCh38
rs774569538 Chr.6: 32181549-32181549 on Build GRCh38
rs139049509 Chr.6: 32181588-32181588 on Build GRCh38
rs149968577 Chr.6: 32181364-32181364 on Build GRCh38
rs774694207 Chr.6: 32181436-32181436 on Build GRCh38
rs761224305 Chr.6: 32181584-32181584 on Build GRCh38
rs753449078 Chr.6: 32181478-32181478 on Build GRCh38
rs2071288 Chr.6: 32181483-32181483 on Build GRCh38
rs780501411 Chr.6: 32181545-32181545 on Build GRCh38
rs751347820 Chr.6: 32181648-32181648 on Build GRCh38
rs367972091 Chr.6: 32181320-32181320 on Build GRCh38
rs745880364 Chr.6: 32181404-32181404 on Build GRCh38
rs570369407 Chr.6: 32181645-32181645 on Build GRCh38

View Full Product Details