Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 209838761-209839343 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DIEXF
Entrez Gene ID
27042
Gene Name
digestive organ expansion factor homolog (zebrafish)
Gene Aliases
C1orf107, DEF, DJ434O14.5, UTP25
UniGene
Hs.194754
Gene Chromosome Location
Chr.1: 209827967-209857565 on Build GRCh38
Transcripts
NM_014388.6 XM_006711275.3 XM_011509418.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
301816 c.1100G>T p.R367L Chr.1: 209838946-209838946 on Build GRCh38
6060779 c.1139G>T p.S380I Chr.1: 209838985-209838985 on Build GRCh38
6885766 c.1102G>A p.V368M Chr.1: 209838948-209838948 on Build GRCh38
1929306 c.1245C>T p.A415A Chr.1: 209839091-209839091 on Build GRCh38
1929304 c.1085G>A p.R362Q Chr.1: 209838931-209838931 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs868520876 Chr.1: 209839197-209839197 on Build GRCh38
rs749040912 Chr.1: 209839173-209839173 on Build GRCh38
rs749595907 Chr.1: 209838893-209838893 on Build GRCh38
rs972863808 Chr.1: 209838928-209838928 on Build GRCh38
rs781329876 Chr.1: 209839150-209839150 on Build GRCh38
rs756816767 Chr.1: 209839143-209839143 on Build GRCh38
rs753377371 Chr.1: 209838989-209838989 on Build GRCh38
rs41275344 Chr.1: 209838863-209838863 on Build GRCh38
rs753057975 Chr.1: 209839111-209839111 on Build GRCh38
rs528239700 Chr.1: 209838985-209838985 on Build GRCh38
rs752173931 Chr.1: 209839079-209839079 on Build GRCh38
rs201097110 Chr.1: 209838877-209838877 on Build GRCh38
rs779723608 Chr.1: 209839168-209839168 on Build GRCh38
rs556550060 Chr.1: 209839293-209839293 on Build GRCh38
rs780616798 Chr.1: 209839165-209839167 on Build GRCh38
rs530246971 Chr.1: 209838965-209838965 on Build GRCh38
rs571163688 Chr.1: 209839037-209839037 on Build GRCh38
rs372518353 Chr.1: 209839091-209839091 on Build GRCh38
rs202202604 Chr.1: 209838881-209838881 on Build GRCh38
rs191609351 Chr.1: 209839221-209839221 on Build GRCh38
rs755377240 Chr.1: 209839104-209839104 on Build GRCh38
rs754319011 Chr.1: 209839100-209839100 on Build GRCh38
rs577652416 Chr.1: 209839258-209839258 on Build GRCh38
rs114160793 Chr.1: 209838974-209838974 on Build GRCh38
rs751920227 Chr.1: 209839000-209839000 on Build GRCh38
rs962024070 Chr.1: 209839279-209839279 on Build GRCh38
rs761960695 Chr.1: 209839074-209839074 on Build GRCh38
rs774310894 Chr.1: 209838820-209838820 on Build GRCh38
rs758000151 Chr.1: 209839118-209839118 on Build GRCh38
rs184123463 Chr.1: 209839275-209839275 on Build GRCh38
rs760337258 Chr.1: 209838905-209838905 on Build GRCh38
rs767522453 Chr.1: 209838977-209838977 on Build GRCh38
rs902849132 Chr.1: 209839049-209839049 on Build GRCh38
rs535947899 Chr.1: 209838783-209838783 on Build GRCh38
rs761990820 Chr.1: 209838975-209838975 on Build GRCh38
rs200728349 Chr.1: 209838930-209838930 on Build GRCh38
rs753650331 Chr.1: 209838854-209838854 on Build GRCh38
rs778587430 Chr.1: 209839001-209839001 on Build GRCh38
rs746473653 Chr.1: 209839036-209839036 on Build GRCh38
rs759341918 Chr.1: 209838955-209838955 on Build GRCh38
rs780152654 Chr.1: 209838870-209838873 on Build GRCh38
rs773243113 Chr.1: 209838945-209838945 on Build GRCh38
rs777170728 Chr.1: 209839026-209839026 on Build GRCh38
rs747726754 Chr.1: 209839008-209839008 on Build GRCh38
rs1040684733 Chr.1: 209838899-209838899 on Build GRCh38
rs776708760 Chr.1: 209839046-209839046 on Build GRCh38
rs879236843 Chr.1: 209838816-209838816 on Build GRCh38
rs34776375 Chr.1: 209839121-209839122 on Build GRCh38
rs907990927 Chr.1: 209839247-209839247 on Build GRCh38
rs747000268 Chr.1: 209838908-209838909 on Build GRCh38
rs770030646 Chr.1: 209838810-209838810 on Build GRCh38
rs754579920 Chr.1: 209839056-209839056 on Build GRCh38
rs139795463 Chr.1: 209838839-209838839 on Build GRCh38
rs963178213 Chr.1: 209839267-209839267 on Build GRCh38
rs1041325560 Chr.1: 209838992-209838992 on Build GRCh38
rs765552386 Chr.1: 209839110-209839110 on Build GRCh38
rs370440596 Chr.1: 209839126-209839126 on Build GRCh38
rs769678559 Chr.1: 209839059-209839059 on Build GRCh38
rs199508028 Chr.1: 209839162-209839162 on Build GRCh38
rs757281285 Chr.1: 209838876-209838876 on Build GRCh38
rs767401440 Chr.1: 209838858-209838858 on Build GRCh38
rs375130580 Chr.1: 209838948-209838948 on Build GRCh38
rs145280991 Chr.1: 209839158-209839158 on Build GRCh38
rs770943643 Chr.1: 209838950-209838950 on Build GRCh38
rs931930543 Chr.1: 209839311-209839311 on Build GRCh38
rs766248359 Chr.1: 209838853-209838853 on Build GRCh38
rs760570349 Chr.1: 209838843-209838843 on Build GRCh38
rs532263458 Chr.1: 209839047-209839047 on Build GRCh38
rs757721846 Chr.1: 209839089-209839089 on Build GRCh38
rs1054314577 Chr.1: 209839033-209839033 on Build GRCh38
rs768251579 Chr.1: 209838910-209838910 on Build GRCh38
rs201756796 Chr.1: 209838951-209838951 on Build GRCh38
rs779720285 Chr.1: 209838813-209838813 on Build GRCh38
rs756600478 Chr.1: 209838883-209838883 on Build GRCh38
rs139412208 Chr.1: 209838914-209838914 on Build GRCh38
rs544642129 Chr.1: 209838929-209838929 on Build GRCh38
rs761157719 Chr.1: 209838826-209838826 on Build GRCh38
rs143030946 Chr.1: 209839122-209839122 on Build GRCh38
rs772112258 Chr.1: 209838827-209838827 on Build GRCh38
rs778531891 Chr.1: 209838927-209838927 on Build GRCh38
rs746313784 Chr.1: 209838805-209838805 on Build GRCh38
rs766928313 Chr.1: 209839098-209839098 on Build GRCh38
rs758342770 Chr.1: 209838858-209838858 on Build GRCh38
rs765971081 Chr.1: 209838946-209838946 on Build GRCh38
rs550583148 Chr.1: 209839066-209839066 on Build GRCh38
rs986334554 Chr.1: 209839316-209839316 on Build GRCh38
rs58524905 Chr.1: 209838800-209838800 on Build GRCh38
rs764697718 Chr.1: 209839064-209839064 on Build GRCh38
rs780415427 Chr.1: 209838887-209838887 on Build GRCh38
rs766805394 Chr.1: 209838987-209838987 on Build GRCh38
rs369162277 Chr.1: 209839025-209839025 on Build GRCh38
rs41275346 Chr.1: 209839092-209839092 on Build GRCh38
rs561160476 Chr.1: 209838981-209838981 on Build GRCh38
rs746247399 Chr.1: 209839298-209839298 on Build GRCh38

View Full Product Details