Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 49638849-49639444 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FMNL3
Entrez Gene ID
91010
Gene Name
formin like 3
Gene Aliases
FHOD3, FRL2, WBP-3, WBP3
UniGene
Hs.179838
Gene Chromosome Location
Chr.12: 49636499-49707414 on Build GRCh38
Transcripts
NM_175736.4 NM_198900.2 XM_005269218.2 XM_011538968.2 XM_011538969.2 XM_011538970.2 XM_011538971.2 XM_011538972.2 XM_011538973.2 XM_011538974.2 NM_001031698.2 NM_012272.2 XM_006719324.3 XM_006719325.3 XM_011538135.2 XM_011538136.2 XM_011538137.2 XM_011538138.2 XM_011538139.2 XM_011538140.2 XM_011538141.1 XM_011538143.2 XM_011538144.2 XM_017019135.1 XM_017019136.1 XM_017019137.1 XM_017019138.1 XM_017019139.1

Gene Symbol
PRPF40B
Entrez Gene ID
25766
Gene Name
pre-mRNA processing factor 40 homolog B
Gene Aliases
HYPC
UniGene
Hs.706827
Gene Chromosome Location
Chr.12: 49622717-49644669 on Build GRCh38
Transcripts
NM_175736.4 NM_198900.2 XM_005269218.2 XM_011538968.2 XM_011538969.2 XM_011538970.2 XM_011538971.2 XM_011538972.2 XM_011538973.2 XM_011538974.2 NM_001031698.2 NM_012272.2 XM_006719324.3 XM_006719325.3 XM_011538135.2 XM_011538136.2 XM_011538137.2 XM_011538138.2 XM_011538139.2 XM_011538140.2 XM_011538141.1 XM_011538143.2 XM_011538144.2 XM_017019135.1 XM_017019136.1 XM_017019137.1 XM_017019138.1 XM_017019139.1

SNP Detail

SNP ID Chromosome Location
rs996106233 Chr.12: 49639018-49639018 on Build GRCh38
rs987750754 Chr.12: 49638895-49638896 on Build GRCh38
rs1044827528 Chr.12: 49639075-49639075 on Build GRCh38
rs767922917 Chr.12: 49639406-49639406 on Build GRCh38
rs541446381 Chr.12: 49639203-49639203 on Build GRCh38
rs561753080 Chr.12: 49639362-49639362 on Build GRCh38
rs1034013000 Chr.12: 49639152-49639152 on Build GRCh38
rs577062684 Chr.12: 49639021-49639021 on Build GRCh38
rs944015637 Chr.12: 49638959-49638959 on Build GRCh38
rs1028772936 Chr.12: 49639389-49639389 on Build GRCh38
rs967546388 Chr.12: 49639182-49639182 on Build GRCh38
rs997213396 Chr.12: 49639171-49639171 on Build GRCh38
rs914736927 Chr.12: 49638871-49638871 on Build GRCh38
rs900390579 Chr.12: 49639004-49639004 on Build GRCh38
rs914746976 Chr.12: 49639172-49639172 on Build GRCh38
rs971060004 Chr.12: 49639121-49639121 on Build GRCh38
rs140570681 Chr.12: 49638904-49638904 on Build GRCh38
rs146710003 Chr.12: 49639412-49639412 on Build GRCh38
rs527623208 Chr.12: 49639290-49639291 on Build GRCh38
rs1027936532 Chr.12: 49639020-49639020 on Build GRCh38
rs921269539 Chr.12: 49639263-49639263 on Build GRCh38
rs1057066812 Chr.12: 49639135-49639135 on Build GRCh38
rs145776505 Chr.12: 49639081-49639081 on Build GRCh38
rs534052720 Chr.12: 49638938-49638938 on Build GRCh38
rs538769965 Chr.12: 49638935-49638935 on Build GRCh38
rs904900935 Chr.12: 49639078-49639078 on Build GRCh38
rs370284480 Chr.12: 49639045-49639045 on Build GRCh38
rs184849677 Chr.12: 49639175-49639175 on Build GRCh38
rs1005063603 Chr.12: 49639049-49639049 on Build GRCh38
rs911752406 Chr.12: 49639197-49639197 on Build GRCh38
rs751239860 Chr.12: 49639218-49639218 on Build GRCh38
rs912615126 Chr.12: 49639211-49639211 on Build GRCh38
rs762497372 Chr.12: 49639154-49639154 on Build GRCh38
rs553939530 Chr.12: 49638987-49638987 on Build GRCh38
rs971988505 Chr.12: 49639207-49639207 on Build GRCh38
rs1022477563 Chr.12: 49639158-49639158 on Build GRCh38
rs765967406 Chr.12: 49639289-49639290 on Build GRCh38
rs759413028 Chr.12: 49639063-49639063 on Build GRCh38
rs911723483 Chr.12: 49638929-49638929 on Build GRCh38
rs764827612 Chr.12: 49639072-49639072 on Build GRCh38
rs548578177 Chr.12: 49638931-49638931 on Build GRCh38
rs1869842 Chr.12: 49639000-49639000 on Build GRCh38
rs990402794 Chr.12: 49639169-49639169 on Build GRCh38
rs763706599 Chr.12: 49639199-49639199 on Build GRCh38
rs1039719868 Chr.12: 49639239-49639239 on Build GRCh38
rs986891423 Chr.12: 49639180-49639180 on Build GRCh38
rs530314383 Chr.12: 49639291-49639292 on Build GRCh38
rs887625724 Chr.12: 49639328-49639328 on Build GRCh38
rs1040278375 Chr.12: 49638962-49638962 on Build GRCh38
rs931921344 Chr.12: 49639308-49639308 on Build GRCh38
rs746864403 Chr.12: 49638922-49638922 on Build GRCh38
rs1048953948 Chr.12: 49639322-49639322 on Build GRCh38
rs1043960370 Chr.12: 49639357-49639357 on Build GRCh38
rs946150089 Chr.12: 49638878-49638878 on Build GRCh38
rs887606331 Chr.12: 49639024-49639024 on Build GRCh38
rs1045167373 Chr.12: 49639233-49639233 on Build GRCh38
rs757758624 Chr.12: 49639361-49639361 on Build GRCh38
rs1004690924 Chr.12: 49639343-49639343 on Build GRCh38
rs953243461 Chr.12: 49638892-49638892 on Build GRCh38
rs770709129 Chr.12: 49639043-49639043 on Build GRCh38
rs892651172 Chr.12: 49639336-49639336 on Build GRCh38
rs2124478 Chr.12: 49639151-49639151 on Build GRCh38
rs772044305 Chr.12: 49638872-49638872 on Build GRCh38

View Full Product Details