Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 89008792-89009390 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GBP3
Entrez Gene ID
2635
Gene Name
guanylate binding protein 3
Gene Aliases
-
UniGene
Hs.720167
Gene Chromosome Location
Chr.1: 89006677-89022880 on Build GRCh38
Transcripts
NM_001319179.1 NM_001319180.1 NM_001319181.1 NM_018284.2 NR_135007.1 XM_005270743.2 XM_006710574.2 XM_011541233.1 XM_011541234.1 XM_017001013.1 XM_017001014.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
400401 c.1523A>G p.Q508R Chr.1: 89009083-89009083 on Build GRCh38
311406 c.1491A>T p.A497A Chr.1: 89009115-89009115 on Build GRCh38
3751431 c.1471T>C p.C491R Chr.1: 89009135-89009135 on Build GRCh38
7078544 c.1578G>A p.V526V Chr.1: 89009028-89009028 on Build GRCh38
5498733 c.1518A>G p.E506E Chr.1: 89009088-89009088 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747796561 Chr.1: 89009020-89009020 on Build GRCh38
rs531689563 Chr.1: 89008846-89008846 on Build GRCh38
rs576596322 Chr.1: 89009227-89009227 on Build GRCh38
rs553573143 Chr.1: 89008984-89008984 on Build GRCh38
rs751818419 Chr.1: 89008963-89008963 on Build GRCh38
rs768362780 Chr.1: 89008894-89008894 on Build GRCh38
rs781574139 Chr.1: 89008975-89008975 on Build GRCh38
rs777631196 Chr.1: 89009301-89009301 on Build GRCh38
rs745829250 Chr.1: 89009186-89009186 on Build GRCh38
rs866354434 Chr.1: 89009036-89009036 on Build GRCh38
rs202154907 Chr.1: 89008973-89008973 on Build GRCh38
rs752297910 Chr.1: 89009215-89009215 on Build GRCh38
rs550723753 Chr.1: 89009096-89009096 on Build GRCh38
rs996777882 Chr.1: 89009230-89009230 on Build GRCh38
rs1004458985 Chr.1: 89009294-89009294 on Build GRCh38
rs144631115 Chr.1: 89009233-89009233 on Build GRCh38
rs993242183 Chr.1: 89009268-89009268 on Build GRCh38
rs748892311 Chr.1: 89008875-89008875 on Build GRCh38
rs939153894 Chr.1: 89008839-89008839 on Build GRCh38
rs138557169 Chr.1: 89009336-89009336 on Build GRCh38
rs867492736 Chr.1: 89008991-89008991 on Build GRCh38
rs867572188 Chr.1: 89009194-89009194 on Build GRCh38
rs781225133 Chr.1: 89009161-89009161 on Build GRCh38
rs201802879 Chr.1: 89009163-89009163 on Build GRCh38
rs754480872 Chr.1: 89009144-89009144 on Build GRCh38
rs755539053 Chr.1: 89009104-89009104 on Build GRCh38
rs374254544 Chr.1: 89008920-89008920 on Build GRCh38
rs140794197 Chr.1: 89008866-89008866 on Build GRCh38
rs773962933 Chr.1: 89008897-89008897 on Build GRCh38
rs774363421 Chr.1: 89009347-89009347 on Build GRCh38
rs371275732 Chr.1: 89009152-89009152 on Build GRCh38
rs367916921 Chr.1: 89009206-89009206 on Build GRCh38
rs183211655 Chr.1: 89008919-89008919 on Build GRCh38
rs761959766 Chr.1: 89009356-89009356 on Build GRCh38
rs768283267 Chr.1: 89008938-89008938 on Build GRCh38
rs746794145 Chr.1: 89008914-89008914 on Build GRCh38
rs148387405 Chr.1: 89009004-89009004 on Build GRCh38
rs1042219166 Chr.1: 89009079-89009079 on Build GRCh38
rs768144664 Chr.1: 89009014-89009014 on Build GRCh38
rs760746460 Chr.1: 89008922-89008922 on Build GRCh38
rs769879227 Chr.1: 89009355-89009356 on Build GRCh38
rs369497629 Chr.1: 89009133-89009133 on Build GRCh38
rs565008059 Chr.1: 89008873-89008873 on Build GRCh38
rs200179940 Chr.1: 89009055-89009055 on Build GRCh38
rs781677372 Chr.1: 89008863-89008863 on Build GRCh38
rs981778901 Chr.1: 89009341-89009341 on Build GRCh38
rs140601989 Chr.1: 89009040-89009040 on Build GRCh38
rs776657841 Chr.1: 89009184-89009184 on Build GRCh38
rs368459851 Chr.1: 89009131-89009131 on Build GRCh38
rs767866856 Chr.1: 89009111-89009111 on Build GRCh38
rs750794401 Chr.1: 89009122-89009122 on Build GRCh38
rs779750411 Chr.1: 89008880-89008881 on Build GRCh38
rs767743790 Chr.1: 89009360-89009360 on Build GRCh38
rs773197183 Chr.1: 89009043-89009043 on Build GRCh38
rs561116140 Chr.1: 89008818-89008818 on Build GRCh38
rs893184631 Chr.1: 89009009-89009009 on Build GRCh38
rs1010872045 Chr.1: 89008872-89008872 on Build GRCh38
rs891336747 Chr.1: 89009275-89009275 on Build GRCh38
rs747825921 Chr.1: 89009159-89009159 on Build GRCh38
rs763018214 Chr.1: 89008961-89008961 on Build GRCh38
rs765392892 Chr.1: 89008928-89008928 on Build GRCh38
rs748015277 Chr.1: 89009354-89009355 on Build GRCh38
rs369942449 Chr.1: 89009080-89009080 on Build GRCh38
rs766742474 Chr.1: 89008962-89008962 on Build GRCh38
rs763112575 Chr.1: 89009078-89009078 on Build GRCh38
rs771914848 Chr.1: 89009166-89009166 on Build GRCh38
rs17433780 Chr.1: 89009135-89009135 on Build GRCh38
rs762068363 Chr.1: 89009105-89009105 on Build GRCh38
rs764179425 Chr.1: 89009085-89009085 on Build GRCh38
rs1043742344 Chr.1: 89008904-89008904 on Build GRCh38
rs928829820 Chr.1: 89009363-89009363 on Build GRCh38
rs768756939 Chr.1: 89009189-89009189 on Build GRCh38
rs775374518 Chr.1: 89009076-89009076 on Build GRCh38
rs777643488 Chr.1: 89009170-89009170 on Build GRCh38
rs771971966 Chr.1: 89009021-89009021 on Build GRCh38
rs1007782985 Chr.1: 89009205-89009205 on Build GRCh38
rs532483814 Chr.1: 89008915-89008915 on Build GRCh38
rs751820644 Chr.1: 89009008-89009008 on Build GRCh38
rs777994567 Chr.1: 89008852-89008852 on Build GRCh38
rs778659868 Chr.1: 89009015-89009015 on Build GRCh38
rs1004748482 Chr.1: 89008932-89008932 on Build GRCh38
rs752891472 Chr.1: 89008931-89008931 on Build GRCh38
rs200954051 Chr.1: 89009003-89009003 on Build GRCh38
rs773520369 Chr.1: 89009366-89009366 on Build GRCh38
rs747404066 Chr.1: 89008857-89008857 on Build GRCh38
rs757642584 Chr.1: 89008861-89008861 on Build GRCh38
rs775041863 Chr.1: 89008871-89008871 on Build GRCh38
rs770939504 Chr.1: 89009177-89009177 on Build GRCh38
rs750919160 Chr.1: 89008981-89008981 on Build GRCh38
rs746814079 Chr.1: 89009175-89009175 on Build GRCh38
rs769259607 Chr.1: 89008867-89008867 on Build GRCh38
rs770996703 Chr.1: 89009061-89009061 on Build GRCh38

View Full Product Details