Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 160998650-160999168 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
F11R
Entrez Gene ID
50848
Gene Name
F11 receptor
Gene Aliases
CD321, JAM, JAM1, JAMA, JCAM, KAT, PAM-1
UniGene
Hs.517293
Gene Chromosome Location
Chr.1: 160995211-161021343 on Build GRCh38
Transcripts
NM_016946.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1193520 c.857G>A p.R286Q Chr.1: 160999050-160999050 on Build GRCh38
7366870 c.869G>A p.R290Q Chr.1: 160999050-160999050 on Build GRCh38
4599585 c.832G>A p.V278M Chr.1: 160999075-160999075 on Build GRCh38
6657663 c.829A>G p.T277A Chr.1: 160999090-160999090 on Build GRCh38
5641417 c.900A>T p.*300C Chr.1: 160998871-160998871 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764558127 Chr.1: 160999029-160999029 on Build GRCh38
rs929342606 Chr.1: 160998800-160998800 on Build GRCh38
rs148045492 Chr.1: 160998926-160998926 on Build GRCh38
rs141740827 Chr.1: 160999010-160999010 on Build GRCh38
rs749191205 Chr.1: 160999132-160999132 on Build GRCh38
rs953292393 Chr.1: 160998728-160998728 on Build GRCh38
rs149044377 Chr.1: 160998886-160998886 on Build GRCh38
rs746832900 Chr.1: 160998956-160998956 on Build GRCh38
rs900693297 Chr.1: 160999049-160999049 on Build GRCh38
rs762669299 Chr.1: 160998883-160998883 on Build GRCh38
rs145312674 Chr.1: 160999083-160999083 on Build GRCh38
rs946449826 Chr.1: 160998713-160998713 on Build GRCh38
rs754154040 Chr.1: 160999051-160999051 on Build GRCh38
rs770473950 Chr.1: 160999133-160999133 on Build GRCh38
rs775432340 Chr.1: 160998851-160998851 on Build GRCh38
rs745871866 Chr.1: 160998946-160998946 on Build GRCh38
rs896472476 Chr.1: 160998776-160998776 on Build GRCh38
rs772181583 Chr.1: 160998831-160998831 on Build GRCh38
rs368329169 Chr.1: 160999027-160999027 on Build GRCh38
rs945515929 Chr.1: 160998881-160998881 on Build GRCh38
rs755922833 Chr.1: 160998966-160998966 on Build GRCh38
rs970510169 Chr.1: 160998753-160998753 on Build GRCh38
rs747284482 Chr.1: 160999086-160999086 on Build GRCh38
rs781044888 Chr.1: 160999108-160999108 on Build GRCh38
rs970464508 Chr.1: 160999028-160999028 on Build GRCh38
rs751710003 Chr.1: 160999050-160999050 on Build GRCh38
rs926264385 Chr.1: 160999080-160999080 on Build GRCh38
rs1016571312 Chr.1: 160998975-160998975 on Build GRCh38
rs979223458 Chr.1: 160998739-160998739 on Build GRCh38
rs746744804 Chr.1: 160998852-160998852 on Build GRCh38
rs765608207 Chr.1: 160999067-160999067 on Build GRCh38
rs764034212 Chr.1: 160998698-160998700 on Build GRCh38
rs886939003 Chr.1: 160999116-160999127 on Build GRCh38
rs926460468 Chr.1: 160998743-160998743 on Build GRCh38
rs772591374 Chr.1: 160998877-160998877 on Build GRCh38
rs764406987 Chr.1: 160998923-160998923 on Build GRCh38
rs374390184 Chr.1: 160998830-160998830 on Build GRCh38
rs779119815 Chr.1: 160998943-160998943 on Build GRCh38
rs780375383 Chr.1: 160998955-160998955 on Build GRCh38
rs559524199 Chr.1: 160998698-160998698 on Build GRCh38
rs757357648 Chr.1: 160998932-160998933 on Build GRCh38
rs375351079 Chr.1: 160998993-160998993 on Build GRCh38
rs528185113 Chr.1: 160998699-160998699 on Build GRCh38
rs762359389 Chr.1: 160999116-160999116 on Build GRCh38
rs776268416 Chr.1: 160998997-160998997 on Build GRCh38
rs756427805 Chr.1: 160998916-160998916 on Build GRCh38
rs755247076 Chr.1: 160999094-160999094 on Build GRCh38
rs146651683 Chr.1: 160998887-160998887 on Build GRCh38
rs761046064 Chr.1: 160998908-160998908 on Build GRCh38
rs937918399 Chr.1: 160998747-160998747 on Build GRCh38
rs41269993 Chr.1: 160998819-160998819 on Build GRCh38
rs538400987 Chr.1: 160999085-160999085 on Build GRCh38
rs370457243 Chr.1: 160998949-160998949 on Build GRCh38
rs373180635 Chr.1: 160998750-160998750 on Build GRCh38
rs563047861 Chr.1: 160998749-160998749 on Build GRCh38
rs776670650 Chr.1: 160998862-160998862 on Build GRCh38
rs998114328 Chr.1: 160998705-160998705 on Build GRCh38
rs913690188 Chr.1: 160998708-160998708 on Build GRCh38
rs777362378 Chr.1: 160999119-160999119 on Build GRCh38
rs201161407 Chr.1: 160998890-160998890 on Build GRCh38
rs191764210 Chr.1: 160999115-160999115 on Build GRCh38
rs757695512 Chr.1: 160998940-160998940 on Build GRCh38
rs370163633 Chr.1: 160998855-160998855 on Build GRCh38
rs1007412792 Chr.1: 160998934-160998934 on Build GRCh38
rs111999502 Chr.1: 160998706-160998706 on Build GRCh38
rs201421335 Chr.1: 160998863-160998863 on Build GRCh38
rs558582675 Chr.1: 160999038-160999038 on Build GRCh38
rs1016083307 Chr.1: 160998740-160998740 on Build GRCh38
rs757789990 Chr.1: 160999054-160999054 on Build GRCh38
rs955067746 Chr.1: 160999096-160999096 on Build GRCh38
rs12047706 Chr.1: 160999092-160999092 on Build GRCh38
rs998262701 Chr.1: 160998921-160998921 on Build GRCh38
rs777529755 Chr.1: 160999129-160999129 on Build GRCh38
rs762318782 Chr.1: 160999025-160999025 on Build GRCh38
rs772790343 Chr.1: 160999013-160999013 on Build GRCh38
rs772185387 Chr.1: 160998758-160998758 on Build GRCh38
rs186871585 Chr.1: 160999009-160999009 on Build GRCh38
rs750540056 Chr.1: 160999069-160999069 on Build GRCh38
rs375303349 Chr.1: 160999033-160999033 on Build GRCh38
rs550614644 Chr.1: 160998874-160998874 on Build GRCh38

View Full Product Details