Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 32607247-32607839 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C21orf59
Entrez Gene ID
56683
Gene Name
chromosome 21 open reading frame 59
Gene Aliases
C21orf48, CILD26, FBB18, Kur
UniGene
Hs.5811
Gene Chromosome Location
Chr.21: 32601674-32612608 on Build GRCh38
Transcripts
NM_021254.2 NR_036552.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6574335 c.329A>C p.Q110P Chr.21: 32607695-32607695 on Build GRCh38
4872823 c.140-1G>T p.? Chr.21: 32607717-32607717 on Build GRCh38
1030204 c.308-1G>T p.? Chr.21: 32607717-32607717 on Build GRCh38
3799862 c.202T>C p.S68P Chr.21: 32607654-32607654 on Build GRCh38
5902153 c.375G>A p.K125K Chr.21: 32607649-32607649 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs934358184 Chr.21: 32607405-32607405 on Build GRCh38
rs749373260 Chr.21: 32607653-32607653 on Build GRCh38
rs750102379 Chr.21: 32607739-32607739 on Build GRCh38
rs930493406 Chr.21: 32607298-32607299 on Build GRCh38
rs113525692 Chr.21: 32607321-32607321 on Build GRCh38
rs993817357 Chr.21: 32607282-32607282 on Build GRCh38
rs113142611 Chr.21: 32607533-32607533 on Build GRCh38
rs753158067 Chr.21: 32607627-32607627 on Build GRCh38
rs891347799 Chr.21: 32607450-32607450 on Build GRCh38
rs989689505 Chr.21: 32607690-32607690 on Build GRCh38
rs373254863 Chr.21: 32607753-32607753 on Build GRCh38
rs869200061 Chr.21: 32607608-32607610 on Build GRCh38
rs952003447 Chr.21: 32607531-32607531 on Build GRCh38
rs564768641 Chr.21: 32607782-32607782 on Build GRCh38
rs897199908 Chr.21: 32607790-32607793 on Build GRCh38
rs570814268 Chr.21: 32607536-32607536 on Build GRCh38
rs966794211 Chr.21: 32607751-32607751 on Build GRCh38
rs552387466 Chr.21: 32607477-32607477 on Build GRCh38
rs1015508194 Chr.21: 32607460-32607460 on Build GRCh38
rs1038066225 Chr.21: 32607497-32607497 on Build GRCh38
rs971300791 Chr.21: 32607567-32607567 on Build GRCh38
rs1013169824 Chr.21: 32607809-32607809 on Build GRCh38
rs184109745 Chr.21: 32607541-32607541 on Build GRCh38
rs915355794 Chr.21: 32607370-32607370 on Build GRCh38
rs554012729 Chr.21: 32607542-32607542 on Build GRCh38
rs112782203 Chr.21: 32607615-32607615 on Build GRCh38
rs750641633 Chr.21: 32607657-32607664 on Build GRCh38
rs1054202845 Chr.21: 32607413-32607413 on Build GRCh38
rs753319975 Chr.21: 32607607-32607607 on Build GRCh38
rs959463259 Chr.21: 32607804-32607804 on Build GRCh38
rs796704338 Chr.21: 32607611-32607612 on Build GRCh38
rs989575118 Chr.21: 32607331-32607331 on Build GRCh38
rs75400278 Chr.21: 32607593-32607593 on Build GRCh38
rs1015116939 Chr.21: 32607574-32607574 on Build GRCh38
rs767677910 Chr.21: 32607633-32607634 on Build GRCh38
rs746046767 Chr.21: 32607683-32607683 on Build GRCh38
rs572291042 Chr.21: 32607550-32607550 on Build GRCh38
rs779245134 Chr.21: 32607677-32607677 on Build GRCh38
rs560584894 Chr.21: 32607595-32607596 on Build GRCh38
rs375465953 Chr.21: 32607644-32607644 on Build GRCh38
rs546042812 Chr.21: 32607451-32607451 on Build GRCh38
rs577484040 Chr.21: 32607592-32607592 on Build GRCh38
rs775624139 Chr.21: 32607709-32607709 on Build GRCh38
rs844994 Chr.21: 32607423-32607423 on Build GRCh38
rs569780918 Chr.21: 32607479-32607479 on Build GRCh38
rs114583237 Chr.21: 32607316-32607316 on Build GRCh38
rs771106055 Chr.21: 32607671-32607671 on Build GRCh38
rs530210896 Chr.21: 32607454-32607454 on Build GRCh38
rs756639568 Chr.21: 32607764-32607764 on Build GRCh38
rs926813133 Chr.21: 32607318-32607318 on Build GRCh38
rs781446380 Chr.21: 32607361-32607361 on Build GRCh38
rs768630554 Chr.21: 32607724-32607724 on Build GRCh38
rs769161712 Chr.21: 32607611-32607612 on Build GRCh38
rs560934494 Chr.21: 32607315-32607315 on Build GRCh38
rs902941603 Chr.21: 32607404-32607404 on Build GRCh38
rs760609015 Chr.21: 32607603-32607603 on Build GRCh38
rs971037424 Chr.21: 32607293-32607293 on Build GRCh38
rs527702535 Chr.21: 32607610-32607611 on Build GRCh38
rs112325415 Chr.21: 32607390-32607390 on Build GRCh38
rs949770319 Chr.21: 32607369-32607369 on Build GRCh38
rs543502175 Chr.21: 32607777-32607777 on Build GRCh38
rs1027065729 Chr.21: 32607505-32607505 on Build GRCh38
rs376482940 Chr.21: 32607741-32607741 on Build GRCh38
rs112029514 Chr.21: 32607554-32607554 on Build GRCh38
rs756577924 Chr.21: 32607634-32607634 on Build GRCh38
rs934312313 Chr.21: 32607647-32607647 on Build GRCh38
rs1005371357 Chr.21: 32607485-32607485 on Build GRCh38
rs537048874 Chr.21: 32607512-32607512 on Build GRCh38
rs893328389 Chr.21: 32607414-32607414 on Build GRCh38
rs763521443 Chr.21: 32607606-32607606 on Build GRCh38
rs200301099 Chr.21: 32607703-32607703 on Build GRCh38
rs761917396 Chr.21: 32607630-32607631 on Build GRCh38
rs771080449 Chr.21: 32607291-32607291 on Build GRCh38
rs924308788 Chr.21: 32607643-32607643 on Build GRCh38
rs961447166 Chr.21: 32607462-32607462 on Build GRCh38
rs369481919 Chr.21: 32607696-32607696 on Build GRCh38
rs962247913 Chr.21: 32607579-32607579 on Build GRCh38
rs78862894 Chr.21: 32607595-32607595 on Build GRCh38
rs140374402 Chr.21: 32607732-32607732 on Build GRCh38
rs191821279 Chr.21: 32607431-32607431 on Build GRCh38
rs973660290 Chr.21: 32607580-32607580 on Build GRCh38
rs753103171 Chr.21: 32607760-32607760 on Build GRCh38
rs11538144 Chr.21: 32607702-32607702 on Build GRCh38
rs755371024 Chr.21: 32607617-32607617 on Build GRCh38
rs370610216 Chr.21: 32607758-32607758 on Build GRCh38
rs543008562 Chr.21: 32607320-32607320 on Build GRCh38
rs761435759 Chr.21: 32607614-32607614 on Build GRCh38
rs186557521 Chr.21: 32607626-32607626 on Build GRCh38
rs759468666 Chr.21: 32607731-32607732 on Build GRCh38
rs995446328 Chr.21: 32607791-32607791 on Build GRCh38
rs971346760 Chr.21: 32607558-32607558 on Build GRCh38
rs889640179 Chr.21: 32607464-32607464 on Build GRCh38
rs879187168 Chr.21: 32607595-32607595 on Build GRCh38
rs200153059 Chr.21: 32607742-32607742 on Build GRCh38
rs1027500323 Chr.21: 32607802-32607803 on Build GRCh38
rs370314274 Chr.21: 32607678-32607678 on Build GRCh38
rs776057146 Chr.21: 32607726-32607726 on Build GRCh38
rs945359351 Chr.21: 32607377-32607377 on Build GRCh38

View Full Product Details