Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 46496849-46497206 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DIP2A
Entrez Gene ID
23181
Gene Name
disco interacting protein 2 homolog A
Gene Aliases
C21orf106, DIP2
UniGene
Hs.189585
Gene Chromosome Location
Chr.21: 46458942-46570900 on Build GRCh38
Transcripts
NM_001146115.1 NM_001146116.1 NM_015151.3 NM_206889.2 NM_206890.2 NM_206891.2 XM_011529492.2 XM_011529501.1 XM_011529502.1 XM_017028291.1 XM_017028292.1 XM_017028293.1 XM_017028294.1 XM_017028295.1 XM_017028296.1 XM_017028297.1 XM_017028298.1 XM_017028299.1 XM_017028300.1 XM_017028301.1 XM_017028302.1 XM_017028303.1 XM_017028304.1 XM_017028305.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3551793 c.377C>T p.S126L Chr.21: 46497081-46497081 on Build GRCh38
1534844 c.108C>T p.A36A Chr.21: 46497004-46497004 on Build GRCh38
5810858 c.284-2A>G p.? Chr.21: 46496986-46496986 on Build GRCh38
3972838 c.378G>A p.S126S Chr.21: 46497082-46497082 on Build GRCh38
3972839 c.378G>A p.S126S Chr.21: 46497082-46497082 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs754034983 Chr.21: 46497113-46497113 on Build GRCh38
rs563754453 Chr.21: 46496990-46496990 on Build GRCh38
rs896513237 Chr.21: 46496880-46496880 on Build GRCh38
rs746861048 Chr.21: 46497003-46497003 on Build GRCh38
rs762230072 Chr.21: 46497085-46497085 on Build GRCh38
rs57652511 Chr.21: 46497035-46497036 on Build GRCh38
rs11315869 Chr.21: 46497135-46497135 on Build GRCh38
rs545767339 Chr.21: 46496975-46496975 on Build GRCh38
rs376553079 Chr.21: 46496999-46496999 on Build GRCh38
rs766400850 Chr.21: 46497106-46497106 on Build GRCh38
rs530313296 Chr.21: 46497182-46497182 on Build GRCh38
rs745352405 Chr.21: 46497006-46497006 on Build GRCh38
rs779014005 Chr.21: 46497171-46497171 on Build GRCh38
rs781230100 Chr.21: 46497151-46497151 on Build GRCh38
rs572277026 Chr.21: 46496918-46496918 on Build GRCh38
rs764402155 Chr.21: 46496945-46496945 on Build GRCh38
rs1014567101 Chr.21: 46496895-46496895 on Build GRCh38
rs755142608 Chr.21: 46497119-46497119 on Build GRCh38
rs868356218 Chr.21: 46497066-46497066 on Build GRCh38
rs752034147 Chr.21: 46496952-46496952 on Build GRCh38
rs957542247 Chr.21: 46496911-46496911 on Build GRCh38
rs745836051 Chr.21: 46497156-46497156 on Build GRCh38
rs751978836 Chr.21: 46496950-46496950 on Build GRCh38
rs756225398 Chr.21: 46496970-46496970 on Build GRCh38
rs369485432 Chr.21: 46497055-46497055 on Build GRCh38
rs993298684 Chr.21: 46496924-46496924 on Build GRCh38
rs561488078 Chr.21: 46497122-46497122 on Build GRCh38
rs373442777 Chr.21: 46497075-46497075 on Build GRCh38
rs757465103 Chr.21: 46497093-46497093 on Build GRCh38
rs951904338 Chr.21: 46496953-46496953 on Build GRCh38
rs769213497 Chr.21: 46497041-46497041 on Build GRCh38
rs976329649 Chr.21: 46497051-46497051 on Build GRCh38
rs397867243 Chr.21: 46497142-46497142 on Build GRCh38
rs762512866 Chr.21: 46497077-46497077 on Build GRCh38
rs372158611 Chr.21: 46496966-46496966 on Build GRCh38
rs767979325 Chr.21: 46497094-46497094 on Build GRCh38
rs781406550 Chr.21: 46497131-46497132 on Build GRCh38
rs1045662464 Chr.21: 46496912-46496912 on Build GRCh38
rs181610004 Chr.21: 46497082-46497082 on Build GRCh38
rs760696574 Chr.21: 46497100-46497100 on Build GRCh38
rs750975896 Chr.21: 46497096-46497096 on Build GRCh38
rs749494495 Chr.21: 46496981-46496981 on Build GRCh38
rs777976003 Chr.21: 46496945-46496946 on Build GRCh38
rs1027320281 Chr.21: 46496935-46496935 on Build GRCh38
rs757770299 Chr.21: 46496961-46496961 on Build GRCh38
rs750494625 Chr.21: 46496968-46496968 on Build GRCh38
rs915766114 Chr.21: 46496937-46496937 on Build GRCh38
rs757169725 Chr.21: 46497140-46497140 on Build GRCh38
rs1008367396 Chr.21: 46497176-46497176 on Build GRCh38
rs375904809 Chr.21: 46497081-46497081 on Build GRCh38
rs775173135 Chr.21: 46497052-46497052 on Build GRCh38
rs150329133 Chr.21: 46496911-46496911 on Build GRCh38
rs139510335 Chr.21: 46497004-46497004 on Build GRCh38
rs751370781 Chr.21: 46497127-46497127 on Build GRCh38

View Full Product Details