Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 11446137-11446649 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
E2F6
Entrez Gene ID
1876
Gene Name
E2F transcription factor 6
Gene Aliases
E2F-6
UniGene
Hs.603093
Gene Chromosome Location
Chr.2: 11444375-11466177 on Build GRCh38
Transcripts
NM_001278275.1 NM_001278276.1 NM_001278277.1 NM_001278278.1 NM_198256.3 NM_212540.2 NR_103490.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1668711 c.823G>T p.E275* Chr.2: 11446500-11446500 on Build GRCh38
4679974 c.829C>A p.L277I Chr.2: 11446494-11446494 on Build GRCh38
4401012 c.806A>C p.N269T Chr.2: 11446517-11446517 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1051083345 Chr.2: 11446333-11446333 on Build GRCh38
rs771744339 Chr.2: 11446493-11446493 on Build GRCh38
rs538288887 Chr.2: 11446546-11446559 on Build GRCh38
rs111978007 Chr.2: 11446572-11446572 on Build GRCh38
rs1012753066 Chr.2: 11446167-11446167 on Build GRCh38
rs112341910 Chr.2: 11446430-11446430 on Build GRCh38
rs781333322 Chr.2: 11446466-11446466 on Build GRCh38
rs377480770 Chr.2: 11446537-11446537 on Build GRCh38
rs764101094 Chr.2: 11446435-11446435 on Build GRCh38
rs1012750737 Chr.2: 11446411-11446411 on Build GRCh38
rs116381336 Chr.2: 11446553-11446553 on Build GRCh38
rs775332275 Chr.2: 11446500-11446500 on Build GRCh38
rs760605766 Chr.2: 11446433-11446433 on Build GRCh38
rs995837854 Chr.2: 11446610-11446610 on Build GRCh38
rs560959422 Chr.2: 11446449-11446449 on Build GRCh38
rs753044171 Chr.2: 11446474-11446474 on Build GRCh38
rs950431119 Chr.2: 11446187-11446187 on Build GRCh38
rs759590056 Chr.2: 11446538-11446538 on Build GRCh38
rs566983632 Chr.2: 11446567-11446567 on Build GRCh38
rs778492314 Chr.2: 11446491-11446491 on Build GRCh38
rs539823662 Chr.2: 11446562-11446562 on Build GRCh38
rs899125750 Chr.2: 11446603-11446603 on Build GRCh38
rs145760514 Chr.2: 11446580-11446580 on Build GRCh38
rs751673273 Chr.2: 11446452-11446452 on Build GRCh38
rs770292671 Chr.2: 11446489-11446489 on Build GRCh38
rs538487274 Chr.2: 11446199-11446199 on Build GRCh38
rs752952391 Chr.2: 11446548-11446548 on Build GRCh38
rs921516713 Chr.2: 11446290-11446290 on Build GRCh38
rs532617402 Chr.2: 11446185-11446185 on Build GRCh38
rs767753132 Chr.2: 11446540-11446540 on Build GRCh38
rs753867441 Chr.2: 11446437-11446437 on Build GRCh38
rs976107301 Chr.2: 11446266-11446266 on Build GRCh38
rs575306694 Chr.2: 11446398-11446398 on Build GRCh38
rs373248052 Chr.2: 11446530-11446530 on Build GRCh38
rs778217325 Chr.2: 11446483-11446483 on Build GRCh38
rs934716148 Chr.2: 11446352-11446352 on Build GRCh38
rs971510549 Chr.2: 11446175-11446175 on Build GRCh38
rs892932874 Chr.2: 11446404-11446404 on Build GRCh38
rs745438972 Chr.2: 11446492-11446492 on Build GRCh38
rs760515748 Chr.2: 11446514-11446514 on Build GRCh38
rs771853817 Chr.2: 11446205-11446205 on Build GRCh38
rs577603226 Chr.2: 11446197-11446197 on Build GRCh38
rs778127463 Chr.2: 11446557-11446557 on Build GRCh38
rs556433219 Chr.2: 11446253-11446253 on Build GRCh38
rs776660326 Chr.2: 11446526-11446526 on Build GRCh38
rs1054503122 Chr.2: 11446604-11446604 on Build GRCh38
rs761853250 Chr.2: 11446528-11446528 on Build GRCh38
rs564874035 Chr.2: 11446565-11446565 on Build GRCh38
rs745362159 Chr.2: 11446566-11446566 on Build GRCh38
rs950463205 Chr.2: 11446488-11446488 on Build GRCh38
rs754945668 Chr.2: 11446475-11446475 on Build GRCh38
rs903294207 Chr.2: 11446351-11446351 on Build GRCh38
rs1022632282 Chr.2: 11446172-11446172 on Build GRCh38
rs867318494 Chr.2: 11446298-11446298 on Build GRCh38
rs569484319 Chr.2: 11446418-11446418 on Build GRCh38
rs757781629 Chr.2: 11446564-11446564 on Build GRCh38
rs552714154 Chr.2: 11446357-11446357 on Build GRCh38
rs755231264 Chr.2: 11446459-11446459 on Build GRCh38
rs981124267 Chr.2: 11446176-11446176 on Build GRCh38
rs1054225226 Chr.2: 11446384-11446384 on Build GRCh38
rs747284605 Chr.2: 11446450-11446451 on Build GRCh38
rs200124287 Chr.2: 11446515-11446515 on Build GRCh38
rs774624009 Chr.2: 11446429-11446429 on Build GRCh38
rs763086890 Chr.2: 11446439-11446439 on Build GRCh38

View Full Product Details