Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 152521660-152522210 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FMNL2
Entrez Gene ID
114793
Gene Name
formin like 2
Gene Aliases
FHOD2
UniGene
Hs.654630
Gene Chromosome Location
Chr.2: 152335170-152649834 on Build GRCh38
Transcripts
NM_052905.3 XM_005246263.2 XM_005246265.3 XM_011510530.1 XM_011510531.1 XM_011510532.2 XM_011510533.2 XM_011510534.1 XM_011510535.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3769792 c.180A>G p.K60K Chr.2: 152522005-152522005 on Build GRCh38
1207298 c.121G>A p.A41T Chr.2: 152521946-152521946 on Build GRCh38
1400175 c.130C>T p.L44L Chr.2: 152521955-152521955 on Build GRCh38
5501326 c.118-7T>A p.? Chr.2: 152521936-152521936 on Build GRCh38
5993900 c.163T>C p.Y55H Chr.2: 152521988-152521988 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs938711959 Chr.2: 152521711-152521711 on Build GRCh38
rs777140418 Chr.2: 152521963-152521963 on Build GRCh38
rs900125674 Chr.2: 152521790-152521790 on Build GRCh38
rs4664593 Chr.2: 152522138-152522138 on Build GRCh38
rs762086112 Chr.2: 152521969-152521969 on Build GRCh38
rs991520604 Chr.2: 152522104-152522104 on Build GRCh38
rs915892069 Chr.2: 152521700-152521700 on Build GRCh38
rs990705011 Chr.2: 152521931-152521931 on Build GRCh38
rs983272578 Chr.2: 152521886-152521886 on Build GRCh38
rs150034689 Chr.2: 152521756-152521756 on Build GRCh38
rs747865885 Chr.2: 152521910-152521910 on Build GRCh38
rs900177481 Chr.2: 152522112-152522112 on Build GRCh38
rs887009364 Chr.2: 152521795-152521795 on Build GRCh38
rs563749669 Chr.2: 152521772-152521772 on Build GRCh38
rs764493438 Chr.2: 152521961-152521961 on Build GRCh38
rs781459845 Chr.2: 152521686-152521686 on Build GRCh38
rs550360898 Chr.2: 152521771-152521771 on Build GRCh38
rs928665638 Chr.2: 152521706-152521706 on Build GRCh38
rs4664592 Chr.2: 152522060-152522060 on Build GRCh38
rs778388273 Chr.2: 152521923-152521926 on Build GRCh38
rs532477610 Chr.2: 152521853-152521853 on Build GRCh38
rs765339810 Chr.2: 152521895-152521895 on Build GRCh38
rs776128288 Chr.2: 152521953-152521953 on Build GRCh38
rs750447695 Chr.2: 152521983-152521983 on Build GRCh38
rs541739709 Chr.2: 152521733-152521733 on Build GRCh38
rs756061850 Chr.2: 152522010-152522010 on Build GRCh38
rs776807439 Chr.2: 152521802-152521802 on Build GRCh38
rs997458643 Chr.2: 152521793-152521793 on Build GRCh38
rs138926754 Chr.2: 152521948-152521948 on Build GRCh38
rs538029579 Chr.2: 152521819-152521819 on Build GRCh38
rs770578373 Chr.2: 152521946-152521946 on Build GRCh38
rs760932380 Chr.2: 152521960-152521960 on Build GRCh38
rs907389191 Chr.2: 152521729-152521729 on Build GRCh38
rs775199622 Chr.2: 152522137-152522137 on Build GRCh38
rs569781284 Chr.2: 152522111-152522111 on Build GRCh38
rs369900467 Chr.2: 152521995-152521995 on Build GRCh38
rs534742882 Chr.2: 152521902-152521902 on Build GRCh38
rs745363513 Chr.2: 152522067-152522067 on Build GRCh38
rs914709638 Chr.2: 152522097-152522097 on Build GRCh38
rs1028027450 Chr.2: 152521905-152521905 on Build GRCh38
rs754822790 Chr.2: 152521898-152521898 on Build GRCh38
rs934788566 Chr.2: 152521744-152521744 on Build GRCh38
rs957811063 Chr.2: 152521924-152521924 on Build GRCh38
rs777393147 Chr.2: 152521937-152521937 on Build GRCh38
rs4664114 Chr.2: 152521945-152521945 on Build GRCh38
rs757733887 Chr.2: 152521928-152521929 on Build GRCh38
rs772424243 Chr.2: 152521950-152521950 on Build GRCh38
rs567691441 Chr.2: 152521726-152521726 on Build GRCh38
rs755973752 Chr.2: 152521927-152521927 on Build GRCh38
rs779327259 Chr.2: 152522043-152522043 on Build GRCh38
rs939898880 Chr.2: 152522183-152522183 on Build GRCh38
rs529366124 Chr.2: 152521806-152521806 on Build GRCh38
rs1053260616 Chr.2: 152521755-152521755 on Build GRCh38
rs900855676 Chr.2: 152521829-152521829 on Build GRCh38
rs749116025 Chr.2: 152522052-152522052 on Build GRCh38
rs756877128 Chr.2: 152522057-152522057 on Build GRCh38
rs752741344 Chr.2: 152522005-152522005 on Build GRCh38
rs73968056 Chr.2: 152522124-152522124 on Build GRCh38
rs375469230 Chr.2: 152521982-152521982 on Build GRCh38
rs758548648 Chr.2: 152521985-152521985 on Build GRCh38
rs754371548 Chr.2: 152521929-152521929 on Build GRCh38
rs566055964 Chr.2: 152521837-152521837 on Build GRCh38
rs200398180 Chr.2: 152522030-152522030 on Build GRCh38
rs144317405 Chr.2: 152521764-152521764 on Build GRCh38
rs886962976 Chr.2: 152522144-152522144 on Build GRCh38

View Full Product Details