Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 24213819-24214412 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHMP4A
Entrez Gene ID
29082
Gene Name
charged multivesicular body protein 4A
Gene Aliases
C14orf123, CHMP4, CHMP4B, HSPC134, SHAX2, SNF7, SNF7-1, VPS32-1, VPS32A
UniGene
Hs.279761
Gene Chromosome Location
Chr.14: 24209581-24213830 on Build GRCh38
Transcripts
NM_014169.3 NM_001199821.1 NM_001199822.1 NM_138476.3

Gene Symbol
MDP1
Entrez Gene ID
145553
Gene Name
magnesium dependent phosphatase 1
Gene Aliases
FN6PASE, MDP-1
UniGene
Hs.220963
Gene Chromosome Location
Chr.14: 24213937-24216070 on Build GRCh38
Transcripts
NM_014169.3 NM_001199821.1 NM_001199822.1 NM_138476.3

Gene Symbol
NEDD8-MDP1
Entrez Gene ID
100528064
Gene Name
NEDD8-MDP1 readthrough
Gene Aliases
-
UniGene
Hs.220963
Gene Chromosome Location
Chr.14: 24213937-24232370 on Build GRCh38
Transcripts
NM_014169.3 NM_001199821.1 NM_001199822.1 NM_138476.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
954965 c.452G>A p.S151N Chr.14: 24214103-24214103 on Build GRCh38
432946 c.421A>G p.I141V Chr.14: 24214134-24214134 on Build GRCh38
3955948 c.495G>C p.L165F Chr.14: 24214060-24214060 on Build GRCh38
1195872 c.400C>G p.L134V Chr.14: 24214313-24214313 on Build GRCh38
954964 c.504C>T p.S168S Chr.14: 24214051-24214051 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761084532 Chr.14: 24214135-24214135 on Build GRCh38
rs1028162999 Chr.14: 24214023-24214023 on Build GRCh38
rs377711229 Chr.14: 24214336-24214336 on Build GRCh38
rs1055128296 Chr.14: 24213958-24213958 on Build GRCh38
rs772685677 Chr.14: 24214099-24214099 on Build GRCh38
rs747885783 Chr.14: 24214067-24214067 on Build GRCh38
rs2295318 Chr.14: 24213919-24213919 on Build GRCh38
rs765269280 Chr.14: 24214166-24214166 on Build GRCh38
rs1054891410 Chr.14: 24214279-24214279 on Build GRCh38
rs754927249 Chr.14: 24214191-24214191 on Build GRCh38
rs936247414 Chr.14: 24214267-24214267 on Build GRCh38
rs529565317 Chr.14: 24214150-24214150 on Build GRCh38
rs774595458 Chr.14: 24213955-24213955 on Build GRCh38
rs778668678 Chr.14: 24214359-24214359 on Build GRCh38
rs764177509 Chr.14: 24213998-24213998 on Build GRCh38
rs201515371 Chr.14: 24214037-24214037 on Build GRCh38
rs1055161752 Chr.14: 24214308-24214308 on Build GRCh38
rs1010307288 Chr.14: 24213879-24213879 on Build GRCh38
rs762088409 Chr.14: 24214154-24214154 on Build GRCh38
rs764385329 Chr.14: 24214145-24214145 on Build GRCh38
rs754875485 Chr.14: 24214050-24214050 on Build GRCh38
rs79864431 Chr.14: 24213960-24213960 on Build GRCh38
rs745322383 Chr.14: 24214261-24214261 on Build GRCh38
rs761027347 Chr.14: 24213978-24213978 on Build GRCh38
rs757926155 Chr.14: 24214374-24214374 on Build GRCh38
rs766355178 Chr.14: 24214039-24214039 on Build GRCh38
rs753987448 Chr.14: 24214034-24214034 on Build GRCh38
rs777020587 Chr.14: 24214322-24214322 on Build GRCh38
rs945398106 Chr.14: 24213856-24213856 on Build GRCh38
rs3215610 Chr.14: 24214061-24214061 on Build GRCh38
rs1042368792 Chr.14: 24213865-24213865 on Build GRCh38
rs773307111 Chr.14: 24214333-24214333 on Build GRCh38
rs193921062 Chr.14: 24214313-24214313 on Build GRCh38
rs748962147 Chr.14: 24214253-24214253 on Build GRCh38
rs370925626 Chr.14: 24214384-24214384 on Build GRCh38
rs770403232 Chr.14: 24214097-24214097 on Build GRCh38
rs768962135 Chr.14: 24214320-24214320 on Build GRCh38
rs778615525 Chr.14: 24214260-24214260 on Build GRCh38
rs976640966 Chr.14: 24214010-24214010 on Build GRCh38
rs772740993 Chr.14: 24214264-24214264 on Build GRCh38
rs2295317 Chr.14: 24214337-24214337 on Build GRCh38
rs548535729 Chr.14: 24214202-24214202 on Build GRCh38
rs776076407 Chr.14: 24214302-24214302 on Build GRCh38
rs374613616 Chr.14: 24214304-24214304 on Build GRCh38
rs976530388 Chr.14: 24214107-24214107 on Build GRCh38
rs200207999 Chr.14: 24214372-24214372 on Build GRCh38
rs398102305 Chr.14: 24214063-24214063 on Build GRCh38
rs756904046 Chr.14: 24214254-24214254 on Build GRCh38
rs760656021 Chr.14: 24214342-24214342 on Build GRCh38
rs533615873 Chr.14: 24213922-24213922 on Build GRCh38
rs751563297 Chr.14: 24214047-24214047 on Build GRCh38
rs953729557 Chr.14: 24214027-24214027 on Build GRCh38
rs755822554 Chr.14: 24214070-24214070 on Build GRCh38
rs939417997 Chr.14: 24213937-24213937 on Build GRCh38
rs950161353 Chr.14: 24214209-24214209 on Build GRCh38
rs1027893118 Chr.14: 24214052-24214052 on Build GRCh38
rs368781852 Chr.14: 24214072-24214072 on Build GRCh38
rs377678343 Chr.14: 24214382-24214382 on Build GRCh38
rs753383510 Chr.14: 24214168-24214168 on Build GRCh38
rs781010478 Chr.14: 24214056-24214056 on Build GRCh38
rs776019197 Chr.14: 24214112-24214112 on Build GRCh38
rs563719275 Chr.14: 24214356-24214356 on Build GRCh38
rs748842541 Chr.14: 24214082-24214082 on Build GRCh38
rs945273625 Chr.14: 24214219-24214219 on Build GRCh38
rs145254894 Chr.14: 24214098-24214098 on Build GRCh38
rs200453640 Chr.14: 24214063-24214063 on Build GRCh38
rs761987435 Chr.14: 24214323-24214323 on Build GRCh38
rs753646844 Chr.14: 24214355-24214355 on Build GRCh38
rs1010777639 Chr.14: 24214306-24214306 on Build GRCh38
rs572434863 Chr.14: 24213860-24213860 on Build GRCh38
rs761454982 Chr.14: 24214190-24214190 on Build GRCh38
rs2875 Chr.14: 24214017-24214017 on Build GRCh38
rs777608388 Chr.14: 24214251-24214251 on Build GRCh38
rs758770893 Chr.14: 24214279-24214279 on Build GRCh38
rs564523733 Chr.14: 24213921-24213921 on Build GRCh38
rs1044678971 Chr.14: 24214243-24214243 on Build GRCh38
rs769182213 Chr.14: 24214385-24214385 on Build GRCh38
rs780202707 Chr.14: 24214351-24214353 on Build GRCh38
rs994949216 Chr.14: 24214000-24214000 on Build GRCh38
rs371040827 Chr.14: 24214351-24214351 on Build GRCh38
rs750942020 Chr.14: 24214106-24214107 on Build GRCh38
rs113997187 Chr.14: 24214250-24214250 on Build GRCh38

View Full Product Details