Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 44562220-44562815 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EIF3J
Entrez Gene ID
8669
Gene Name
eukaryotic translation initiation factor 3 subunit J
Gene Aliases
EIF3S1, eIF3-alpha, eIF3-p35
UniGene
Hs.404056
Gene Chromosome Location
Chr.15: 44537068-44562803 on Build GRCh38
Transcripts
NM_001284335.1 NM_001284336.1 NM_003758.3 NM_001160227.1 NM_025137.3 XM_006720700.1 XM_017022634.1 XM_017022636.1

Gene Symbol
SPG11
Entrez Gene ID
80208
Gene Name
spastic paraplegia 11 (autosomal recessive)
Gene Aliases
ALS5, CMT2X, KIAA1840
UniGene
Hs.656271
Gene Chromosome Location
Chr.15: 44562696-44663678 on Build GRCh38
Transcripts
NM_001284335.1 NM_001284336.1 NM_003758.3 NM_001160227.1 NM_025137.3 XM_006720700.1 XM_017022634.1 XM_017022636.1

SNP Detail

SNP ID Chromosome Location
rs909374249 Chr.15: 44562316-44562316 on Build GRCh38
rs934135471 Chr.15: 44562442-44562442 on Build GRCh38
rs543388811 Chr.15: 44562755-44562755 on Build GRCh38
rs561296743 Chr.15: 44562573-44562573 on Build GRCh38
rs182043061 Chr.15: 44562482-44562482 on Build GRCh38
rs759682552 Chr.15: 44562708-44562709 on Build GRCh38
rs914167002 Chr.15: 44562751-44562751 on Build GRCh38
rs1036429609 Chr.15: 44562339-44562339 on Build GRCh38
rs886051174 Chr.15: 44562749-44562750 on Build GRCh38
rs1005510619 Chr.15: 44562595-44562595 on Build GRCh38
rs919428815 Chr.15: 44562674-44562674 on Build GRCh38
rs563167899 Chr.15: 44562244-44562244 on Build GRCh38
rs967093427 Chr.15: 44562562-44562562 on Build GRCh38
rs534829835 Chr.15: 44562641-44562641 on Build GRCh38
rs988415393 Chr.15: 44562476-44562477 on Build GRCh38
rs571174731 Chr.15: 44562370-44562371 on Build GRCh38
rs190869234 Chr.15: 44562405-44562405 on Build GRCh38
rs967021161 Chr.15: 44562257-44562257 on Build GRCh38
rs904266667 Chr.15: 44562249-44562249 on Build GRCh38
rs531788566 Chr.15: 44562490-44562490 on Build GRCh38
rs774272508 Chr.15: 44562357-44562357 on Build GRCh38
rs905192530 Chr.15: 44562710-44562710 on Build GRCh38
rs895733754 Chr.15: 44562294-44562295 on Build GRCh38
rs373513475 Chr.15: 44562498-44562498 on Build GRCh38
rs528969919 Chr.15: 44562259-44562259 on Build GRCh38
rs932210863 Chr.15: 44562302-44562302 on Build GRCh38
rs949438142 Chr.15: 44562691-44562691 on Build GRCh38
rs975830151 Chr.15: 44562572-44562572 on Build GRCh38
rs141191303 Chr.15: 44562390-44562391 on Build GRCh38
rs1014038511 Chr.15: 44562574-44562575 on Build GRCh38
rs150223479 Chr.15: 44562774-44562776 on Build GRCh38
rs893012769 Chr.15: 44562749-44562749 on Build GRCh38
rs904275681 Chr.15: 44562554-44562554 on Build GRCh38
rs74935427 Chr.15: 44562291-44562291 on Build GRCh38
rs886051173 Chr.15: 44562722-44562723 on Build GRCh38
rs764147365 Chr.15: 44562457-44562457 on Build GRCh38
rs988302418 Chr.15: 44562749-44562750 on Build GRCh38
rs761486536 Chr.15: 44562456-44562457 on Build GRCh38
rs574359966 Chr.15: 44562276-44562279 on Build GRCh38
rs777573145 Chr.15: 44562643-44562643 on Build GRCh38
rs922767393 Chr.15: 44562425-44562425 on Build GRCh38
rs897885940 Chr.15: 44562349-44562349 on Build GRCh38
rs1025495615 Chr.15: 44562320-44562323 on Build GRCh38
rs1055703962 Chr.15: 44562733-44562733 on Build GRCh38
rs964120742 Chr.15: 44562396-44562396 on Build GRCh38
rs909476778 Chr.15: 44562647-44562647 on Build GRCh38
rs1055937262 Chr.15: 44562566-44562566 on Build GRCh38
rs975715776 Chr.15: 44562277-44562277 on Build GRCh38
rs1001453704 Chr.15: 44562409-44562409 on Build GRCh38
rs535949692 Chr.15: 44562510-44562510 on Build GRCh38
rs1046502069 Chr.15: 44562695-44562695 on Build GRCh38
rs1046342811 Chr.15: 44562402-44562402 on Build GRCh38
rs942231650 Chr.15: 44562324-44562324 on Build GRCh38
rs527880357 Chr.15: 44562327-44562327 on Build GRCh38
rs886991821 Chr.15: 44562594-44562594 on Build GRCh38
rs547643077 Chr.15: 44562399-44562399 on Build GRCh38
rs1036466116 Chr.15: 44562671-44562671 on Build GRCh38
rs945731835 Chr.15: 44562512-44562512 on Build GRCh38
rs937084503 Chr.15: 44562555-44562555 on Build GRCh38
rs571137659 Chr.15: 44562400-44562400 on Build GRCh38
rs555901905 Chr.15: 44562513-44562513 on Build GRCh38
rs577849052 Chr.15: 44562732-44562732 on Build GRCh38
rs770650460 Chr.15: 44562668-44562668 on Build GRCh38
rs1011852470 Chr.15: 44562528-44562528 on Build GRCh38
rs199515547 Chr.15: 44562389-44562390 on Build GRCh38
rs1002641133 Chr.15: 44562717-44562717 on Build GRCh38
rs780275087 Chr.15: 44562329-44562329 on Build GRCh38
rs2288342 Chr.15: 44562587-44562587 on Build GRCh38
rs186069685 Chr.15: 44562729-44562729 on Build GRCh38
rs749067158 Chr.15: 44562656-44562656 on Build GRCh38
rs550125864 Chr.15: 44562407-44562407 on Build GRCh38
rs984078820 Chr.15: 44562389-44562389 on Build GRCh38
rs945676429 Chr.15: 44562243-44562243 on Build GRCh38
rs561940024 Chr.15: 44562304-44562304 on Build GRCh38
rs1051126836 Chr.15: 44562305-44562305 on Build GRCh38
rs760360481 Chr.15: 44562734-44562734 on Build GRCh38
rs978854154 Chr.15: 44562780-44562780 on Build GRCh38
rs1020140298 Chr.15: 44562546-44562546 on Build GRCh38

View Full Product Details