Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 34940154-34940707 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCT6B
Entrez Gene ID
10693
Gene Name
chaperonin containing TCP1 subunit 6B
Gene Aliases
CCT-zeta-2, CCTZ-2, Cctz2, TCP-1-zeta-2, TSA303
UniGene
Hs.73072
Gene Chromosome Location
Chr.17: 34927859-34981221 on Build GRCh38
Transcripts
NM_001193529.2 NM_001193530.1 NM_006584.3 XM_011524203.2 XM_017024023.1 XM_017024024.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4853308 c.892G>C p.D298H Chr.17: 34940615-34940615 on Build GRCh38
977755 c.953G>T p.R318I Chr.17: 34940554-34940554 on Build GRCh38
3421421 c.893A>G p.D298G Chr.17: 34940614-34940614 on Build GRCh38
1679717 c.887G>A p.G296E Chr.17: 34940620-34940620 on Build GRCh38
1159743 c.955_956insA p.R319fs*19 Chr.17: 34940551-34940552 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs371134038 Chr.17: 34940665-34940665 on Build GRCh38
rs888151875 Chr.17: 34940367-34940367 on Build GRCh38
rs557186621 Chr.17: 34940446-34940446 on Build GRCh38
rs762726999 Chr.17: 34940582-34940582 on Build GRCh38
rs546730960 Chr.17: 34940207-34940207 on Build GRCh38
rs1024116560 Chr.17: 34940182-34940182 on Build GRCh38
rs762018220 Chr.17: 34940601-34940601 on Build GRCh38
rs567919309 Chr.17: 34940567-34940567 on Build GRCh38
rs140830057 Chr.17: 34940186-34940189 on Build GRCh38
rs971912940 Chr.17: 34940197-34940197 on Build GRCh38
rs766539535 Chr.17: 34940613-34940613 on Build GRCh38
rs534748628 Chr.17: 34940420-34940420 on Build GRCh38
rs766179313 Chr.17: 34940621-34940621 on Build GRCh38
rs112012333 Chr.17: 34940593-34940593 on Build GRCh38
rs777694336 Chr.17: 34940537-34940537 on Build GRCh38
rs538125196 Chr.17: 34940502-34940502 on Build GRCh38
rs984750339 Chr.17: 34940277-34940277 on Build GRCh38
rs750946380 Chr.17: 34940504-34940504 on Build GRCh38
rs774006298 Chr.17: 34940592-34940592 on Build GRCh38
rs1054018950 Chr.17: 34940412-34940412 on Build GRCh38
rs552331178 Chr.17: 34940373-34940373 on Build GRCh38
rs755296208 Chr.17: 34940517-34940517 on Build GRCh38
rs532505804 Chr.17: 34940345-34940345 on Build GRCh38
rs752992674 Chr.17: 34940645-34940645 on Build GRCh38
rs769085819 Chr.17: 34940212-34940212 on Build GRCh38
rs543681451 Chr.17: 34940290-34940290 on Build GRCh38
rs879523192 Chr.17: 34940416-34940416 on Build GRCh38
rs754025813 Chr.17: 34940619-34940619 on Build GRCh38
rs745837995 Chr.17: 34940575-34940575 on Build GRCh38
rs779206308 Chr.17: 34940644-34940644 on Build GRCh38
rs769488342 Chr.17: 34940355-34940355 on Build GRCh38
rs750316975 Chr.17: 34940373-34940374 on Build GRCh38
rs775021290 Chr.17: 34940586-34940586 on Build GRCh38
rs776152186 Chr.17: 34940258-34940258 on Build GRCh38
rs772892812 Chr.17: 34940389-34940389 on Build GRCh38
rs923449738 Chr.17: 34940313-34940313 on Build GRCh38
rs755118569 Chr.17: 34940624-34940624 on Build GRCh38
rs569130814 Chr.17: 34940376-34940376 on Build GRCh38
rs999823923 Chr.17: 34940397-34940397 on Build GRCh38
rs145620587 Chr.17: 34940678-34940678 on Build GRCh38
rs952023275 Chr.17: 34940252-34940252 on Build GRCh38
rs1033052716 Chr.17: 34940462-34940462 on Build GRCh38
rs1036510040 Chr.17: 34940289-34940289 on Build GRCh38
rs757769698 Chr.17: 34940500-34940500 on Build GRCh38
rs769164222 Chr.17: 34940580-34940580 on Build GRCh38
rs533777774 Chr.17: 34940652-34940652 on Build GRCh38
rs566618329 Chr.17: 34940220-34940220 on Build GRCh38
rs1020143792 Chr.17: 34940419-34940420 on Build GRCh38
rs917473463 Chr.17: 34940219-34940219 on Build GRCh38
rs202071383 Chr.17: 34940515-34940515 on Build GRCh38
rs141918585 Chr.17: 34940552-34940552 on Build GRCh38
rs146039907 Chr.17: 34940588-34940588 on Build GRCh38
rs373539642 Chr.17: 34940200-34940205 on Build GRCh38
rs773292589 Chr.17: 34940653-34940654 on Build GRCh38
rs894413670 Chr.17: 34940414-34940414 on Build GRCh38
rs756110344 Chr.17: 34940511-34940511 on Build GRCh38
rs1040964505 Chr.17: 34940312-34940312 on Build GRCh38
rs781087802 Chr.17: 34940653-34940653 on Build GRCh38
rs1041465005 Chr.17: 34940359-34940359 on Build GRCh38
rs749503156 Chr.17: 34940516-34940516 on Build GRCh38
rs9904027 Chr.17: 34940496-34940496 on Build GRCh38
rs909606362 Chr.17: 34940628-34940628 on Build GRCh38
rs950212609 Chr.17: 34940250-34940250 on Build GRCh38
rs563468247 Chr.17: 34940549-34940551 on Build GRCh38
rs943574723 Chr.17: 34940305-34940305 on Build GRCh38
rs767958063 Chr.17: 34940605-34940605 on Build GRCh38
rs1000321814 Chr.17: 34940441-34940441 on Build GRCh38
rs756697013 Chr.17: 34940610-34940610 on Build GRCh38
rs747565057 Chr.17: 34940188-34940188 on Build GRCh38
rs1012714398 Chr.17: 34940179-34940179 on Build GRCh38
rs745761473 Chr.17: 34940664-34940664 on Build GRCh38

View Full Product Details