Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 113413513-113414069 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C9orf43
Entrez Gene ID
257169
Gene Name
chromosome 9 open reading frame 43
Gene Aliases
-
UniGene
Hs.632691
Gene Chromosome Location
Chr.9: 113410644-113429684 on Build GRCh38
Transcripts
NM_001278629.1 NM_001278630.1 NM_152786.2 XM_011518477.1 XM_011518478.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6876002 c.137C>T p.P46L Chr.9: 113413630-113413630 on Build GRCh38
119663 c.30C>G p.D10E Chr.9: 113413523-113413523 on Build GRCh38
6876003 c.138C>T p.P46P Chr.9: 113413631-113413631 on Build GRCh38
4163236 c.104A>T p.H35L Chr.9: 113413597-113413597 on Build GRCh38
5503515 c.140T>A p.L47Q Chr.9: 113413633-113413633 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs201453096 Chr.9: 113413853-113413853 on Build GRCh38
rs1050885321 Chr.9: 113413582-113413582 on Build GRCh38
rs776976021 Chr.9: 113413896-113413896 on Build GRCh38
rs767236002 Chr.9: 113414025-113414025 on Build GRCh38
rs772199543 Chr.9: 113413610-113413610 on Build GRCh38
rs750486753 Chr.9: 113413839-113413839 on Build GRCh38
rs35171810 Chr.9: 113413566-113413567 on Build GRCh38
rs781625652 Chr.9: 113413927-113413927 on Build GRCh38
rs762207548 Chr.9: 113413897-113413897 on Build GRCh38
rs764364793 Chr.9: 113413917-113413928 on Build GRCh38
rs759438202 Chr.9: 113413912-113413915 on Build GRCh38
rs1007331891 Chr.9: 113413744-113413744 on Build GRCh38
rs776757791 Chr.9: 113413751-113413751 on Build GRCh38
rs538917360 Chr.9: 113413803-113413803 on Build GRCh38
rs965819274 Chr.9: 113413794-113413794 on Build GRCh38
rs200426143 Chr.9: 113413655-113413655 on Build GRCh38
rs924405143 Chr.9: 113413809-113413809 on Build GRCh38
rs370827156 Chr.9: 113413909-113413909 on Build GRCh38
rs761863039 Chr.9: 113413630-113413630 on Build GRCh38
rs138681690 Chr.9: 113413882-113413882 on Build GRCh38
rs140393057 Chr.9: 113413578-113413578 on Build GRCh38
rs770171369 Chr.9: 113413961-113413962 on Build GRCh38
rs548386317 Chr.9: 113413624-113413624 on Build GRCh38
rs757769945 Chr.9: 113413676-113413676 on Build GRCh38
rs1055047826 Chr.9: 113413987-113413987 on Build GRCh38
rs995450793 Chr.9: 113413697-113413697 on Build GRCh38
rs768544169 Chr.9: 113413595-113413595 on Build GRCh38
rs182482838 Chr.9: 113413787-113413787 on Build GRCh38
rs375848085 Chr.9: 113413745-113413745 on Build GRCh38
rs750844682 Chr.9: 113413904-113413904 on Build GRCh38
rs879080563 Chr.9: 113413776-113413776 on Build GRCh38
rs150194451 Chr.9: 113413778-113413778 on Build GRCh38
rs757749488 Chr.9: 113413825-113413825 on Build GRCh38
rs766245523 Chr.9: 113413694-113413697 on Build GRCh38
rs143679220 Chr.9: 113413799-113413799 on Build GRCh38
rs778187103 Chr.9: 113413584-113413584 on Build GRCh38
rs762605848 Chr.9: 113413642-113413642 on Build GRCh38
rs756627091 Chr.9: 113413581-113413581 on Build GRCh38
rs766302755 Chr.9: 113413910-113413910 on Build GRCh38
rs371996976 Chr.9: 113413688-113413688 on Build GRCh38
rs574145967 Chr.9: 113413950-113413950 on Build GRCh38
rs536455879 Chr.9: 113413670-113413670 on Build GRCh38
rs753606766 Chr.9: 113413754-113413754 on Build GRCh38
rs187329744 Chr.9: 113413915-113413915 on Build GRCh38
rs959251537 Chr.9: 113413769-113413769 on Build GRCh38
rs554375741 Chr.9: 113413920-113413920 on Build GRCh38
rs761775195 Chr.9: 113413764-113413764 on Build GRCh38
rs778907937 Chr.9: 113413679-113413679 on Build GRCh38
rs768285318 Chr.9: 113413762-113413762 on Build GRCh38
rs895020231 Chr.9: 113414010-113414010 on Build GRCh38
rs372019948 Chr.9: 113413611-113413611 on Build GRCh38
rs766001704 Chr.9: 113413653-113413653 on Build GRCh38
rs377584622 Chr.9: 113413793-113413793 on Build GRCh38
rs770952202 Chr.9: 113413599-113413599 on Build GRCh38
rs575127978 Chr.9: 113413878-113413878 on Build GRCh38
rs749272442 Chr.9: 113413865-113413865 on Build GRCh38
rs958019313 Chr.9: 113413850-113413850 on Build GRCh38
rs558548983 Chr.9: 113413864-113413864 on Build GRCh38
rs770309069 Chr.9: 113413871-113413871 on Build GRCh38
rs776757925 Chr.9: 113413628-113413628 on Build GRCh38
rs767216305 Chr.9: 113413879-113413879 on Build GRCh38
rs749352310 Chr.9: 113413591-113413591 on Build GRCh38
rs375354869 Chr.9: 113413632-113413632 on Build GRCh38
rs769847240 Chr.9: 113413856-113413856 on Build GRCh38
rs35207865 Chr.9: 113413785-113413785 on Build GRCh38
rs750037166 Chr.9: 113414043-113414043 on Build GRCh38
rs755255430 Chr.9: 113413846-113413846 on Build GRCh38
rs922165268 Chr.9: 113413861-113413861 on Build GRCh38
rs538399898 Chr.9: 113414002-113414002 on Build GRCh38
rs772335399 Chr.9: 113413729-113413729 on Build GRCh38
rs760728739 Chr.9: 113413668-113413669 on Build GRCh38
rs776598879 Chr.9: 113413763-113413763 on Build GRCh38
rs200045945 Chr.9: 113413654-113413654 on Build GRCh38
rs768605987 Chr.9: 113413616-113413616 on Build GRCh38
rs749954086 Chr.9: 113413636-113413636 on Build GRCh38
rs954038039 Chr.9: 113413735-113413735 on Build GRCh38
rs369075240 Chr.9: 113413779-113413779 on Build GRCh38
rs995144676 Chr.9: 113413954-113413954 on Build GRCh38
rs765802524 Chr.9: 113413826-113413826 on Build GRCh38
rs373536157 Chr.9: 113413743-113413743 on Build GRCh38
rs892765376 Chr.9: 113413593-113413593 on Build GRCh38
rs367881038 Chr.9: 113413574-113413574 on Build GRCh38
rs974979462 Chr.9: 113413858-113413858 on Build GRCh38
rs765712502 Chr.9: 113413901-113413901 on Build GRCh38
rs772989087 Chr.9: 113413599-113413599 on Build GRCh38
rs781072419 Chr.9: 113413848-113413848 on Build GRCh38
rs758456205 Chr.9: 113413840-113413840 on Build GRCh38
rs759268837 Chr.9: 113413795-113413795 on Build GRCh38
rs758973957 Chr.9: 113413905-113413905 on Build GRCh38
rs764961326 Chr.9: 113413541-113413541 on Build GRCh38
rs1039244418 Chr.9: 113413575-113413575 on Build GRCh38
rs756867505 Chr.9: 113413532-113413532 on Build GRCh38
rs995677110 Chr.9: 113413698-113413698 on Build GRCh38
rs367568001 Chr.9: 113413602-113413602 on Build GRCh38
rs978197731 Chr.9: 113413813-113413813 on Build GRCh38
rs955858759 Chr.9: 113413824-113413824 on Build GRCh38
rs754306786 Chr.9: 113413657-113413657 on Build GRCh38
rs202243408 Chr.9: 113413921-113413921 on Build GRCh38
rs780202684 Chr.9: 113413844-113413844 on Build GRCh38
rs995667187 Chr.9: 113414006-113414006 on Build GRCh38
rs773920376 Chr.9: 113413874-113413874 on Build GRCh38
rs1027233555 Chr.9: 113413703-113413703 on Build GRCh38
rs897327681 Chr.9: 113413976-113413976 on Build GRCh38
rs773877768 Chr.9: 113413603-113413603 on Build GRCh38
rs1036787535 Chr.9: 113413963-113413963 on Build GRCh38

View Full Product Details