Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 152863614-152864184 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARFIP1
Entrez Gene ID
27236
Gene Name
ADP ribosylation factor interacting protein 1
Gene Aliases
HSU52521
UniGene
Hs.416089
Gene Chromosome Location
Chr.4: 152779937-152911912 on Build GRCh38
Transcripts
NM_001025593.2 NM_001025595.2 NM_001287431.1 NM_001287432.1 NM_001287433.1 NM_014447.3 XM_011531868.2 XM_011531869.2 XM_011531870.2 XM_011531871.2 XM_011531872.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4470744 c.167C>T p.T56I Chr.4: 152863679-152863679 on Build GRCh38
3365525 c.107C>A p.S36* Chr.4: 152863619-152863619 on Build GRCh38
4532801 c.191G>C p.G64A Chr.4: 152863703-152863703 on Build GRCh38
5883954 c.149C>G p.S50C Chr.4: 152863661-152863661 on Build GRCh38
5883953 c.149C>G p.S50C Chr.4: 152863661-152863661 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs552053120 Chr.4: 152863908-152863908 on Build GRCh38
rs1009400624 Chr.4: 152863988-152863988 on Build GRCh38
rs770036152 Chr.4: 152863736-152863736 on Build GRCh38
rs765078538 Chr.4: 152864130-152864130 on Build GRCh38
rs28410523 Chr.4: 152864149-152864149 on Build GRCh38
rs975322536 Chr.4: 152863781-152863781 on Build GRCh38
rs747539544 Chr.4: 152863676-152863676 on Build GRCh38
rs556107481 Chr.4: 152863861-152863861 on Build GRCh38
rs1005737472 Chr.4: 152863986-152863991 on Build GRCh38
rs775492660 Chr.4: 152863765-152863765 on Build GRCh38
rs1055395197 Chr.4: 152863868-152863868 on Build GRCh38
rs774345134 Chr.4: 152864022-152864022 on Build GRCh38
rs80259950 Chr.4: 152863793-152863793 on Build GRCh38
rs886493345 Chr.4: 152863926-152863926 on Build GRCh38
rs561563117 Chr.4: 152864098-152864098 on Build GRCh38
rs770973564 Chr.4: 152863763-152863763 on Build GRCh38
rs536318352 Chr.4: 152863705-152863705 on Build GRCh38
rs1008893153 Chr.4: 152863722-152863722 on Build GRCh38
rs542267313 Chr.4: 152863935-152863935 on Build GRCh38
rs989297970 Chr.4: 152863807-152863807 on Build GRCh38
rs761542583 Chr.4: 152864129-152864129 on Build GRCh38
rs1048399844 Chr.4: 152863912-152863912 on Build GRCh38
rs921068684 Chr.4: 152863783-152863783 on Build GRCh38
rs572547658 Chr.4: 152864020-152864020 on Build GRCh38
rs965287491 Chr.4: 152863739-152863739 on Build GRCh38
rs368596152 Chr.4: 152863639-152863639 on Build GRCh38
rs1018925486 Chr.4: 152864012-152864012 on Build GRCh38
rs62319916 Chr.4: 152864095-152864095 on Build GRCh38
rs796127993 Chr.4: 152864103-152864105 on Build GRCh38
rs780601305 Chr.4: 152863673-152863673 on Build GRCh38
rs749980564 Chr.4: 152863653-152863653 on Build GRCh38
rs183829445 Chr.4: 152864096-152864096 on Build GRCh38
rs751628195 Chr.4: 152863997-152863998 on Build GRCh38
rs1046731425 Chr.4: 152863873-152863873 on Build GRCh38
rs1021214816 Chr.4: 152864112-152864112 on Build GRCh38
rs147195311 Chr.4: 152863691-152863691 on Build GRCh38
rs577114597 Chr.4: 152863797-152863797 on Build GRCh38
rs749541326 Chr.4: 152863737-152863737 on Build GRCh38
rs927620897 Chr.4: 152864145-152864145 on Build GRCh38
rs538185445 Chr.4: 152863835-152863835 on Build GRCh38
rs981765990 Chr.4: 152863829-152863829 on Build GRCh38
rs938220209 Chr.4: 152863854-152863854 on Build GRCh38
rs989266666 Chr.4: 152864101-152864101 on Build GRCh38
rs994892017 Chr.4: 152863686-152863686 on Build GRCh38
rs114169627 Chr.4: 152863790-152863790 on Build GRCh38
rs899442492 Chr.4: 152863871-152863871 on Build GRCh38
rs141673805 Chr.4: 152863709-152863709 on Build GRCh38
rs374694647 Chr.4: 152863960-152863960 on Build GRCh38
rs534134939 Chr.4: 152863743-152863743 on Build GRCh38
rs758904927 Chr.4: 152863666-152863666 on Build GRCh38
rs979292584 Chr.4: 152864103-152864103 on Build GRCh38
rs945153252 Chr.4: 152863813-152863813 on Build GRCh38
rs560384979 Chr.4: 152863944-152863944 on Build GRCh38
rs371942166 Chr.4: 152863694-152863694 on Build GRCh38
rs773210934 Chr.4: 152863740-152863740 on Build GRCh38
rs927622020 Chr.4: 152863836-152863836 on Build GRCh38
rs1016682732 Chr.4: 152864030-152864030 on Build GRCh38
rs1039870081 Chr.4: 152864152-152864152 on Build GRCh38
rs1043130586 Chr.4: 152863706-152863706 on Build GRCh38
rs1019373469 Chr.4: 152863733-152863733 on Build GRCh38
rs900517070 Chr.4: 152864046-152864046 on Build GRCh38
rs982287143 Chr.4: 152864133-152864133 on Build GRCh38
rs913648978 Chr.4: 152863809-152863809 on Build GRCh38
rs771012668 Chr.4: 152863756-152863756 on Build GRCh38
rs200431909 Chr.4: 152864108-152864108 on Build GRCh38
rs567015324 Chr.4: 152863723-152863723 on Build GRCh38
rs777993394 Chr.4: 152863662-152863662 on Build GRCh38
rs926049286 Chr.4: 152864128-152864128 on Build GRCh38
rs540044203 Chr.4: 152864034-152864034 on Build GRCh38

View Full Product Details