Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 394412-395001 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IRF4
Entrez Gene ID
3662
Gene Name
interferon regulatory factor 4
Gene Aliases
LSIRF, MUM1, NF-EM5, SHEP8
UniGene
Hs.401013
Gene Chromosome Location
Chr.6: 391739-411443 on Build GRCh38
Transcripts
NM_001195286.1 NM_002460.3 NR_046000.2 XM_006715090.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5925276 c.349G>A p.D117N Chr.6: 394953-394953 on Build GRCh38
3077774 c.295T>C p.C99R Chr.6: 394899-394899 on Build GRCh38
6961314 c.319T>A p.F107I Chr.6: 394923-394923 on Build GRCh38
6933271 c.292C>T p.R98W Chr.6: 394896-394896 on Build GRCh38
7201950 c.217-2A>T p.? Chr.6: 394819-394819 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs931864176 Chr.6: 394431-394431 on Build GRCh38
rs920453866 Chr.6: 394727-394727 on Build GRCh38
rs775268870 Chr.6: 394829-394829 on Build GRCh38
rs561546511 Chr.6: 394768-394768 on Build GRCh38
rs1039409170 Chr.6: 394673-394673 on Build GRCh38
rs770799573 Chr.6: 394797-394797 on Build GRCh38
rs768584121 Chr.6: 394791-394791 on Build GRCh38
rs779109423 Chr.6: 394832-394832 on Build GRCh38
rs374779996 Chr.6: 394593-394593 on Build GRCh38
rs778712289 Chr.6: 394967-394967 on Build GRCh38
rs144593192 Chr.6: 394866-394866 on Build GRCh38
rs556006415 Chr.6: 394488-394488 on Build GRCh38
rs200743910 Chr.6: 394632-394632 on Build GRCh38
rs962415583 Chr.6: 394707-394707 on Build GRCh38
rs753842732 Chr.6: 394941-394941 on Build GRCh38
rs1035715640 Chr.6: 394495-394495 on Build GRCh38
rs868152149 Chr.6: 394681-394681 on Build GRCh38
rs200303421 Chr.6: 394918-394918 on Build GRCh38
rs145478558 Chr.6: 394889-394889 on Build GRCh38
rs202124383 Chr.6: 394867-394867 on Build GRCh38
rs2797308 Chr.6: 394711-394711 on Build GRCh38
rs951202749 Chr.6: 394463-394463 on Build GRCh38
rs977662507 Chr.6: 394590-394590 on Build GRCh38
rs371768937 Chr.6: 394796-394796 on Build GRCh38
rs749876602 Chr.6: 394783-394783 on Build GRCh38
rs570153443 Chr.6: 394528-394528 on Build GRCh38
rs982074949 Chr.6: 394703-394703 on Build GRCh38
rs754867881 Chr.6: 394823-394823 on Build GRCh38
rs999700699 Chr.6: 394612-394612 on Build GRCh38
rs958973315 Chr.6: 394522-394522 on Build GRCh38
rs377399870 Chr.6: 394443-394443 on Build GRCh38
rs944381620 Chr.6: 394942-394942 on Build GRCh38
rs756358063 Chr.6: 394919-394919 on Build GRCh38
rs540377898 Chr.6: 394536-394536 on Build GRCh38
rs772059663 Chr.6: 394804-394804 on Build GRCh38
rs1015012935 Chr.6: 394704-394704 on Build GRCh38
rs367593784 Chr.6: 394784-394784 on Build GRCh38
rs947702977 Chr.6: 394578-394578 on Build GRCh38
rs756125464 Chr.6: 394795-394795 on Build GRCh38
rs756489003 Chr.6: 394695-394695 on Build GRCh38
rs374588785 Chr.6: 394932-394932 on Build GRCh38
rs767879003 Chr.6: 394902-394902 on Build GRCh38
rs2671428 Chr.6: 394643-394643 on Build GRCh38
rs745876631 Chr.6: 394981-394981 on Build GRCh38
rs777434501 Chr.6: 394547-394547 on Build GRCh38
rs1050844890 Chr.6: 394622-394622 on Build GRCh38
rs770415537 Chr.6: 394621-394621 on Build GRCh38
rs548038312 Chr.6: 394757-394757 on Build GRCh38
rs944371595 Chr.6: 394554-394554 on Build GRCh38
rs1012531858 Chr.6: 394666-394666 on Build GRCh38
rs773571916 Chr.6: 394917-394917 on Build GRCh38
rs112896428 Chr.6: 394958-394958 on Build GRCh38
rs148625904 Chr.6: 394648-394648 on Build GRCh38
rs560282242 Chr.6: 394603-394603 on Build GRCh38
rs1035747182 Chr.6: 394801-394801 on Build GRCh38
rs1023874449 Chr.6: 394689-394689 on Build GRCh38
rs902896036 Chr.6: 394611-394611 on Build GRCh38
rs761433144 Chr.6: 394874-394874 on Build GRCh38
rs535745683 Chr.6: 394476-394476 on Build GRCh38
rs995463111 Chr.6: 394697-394697 on Build GRCh38
rs781384052 Chr.6: 394790-394790 on Build GRCh38
rs116609963 Chr.6: 394880-394880 on Build GRCh38
rs745622741 Chr.6: 394802-394802 on Build GRCh38
rs760417834 Chr.6: 394847-394847 on Build GRCh38

View Full Product Details