Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 95537618-95538174 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ASB4
Entrez Gene ID
51666
Gene Name
ankyrin repeat and SOCS box containing 4
Gene Aliases
ASB-4
UniGene
Hs.602765
Gene Chromosome Location
Chr.7: 95485439-95544738 on Build GRCh38
Transcripts
NM_016116.2 XM_017012303.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
748419 c.1213C>T p.P405S Chr.7: 95537691-95537691 on Build GRCh38
4524363 c.1268G>A p.G423E Chr.7: 95537746-95537746 on Build GRCh38
748420 c.1210A>T p.I404F Chr.7: 95537688-95537688 on Build GRCh38
3883565 c.1246T>C p.Y416H Chr.7: 95537724-95537724 on Build GRCh38
7423135 c.1143A>T p.P381P Chr.7: 95537621-95537621 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs553028152 Chr.7: 95538094-95538094 on Build GRCh38
rs368628140 Chr.7: 95537678-95537678 on Build GRCh38
rs185064872 Chr.7: 95537889-95537889 on Build GRCh38
rs887310905 Chr.7: 95537923-95537923 on Build GRCh38
rs267601648 Chr.7: 95537691-95537691 on Build GRCh38
rs757029594 Chr.7: 95537758-95537758 on Build GRCh38
rs567577566 Chr.7: 95537831-95537831 on Build GRCh38
rs577035251 Chr.7: 95537998-95537998 on Build GRCh38
rs370213105 Chr.7: 95537940-95537940 on Build GRCh38
rs780238414 Chr.7: 95537647-95537647 on Build GRCh38
rs533803896 Chr.7: 95537931-95537931 on Build GRCh38
rs758587757 Chr.7: 95537646-95537646 on Build GRCh38
rs958589725 Chr.7: 95538103-95538103 on Build GRCh38
rs6465467 Chr.7: 95537892-95537892 on Build GRCh38
rs967209385 Chr.7: 95538055-95538055 on Build GRCh38
rs780009942 Chr.7: 95537754-95537754 on Build GRCh38
rs1047133244 Chr.7: 95537913-95537913 on Build GRCh38
rs760591550 Chr.7: 95537736-95537736 on Build GRCh38
rs868373034 Chr.7: 95538030-95538030 on Build GRCh38
rs763947521 Chr.7: 95537738-95537738 on Build GRCh38
rs770461882 Chr.7: 95537702-95537702 on Build GRCh38
rs773741911 Chr.7: 95537806-95537806 on Build GRCh38
rs778074001 Chr.7: 95537799-95537799 on Build GRCh38
rs562534305 Chr.7: 95538091-95538091 on Build GRCh38
rs561081117 Chr.7: 95538133-95538133 on Build GRCh38
rs267601649 Chr.7: 95537707-95537707 on Build GRCh38
rs978560221 Chr.7: 95538063-95538063 on Build GRCh38
rs189393227 Chr.7: 95537902-95537902 on Build GRCh38
rs377035609 Chr.7: 95537801-95537801 on Build GRCh38
rs747216323 Chr.7: 95537650-95537650 on Build GRCh38
rs753855421 Chr.7: 95537745-95537745 on Build GRCh38
rs28414348 Chr.7: 95538122-95538122 on Build GRCh38
rs763421697 Chr.7: 95537699-95537699 on Build GRCh38
rs941490681 Chr.7: 95537929-95537929 on Build GRCh38
rs147762255 Chr.7: 95537729-95537729 on Build GRCh38
rs751508144 Chr.7: 95537655-95537657 on Build GRCh38
rs546087938 Chr.7: 95538050-95538050 on Build GRCh38
rs771373292 Chr.7: 95537802-95537802 on Build GRCh38
rs201104760 Chr.7: 95537728-95537728 on Build GRCh38
rs149668942 Chr.7: 95537721-95537721 on Build GRCh38
rs549154065 Chr.7: 95537739-95537739 on Build GRCh38
rs773757488 Chr.7: 95537687-95537687 on Build GRCh38
rs770130688 Chr.7: 95537682-95537682 on Build GRCh38
rs372903179 Chr.7: 95537776-95537776 on Build GRCh38
rs934730944 Chr.7: 95538002-95538002 on Build GRCh38
rs987221083 Chr.7: 95537714-95537714 on Build GRCh38
rs991407775 Chr.7: 95538121-95538121 on Build GRCh38
rs768863503 Chr.7: 95537659-95537659 on Build GRCh38
rs553817885 Chr.7: 95537964-95537964 on Build GRCh38
rs773865308 Chr.7: 95537704-95537704 on Build GRCh38
rs191411461 Chr.7: 95537841-95537841 on Build GRCh38
rs567320199 Chr.7: 95537674-95537674 on Build GRCh38
rs569440979 Chr.7: 95537858-95537858 on Build GRCh38
rs757210569 Chr.7: 95537751-95537751 on Build GRCh38
rs890116427 Chr.7: 95538009-95538009 on Build GRCh38
rs753769496 Chr.7: 95537642-95537642 on Build GRCh38
rs752274771 Chr.7: 95537735-95537735 on Build GRCh38
rs539343126 Chr.7: 95537810-95537810 on Build GRCh38
rs1013022962 Chr.7: 95538110-95538110 on Build GRCh38
rs933909348 Chr.7: 95537977-95537977 on Build GRCh38
rs755007826 Chr.7: 95537761-95537761 on Build GRCh38
rs538388292 Chr.7: 95538129-95538129 on Build GRCh38
rs1029098076 Chr.7: 95538004-95538004 on Build GRCh38
rs1023172572 Chr.7: 95537686-95537686 on Build GRCh38
rs913087125 Chr.7: 95537726-95537726 on Build GRCh38
rs868247987 Chr.7: 95537672-95537672 on Build GRCh38
rs758067988 Chr.7: 95538046-95538046 on Build GRCh38
rs748348441 Chr.7: 95537768-95537768 on Build GRCh38
rs900065337 Chr.7: 95537986-95537986 on Build GRCh38
rs761686052 Chr.7: 95537687-95537687 on Build GRCh38

View Full Product Details