Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 96027980-96028572 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DYNC1I1
Entrez Gene ID
1780
Gene Name
dynein cytoplasmic 1 intermediate chain 1
Gene Aliases
DNCI1, DNCIC1
UniGene
Hs.440364
Gene Chromosome Location
Chr.7: 95772506-96110322 on Build GRCh38
Transcripts
NM_001135556.1 NM_001135557.1 NM_001278421.1 NM_001278422.1 NM_004411.4 XM_011515861.1 XM_011515862.1 XM_011515863.2 XM_017011804.1 XM_017011805.1 XM_017011806.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4679805 c.1053C>G p.F351L Chr.7: 96028207-96028207 on Build GRCh38
6650589 c.942C>G p.F314L Chr.7: 96028207-96028207 on Build GRCh38
5830770 c.1027C>T p.R343W Chr.7: 96028292-96028292 on Build GRCh38
4679806 c.1139G>A p.R380Q Chr.7: 96028293-96028293 on Build GRCh38
6515273 c.1037T>A p.V346D Chr.7: 96028191-96028191 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764966313 Chr.7: 96028280-96028280 on Build GRCh38
rs764683489 Chr.7: 96028257-96028257 on Build GRCh38
rs201240633 Chr.7: 96028189-96028189 on Build GRCh38
rs913296254 Chr.7: 96028258-96028258 on Build GRCh38
rs751344917 Chr.7: 96028331-96028331 on Build GRCh38
rs542606937 Chr.7: 96028038-96028038 on Build GRCh38
rs1004528534 Chr.7: 96028420-96028420 on Build GRCh38
rs771229270 Chr.7: 96028207-96028207 on Build GRCh38
rs754978607 Chr.7: 96028337-96028337 on Build GRCh38
rs756202767 Chr.7: 96028199-96028199 on Build GRCh38
rs1001037455 Chr.7: 96028375-96028375 on Build GRCh38
rs372905393 Chr.7: 96028281-96028281 on Build GRCh38
rs778373700 Chr.7: 96028350-96028350 on Build GRCh38
rs531111773 Chr.7: 96028075-96028075 on Build GRCh38
rs886175566 Chr.7: 96028416-96028416 on Build GRCh38
rs899115388 Chr.7: 96028445-96028445 on Build GRCh38
rs746262928 Chr.7: 96028211-96028211 on Build GRCh38
rs754077557 Chr.7: 96028101-96028101 on Build GRCh38
rs370643324 Chr.7: 96028240-96028240 on Build GRCh38
rs950050593 Chr.7: 96028138-96028138 on Build GRCh38
rs546629907 Chr.7: 96028320-96028320 on Build GRCh38
rs181323240 Chr.7: 96028354-96028354 on Build GRCh38
rs745307681 Chr.7: 96028393-96028393 on Build GRCh38
rs761198234 Chr.7: 96028277-96028277 on Build GRCh38
rs17853779 Chr.7: 96028202-96028202 on Build GRCh38
rs902089808 Chr.7: 96028265-96028265 on Build GRCh38
rs1023078526 Chr.7: 96028425-96028425 on Build GRCh38
rs753557317 Chr.7: 96028169-96028169 on Build GRCh38
rs1008707661 Chr.7: 96028550-96028550 on Build GRCh38
rs776347302 Chr.7: 96028274-96028274 on Build GRCh38
rs755252314 Chr.7: 96028134-96028134 on Build GRCh38
rs772281540 Chr.7: 96028233-96028233 on Build GRCh38
rs779245715 Chr.7: 96028136-96028136 on Build GRCh38
rs376877688 Chr.7: 96028338-96028338 on Build GRCh38
rs150280272 Chr.7: 96028071-96028071 on Build GRCh38
rs573722815 Chr.7: 96028027-96028027 on Build GRCh38
rs190121247 Chr.7: 96028023-96028023 on Build GRCh38
rs368049969 Chr.7: 96028503-96028503 on Build GRCh38
rs950513289 Chr.7: 96028522-96028522 on Build GRCh38
rs567309321 Chr.7: 96028144-96028144 on Build GRCh38
rs565359613 Chr.7: 96028074-96028074 on Build GRCh38
rs537161457 Chr.7: 96028018-96028018 on Build GRCh38
rs981278230 Chr.7: 96028142-96028142 on Build GRCh38
rs950194968 Chr.7: 96028390-96028390 on Build GRCh38
rs376275541 Chr.7: 96028203-96028203 on Build GRCh38
rs78731424 Chr.7: 96028253-96028253 on Build GRCh38
rs145322007 Chr.7: 96028145-96028146 on Build GRCh38
rs778426093 Chr.7: 96028173-96028173 on Build GRCh38
rs758219234 Chr.7: 96028286-96028286 on Build GRCh38
rs552410088 Chr.7: 96028378-96028378 on Build GRCh38
rs371014160 Chr.7: 96028073-96028073 on Build GRCh38
rs1032730580 Chr.7: 96028392-96028392 on Build GRCh38
rs560963017 Chr.7: 96028198-96028198 on Build GRCh38
rs762235826 Chr.7: 96028168-96028169 on Build GRCh38
rs753204180 Chr.7: 96028139-96028139 on Build GRCh38
rs759784080 Chr.7: 96028127-96028127 on Build GRCh38
rs981733973 Chr.7: 96028435-96028435 on Build GRCh38
rs375850948 Chr.7: 96028321-96028321 on Build GRCh38
rs954536370 Chr.7: 96028242-96028242 on Build GRCh38
rs73399006 Chr.7: 96028049-96028049 on Build GRCh38
rs954721217 Chr.7: 96028532-96028532 on Build GRCh38
rs534810161 Chr.7: 96028406-96028406 on Build GRCh38
rs774640094 Chr.7: 96028144-96028145 on Build GRCh38
rs532323215 Chr.7: 96028355-96028355 on Build GRCh38
rs767708769 Chr.7: 96028172-96028172 on Build GRCh38
rs760290749 Chr.7: 96028145-96028145 on Build GRCh38
rs763628350 Chr.7: 96028152-96028152 on Build GRCh38
rs917406662 Chr.7: 96028384-96028384 on Build GRCh38
rs915824784 Chr.7: 96028107-96028107 on Build GRCh38
rs369991411 Chr.7: 96028172-96028172 on Build GRCh38
rs774410304 Chr.7: 96028209-96028209 on Build GRCh38
rs141086884 Chr.7: 96028058-96028058 on Build GRCh38
rs996505540 Chr.7: 96028488-96028488 on Build GRCh38
rs113331208 Chr.7: 96028426-96028426 on Build GRCh38
rs909293554 Chr.7: 96028539-96028539 on Build GRCh38
rs750281458 Chr.7: 96028185-96028185 on Build GRCh38
rs776143490 Chr.7: 96028239-96028239 on Build GRCh38

View Full Product Details