Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 65603342-65603881 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARMC1
Entrez Gene ID
55156
Gene Name
armadillo repeat containing 1
Gene Aliases
Arcp
UniGene
Hs.269542
Gene Chromosome Location
Chr.8: 65602456-65634217 on Build GRCh38
Transcripts
NM_001286702.1 NM_018120.5 XM_005251264.1 XM_017013605.1

SNP Detail

SNP ID Chromosome Location
rs756854669 Chr.8: 65603489-65603489 on Build GRCh38
rs189891136 Chr.8: 65603839-65603839 on Build GRCh38
rs1041700613 Chr.8: 65603557-65603557 on Build GRCh38
rs1025349800 Chr.8: 65603727-65603727 on Build GRCh38
rs1053088 Chr.8: 65603548-65603548 on Build GRCh38
rs984804467 Chr.8: 65603784-65603784 on Build GRCh38
rs959185687 Chr.8: 65603832-65603832 on Build GRCh38
rs904497337 Chr.8: 65603578-65603578 on Build GRCh38
rs75653646 Chr.8: 65603852-65603852 on Build GRCh38
rs1053087 Chr.8: 65603630-65603630 on Build GRCh38
rs937507950 Chr.8: 65603499-65603499 on Build GRCh38
rs530025703 Chr.8: 65603660-65603660 on Build GRCh38
rs150129252 Chr.8: 65603470-65603470 on Build GRCh38
rs1006711336 Chr.8: 65603634-65603635 on Build GRCh38
rs368147598 Chr.8: 65603520-65603520 on Build GRCh38
rs749905275 Chr.8: 65603626-65603626 on Build GRCh38
rs80289702 Chr.8: 65603831-65603831 on Build GRCh38
rs374215308 Chr.8: 65603466-65603466 on Build GRCh38
rs1053085 Chr.8: 65603753-65603753 on Build GRCh38
rs900926270 Chr.8: 65603684-65603687 on Build GRCh38
rs182288000 Chr.8: 65603417-65603417 on Build GRCh38
rs946108861 Chr.8: 65603556-65603556 on Build GRCh38
rs552805450 Chr.8: 65603549-65603549 on Build GRCh38
rs927494612 Chr.8: 65603473-65603473 on Build GRCh38
rs746802249 Chr.8: 65603677-65603677 on Build GRCh38
rs542543575 Chr.8: 65603423-65603423 on Build GRCh38
rs753201719 Chr.8: 65603478-65603478 on Build GRCh38
rs561367630 Chr.8: 65603391-65603391 on Build GRCh38
rs993353393 Chr.8: 65603531-65603531 on Build GRCh38
rs372049744 Chr.8: 65603527-65603527 on Build GRCh38
rs185131404 Chr.8: 65603635-65603635 on Build GRCh38
rs965069804 Chr.8: 65603373-65603373 on Build GRCh38
rs993884633 Chr.8: 65603702-65603702 on Build GRCh38

View Full Product Details