Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 126552714-126553309 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM84B
Entrez Gene ID
157638
Gene Name
family with sequence similarity 84 member B
Gene Aliases
BCMP101, NSE2
UniGene
Hs.741352
Gene Chromosome Location
Chr.8: 126552438-126558499 on Build GRCh38
Transcripts
NM_174911.4 XM_017013107.1 XM_017013108.1

SNP Detail

SNP ID Chromosome Location
rs565554501 Chr.8: 126553246-126553246 on Build GRCh38
rs936846295 Chr.8: 126552942-126552943 on Build GRCh38
rs575178845 Chr.8: 126553261-126553261 on Build GRCh38
rs915360674 Chr.8: 126553233-126553233 on Build GRCh38
rs146635007 Chr.8: 126552739-126552739 on Build GRCh38
rs956651386 Chr.8: 126553183-126553183 on Build GRCh38
rs1026909615 Chr.8: 126553032-126553032 on Build GRCh38
rs923529777 Chr.8: 126552870-126552870 on Build GRCh38
rs554714906 Chr.8: 126552839-126552839 on Build GRCh38
rs1024123045 Chr.8: 126553139-126553139 on Build GRCh38
rs766070185 Chr.8: 126552957-126552957 on Build GRCh38
rs1012736926 Chr.8: 126553129-126553129 on Build GRCh38
rs749821060 Chr.8: 126552807-126552808 on Build GRCh38
rs185754471 Chr.8: 126552770-126552770 on Build GRCh38
rs541602803 Chr.8: 126553140-126553140 on Build GRCh38
rs181935525 Chr.8: 126553081-126553081 on Build GRCh38
rs866323375 Chr.8: 126553099-126553099 on Build GRCh38
rs533744584 Chr.8: 126553212-126553212 on Build GRCh38
rs905104353 Chr.8: 126552978-126552978 on Build GRCh38
rs897462244 Chr.8: 126552956-126552956 on Build GRCh38
rs977335496 Chr.8: 126553155-126553155 on Build GRCh38
rs539884163 Chr.8: 126553000-126553000 on Build GRCh38
rs778047941 Chr.8: 126553067-126553067 on Build GRCh38
rs555277916 Chr.8: 126553198-126553198 on Build GRCh38
rs867263824 Chr.8: 126552754-126552754 on Build GRCh38
rs749605829 Chr.8: 126553080-126553080 on Build GRCh38
rs932265905 Chr.8: 126552921-126552921 on Build GRCh38
rs11539702 Chr.8: 126552939-126552939 on Build GRCh38
rs1050572682 Chr.8: 126552823-126552823 on Build GRCh38
rs16901511 Chr.8: 126553028-126553028 on Build GRCh38
rs765220874 Chr.8: 126552808-126552808 on Build GRCh38
rs976502883 Chr.8: 126552869-126552869 on Build GRCh38
rs888241230 Chr.8: 126553074-126553074 on Build GRCh38
rs989570207 Chr.8: 126553191-126553191 on Build GRCh38
rs986749825 Chr.8: 126553274-126553274 on Build GRCh38
rs892230847 Chr.8: 126552950-126552950 on Build GRCh38
rs138144332 Chr.8: 126552940-126552940 on Build GRCh38
rs952099633 Chr.8: 126552814-126552815 on Build GRCh38
rs1036468635 Chr.8: 126552944-126552944 on Build GRCh38
rs757916008 Chr.8: 126552942-126552942 on Build GRCh38
rs1046509037 Chr.8: 126552974-126552974 on Build GRCh38
rs1031436711 Chr.8: 126553165-126553165 on Build GRCh38
rs190225865 Chr.8: 126552814-126552814 on Build GRCh38
rs962014401 Chr.8: 126552855-126552855 on Build GRCh38

View Full Product Details