Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 22450876-22451426 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DMRTA1
Entrez Gene ID
63951
Gene Name
DMRT like family A1
Gene Aliases
DMO, DMRT4
UniGene
Hs.371976
Gene Chromosome Location
Chr.9: 22446841-22452473 on Build GRCh38
Transcripts
NM_022160.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5903702 c.730C>T p.H244Y Chr.9: 22451126-22451126 on Build GRCh38
5876078 c.706C>T p.Q236* Chr.9: 22451102-22451102 on Build GRCh38
3092369 c.793T>C p.S265P Chr.9: 22451189-22451189 on Build GRCh38
4588743 c.676G>C p.E226Q Chr.9: 22451072-22451072 on Build GRCh38
5803712 c.794C>T p.S265L Chr.9: 22451190-22451190 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs745339156 Chr.9: 22451294-22451294 on Build GRCh38
rs202197256 Chr.9: 22451350-22451350 on Build GRCh38
rs777522680 Chr.9: 22451191-22451191 on Build GRCh38
rs752706554 Chr.9: 22451237-22451237 on Build GRCh38
rs760350204 Chr.9: 22451046-22451046 on Build GRCh38
rs775961973 Chr.9: 22451049-22451049 on Build GRCh38
rs199550791 Chr.9: 22451131-22451131 on Build GRCh38
rs756763306 Chr.9: 22451195-22451195 on Build GRCh38
rs368798749 Chr.9: 22451328-22451328 on Build GRCh38
rs201374002 Chr.9: 22451232-22451232 on Build GRCh38
rs899760825 Chr.9: 22451387-22451387 on Build GRCh38
rs142330994 Chr.9: 22451099-22451099 on Build GRCh38
rs748101368 Chr.9: 22451010-22451010 on Build GRCh38
rs763988468 Chr.9: 22451256-22451256 on Build GRCh38
rs112667674 Chr.9: 22451372-22451372 on Build GRCh38
rs771724739 Chr.9: 22451107-22451107 on Build GRCh38
rs137914267 Chr.9: 22451356-22451356 on Build GRCh38
rs962865767 Chr.9: 22451233-22451233 on Build GRCh38
rs61729546 Chr.9: 22451340-22451340 on Build GRCh38
rs1015081364 Chr.9: 22451267-22451267 on Build GRCh38
rs74769337 Chr.9: 22451228-22451228 on Build GRCh38
rs746675446 Chr.9: 22451193-22451193 on Build GRCh38
rs551088682 Chr.9: 22451363-22451363 on Build GRCh38
rs181512603 Chr.9: 22451185-22451185 on Build GRCh38
rs768996476 Chr.9: 22451125-22451125 on Build GRCh38
rs772447910 Chr.9: 22451222-22451222 on Build GRCh38
rs1000721470 Chr.9: 22450963-22450963 on Build GRCh38
rs748876077 Chr.9: 22450992-22450992 on Build GRCh38
rs750111980 Chr.9: 22451380-22451380 on Build GRCh38
rs776947180 Chr.9: 22451349-22451349 on Build GRCh38
rs779775461 Chr.9: 22451026-22451026 on Build GRCh38
rs1038599209 Chr.9: 22451048-22451048 on Build GRCh38
rs543015454 Chr.9: 22451147-22451147 on Build GRCh38
rs777374100 Chr.9: 22451108-22451108 on Build GRCh38
rs779865358 Chr.9: 22451077-22451077 on Build GRCh38
rs771032631 Chr.9: 22451348-22451348 on Build GRCh38
rs778714494 Chr.9: 22451092-22451092 on Build GRCh38
rs369778210 Chr.9: 22451358-22451358 on Build GRCh38
rs777892111 Chr.9: 22451281-22451283 on Build GRCh38
rs370079512 Chr.9: 22451040-22451040 on Build GRCh38
rs761541718 Chr.9: 22451039-22451040 on Build GRCh38
rs34324424 Chr.9: 22451230-22451230 on Build GRCh38
rs1033016510 Chr.9: 22450954-22450954 on Build GRCh38
rs758459142 Chr.9: 22451178-22451179 on Build GRCh38
rs146478718 Chr.9: 22451009-22451009 on Build GRCh38
rs368374702 Chr.9: 22451376-22451376 on Build GRCh38
rs1041549610 Chr.9: 22451182-22451182 on Build GRCh38
rs745441990 Chr.9: 22451204-22451204 on Build GRCh38
rs773362908 Chr.9: 22451223-22451223 on Build GRCh38
rs143713234 Chr.9: 22451203-22451203 on Build GRCh38
rs751379079 Chr.9: 22451369-22451369 on Build GRCh38
rs753740634 Chr.9: 22451181-22451181 on Build GRCh38
rs762982635 Chr.9: 22451362-22451362 on Build GRCh38
rs371496399 Chr.9: 22451384-22451384 on Build GRCh38
rs936560925 Chr.9: 22451215-22451215 on Build GRCh38
rs758212329 Chr.9: 22451239-22451239 on Build GRCh38
rs780948456 Chr.9: 22451287-22451287 on Build GRCh38
rs142995523 Chr.9: 22451270-22451270 on Build GRCh38
rs781198333 Chr.9: 22450935-22450935 on Build GRCh38
rs773007196 Chr.9: 22451042-22451042 on Build GRCh38
rs1029635429 Chr.9: 22451212-22451212 on Build GRCh38
rs777036865 Chr.9: 22451211-22451211 on Build GRCh38
rs747600792 Chr.9: 22451039-22451039 on Build GRCh38
rs145718826 Chr.9: 22451121-22451121 on Build GRCh38
rs371171820 Chr.9: 22451344-22451344 on Build GRCh38
rs752699944 Chr.9: 22451109-22451110 on Build GRCh38
rs150186157 Chr.9: 22451118-22451118 on Build GRCh38
rs754863697 Chr.9: 22451183-22451183 on Build GRCh38
rs765451375 Chr.9: 22451103-22451105 on Build GRCh38
rs369631878 Chr.9: 22451048-22451049 on Build GRCh38
rs267602201 Chr.9: 22451304-22451304 on Build GRCh38
rs770727275 Chr.9: 22451089-22451089 on Build GRCh38
rs1006426815 Chr.9: 22451266-22451266 on Build GRCh38
rs755916609 Chr.9: 22451018-22451018 on Build GRCh38
rs771947601 Chr.9: 22451268-22451268 on Build GRCh38
rs370396351 Chr.9: 22451076-22451076 on Build GRCh38
rs79094105 Chr.9: 22450989-22450989 on Build GRCh38
rs976230965 Chr.9: 22451172-22451172 on Build GRCh38
rs141039588 Chr.9: 22451151-22451151 on Build GRCh38
rs41270141 Chr.9: 22451040-22451040 on Build GRCh38
rs952326505 Chr.9: 22451259-22451259 on Build GRCh38
rs749280346 Chr.9: 22451028-22451028 on Build GRCh38
rs770309947 Chr.9: 22451047-22451047 on Build GRCh38
rs751726182 Chr.9: 22451059-22451059 on Build GRCh38
rs267602200 Chr.9: 22451124-22451124 on Build GRCh38
rs145152745 Chr.9: 22451278-22451278 on Build GRCh38
rs748471195 Chr.9: 22451220-22451220 on Build GRCh38
rs767694946 Chr.9: 22451140-22451140 on Build GRCh38
rs949805564 Chr.9: 22451354-22451354 on Build GRCh38
rs770682109 Chr.9: 22451120-22451120 on Build GRCh38
rs778537010 Chr.9: 22451038-22451038 on Build GRCh38
rs138782225 Chr.9: 22451402-22451402 on Build GRCh38
rs563802 Chr.9: 22451032-22451032 on Build GRCh38
rs372547221 Chr.9: 22450940-22450940 on Build GRCh38
rs1001508505 Chr.9: 22451244-22451244 on Build GRCh38
rs541190708 Chr.9: 22451053-22451053 on Build GRCh38
rs760888914 Chr.9: 22451227-22451227 on Build GRCh38
rs139030144 Chr.9: 22451338-22451338 on Build GRCh38
rs368428828 Chr.9: 22451217-22451217 on Build GRCh38
rs753555375 Chr.9: 22451088-22451088 on Build GRCh38
rs1033333947 Chr.9: 22451188-22451188 on Build GRCh38
rs763561805 Chr.9: 22451052-22451052 on Build GRCh38
rs750397300 Chr.9: 22451040-22451041 on Build GRCh38
rs1034283321 Chr.9: 22451248-22451248 on Build GRCh38
rs931952036 Chr.9: 22450918-22450918 on Build GRCh38
rs150709609 Chr.9: 22451343-22451343 on Build GRCh38
rs766457846 Chr.9: 22451173-22451173 on Build GRCh38
rs748778435 Chr.9: 22451072-22451072 on Build GRCh38
rs779618649 Chr.9: 22451392-22451392 on Build GRCh38
rs200749823 Chr.9: 22451385-22451385 on Build GRCh38
rs779238351 Chr.9: 22451324-22451324 on Build GRCh38
rs148944998 Chr.9: 22451186-22451186 on Build GRCh38
rs755681561 Chr.9: 22451301-22451301 on Build GRCh38
rs764433058 Chr.9: 22451127-22451127 on Build GRCh38
rs964353908 Chr.9: 22450983-22450983 on Build GRCh38
rs996752450 Chr.9: 22451400-22451400 on Build GRCh38

View Full Product Details