Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 13326276-13326809 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARNTL
Entrez Gene ID
406
Gene Name
aryl hydrocarbon receptor nuclear translocator like
Gene Aliases
BMAL1, BMAL1c, JAP3, MOP3, PASD3, TIC, bHLHe5
UniGene
Hs.65734
Gene Chromosome Location
Chr.11: 13276613-13387268 on Build GRCh38
Transcripts
NM_001030272.2 NM_001030273.2 NM_001178.5 NM_001297719.1 NM_001297722.1 NM_001297724.1 XM_006718233.3 XM_006718234.1 XM_011520105.2 XM_011520107.2 XM_011520108.1 XM_011520109.1 XM_011520110.1 XM_011520111.1 XM_011520112.2 XM_011520113.1 XM_017017738.1 XM_017017739.1 XM_017017740.1 XM_017017741.1 XM_017017742.1 XM_017017743.1 XM_017017744.1 XM_017017745.1 XM_017017746.1 XM_017017747.1

SNP Detail

SNP ID Chromosome Location
rs372995148 Chr.11: 13326741-13326741 on Build GRCh38
rs757666582 Chr.11: 13326601-13326601 on Build GRCh38
rs951925489 Chr.11: 13326508-13326508 on Build GRCh38
rs1054651455 Chr.11: 13326347-13326347 on Build GRCh38
rs540660374 Chr.11: 13326359-13326359 on Build GRCh38
rs896985162 Chr.11: 13326618-13326618 on Build GRCh38
rs775520851 Chr.11: 13326728-13326729 on Build GRCh38
rs1054816054 Chr.11: 13326562-13326562 on Build GRCh38
rs1013513637 Chr.11: 13326603-13326603 on Build GRCh38
rs546646334 Chr.11: 13326695-13326695 on Build GRCh38
rs1007491608 Chr.11: 13326662-13326662 on Build GRCh38
rs780289251 Chr.11: 13326726-13326729 on Build GRCh38
rs901527011 Chr.11: 13326744-13326744 on Build GRCh38
rs560888031 Chr.11: 13326668-13326668 on Build GRCh38
rs576955556 Chr.11: 13326418-13326418 on Build GRCh38
rs564600672 Chr.11: 13326457-13326457 on Build GRCh38
rs377273591 Chr.11: 13326742-13326742 on Build GRCh38
rs910784353 Chr.11: 13326390-13326390 on Build GRCh38
rs914847742 Chr.11: 13326619-13326619 on Build GRCh38
rs562610795 Chr.11: 13326473-13326473 on Build GRCh38
rs1014583719 Chr.11: 13326528-13326530 on Build GRCh38
rs896377159 Chr.11: 13326583-13326583 on Build GRCh38
rs975859068 Chr.11: 13326567-13326567 on Build GRCh38
rs922130187 Chr.11: 13326330-13326330 on Build GRCh38
rs67286103 Chr.11: 13326726-13326727 on Build GRCh38
rs199836202 Chr.11: 13326688-13326689 on Build GRCh38
rs11022766 Chr.11: 13326702-13326702 on Build GRCh38
rs953558508 Chr.11: 13326611-13326611 on Build GRCh38
rs73421894 Chr.11: 13326629-13326629 on Build GRCh38
rs995780670 Chr.11: 13326621-13326621 on Build GRCh38
rs928657372 Chr.11: 13326638-13326638 on Build GRCh38
rs61882381 Chr.11: 13326696-13326696 on Build GRCh38
rs1004590495 Chr.11: 13326509-13326509 on Build GRCh38
rs183970477 Chr.11: 13326435-13326435 on Build GRCh38
rs142987849 Chr.11: 13326377-13326377 on Build GRCh38
rs57010022 Chr.11: 13326739-13326740 on Build GRCh38
rs952153003 Chr.11: 13326645-13326645 on Build GRCh38
rs372644424 Chr.11: 13326755-13326755 on Build GRCh38
rs966662433 Chr.11: 13326687-13326687 on Build GRCh38
rs527969563 Chr.11: 13326694-13326694 on Build GRCh38
rs1012059055 Chr.11: 13326464-13326464 on Build GRCh38
rs1043479236 Chr.11: 13326458-13326458 on Build GRCh38
rs925055960 Chr.11: 13326471-13326471 on Build GRCh38
rs551060109 Chr.11: 13326302-13326302 on Build GRCh38
rs907316418 Chr.11: 13326383-13326383 on Build GRCh38
rs530327853 Chr.11: 13326573-13326573 on Build GRCh38
rs1027734603 Chr.11: 13326354-13326354 on Build GRCh38
rs748013984 Chr.11: 13326631-13326631 on Build GRCh38
rs369418940 Chr.11: 13326739-13326739 on Build GRCh38
rs978044703 Chr.11: 13326630-13326630 on Build GRCh38
rs756179773 Chr.11: 13326767-13326767 on Build GRCh38
rs972177482 Chr.11: 13326296-13326296 on Build GRCh38
rs932112911 Chr.11: 13326336-13326336 on Build GRCh38
rs966337097 Chr.11: 13326417-13326417 on Build GRCh38
rs201906845 Chr.11: 13326701-13326702 on Build GRCh38
rs921652568 Chr.11: 13326574-13326574 on Build GRCh38
rs976689639 Chr.11: 13326317-13326317 on Build GRCh38
rs914176414 Chr.11: 13326699-13326699 on Build GRCh38
rs887728556 Chr.11: 13326727-13326727 on Build GRCh38
rs1037233088 Chr.11: 13326604-13326604 on Build GRCh38

View Full Product Details