Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 10752458-10753057 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CTR9
Entrez Gene ID
9646
Gene Name
CTR9 homolog, Paf1/RNA polymerase II complex component
Gene Aliases
SH2BP1, TSBP, p150, p150TSP
UniGene
Hs.725151
Gene Chromosome Location
Chr.11: 10750987-10779755 on Build GRCh38
Transcripts
NM_014633.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1604026 c.46-7A>G p.? Chr.11: 10752665-10752665 on Build GRCh38
4456103 c.72_73AC>CT p.L24_P25>FS Chr.11: 10752698-10752699 on Build GRCh38
4507552 c.74C>T p.P25L Chr.11: 10752700-10752700 on Build GRCh38
1202627 c.143C>T p.A48V Chr.11: 10752769-10752769 on Build GRCh38
5625084 c.68_69insG p.L24fs*6 Chr.11: 10752694-10752695 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs144498813 Chr.11: 10752879-10752879 on Build GRCh38
rs368879631 Chr.11: 10752633-10752633 on Build GRCh38
rs533330669 Chr.11: 10752527-10752527 on Build GRCh38
rs745549347 Chr.11: 10752665-10752666 on Build GRCh38
rs750362295 Chr.11: 10752639-10752639 on Build GRCh38
rs940002356 Chr.11: 10752594-10752594 on Build GRCh38
rs1000085934 Chr.11: 10752882-10752882 on Build GRCh38
rs1049368857 Chr.11: 10752618-10752618 on Build GRCh38
rs953944123 Chr.11: 10752602-10752602 on Build GRCh38
rs535207885 Chr.11: 10752573-10752573 on Build GRCh38
rs372627965 Chr.11: 10752808-10752808 on Build GRCh38
rs77924039 Chr.11: 10752485-10752485 on Build GRCh38
rs770281838 Chr.11: 10752888-10752888 on Build GRCh38
rs540034855 Chr.11: 10752743-10752743 on Build GRCh38
rs938268850 Chr.11: 10752806-10752806 on Build GRCh38
rs949504090 Chr.11: 10752658-10752658 on Build GRCh38
rs921397422 Chr.11: 10752553-10752553 on Build GRCh38
rs917021899 Chr.11: 10752654-10752654 on Build GRCh38
rs774921914 Chr.11: 10752659-10752659 on Build GRCh38
rs955710732 Chr.11: 10752898-10752898 on Build GRCh38
rs930560306 Chr.11: 10752997-10752997 on Build GRCh38
rs547193282 Chr.11: 10752529-10752529 on Build GRCh38
rs755621124 Chr.11: 10752798-10752799 on Build GRCh38
rs755268144 Chr.11: 10752689-10752689 on Build GRCh38
rs921439546 Chr.11: 10752799-10752799 on Build GRCh38
rs781265429 Chr.11: 10752690-10752690 on Build GRCh38
rs763869251 Chr.11: 10752787-10752787 on Build GRCh38
rs188933909 Chr.11: 10752829-10752829 on Build GRCh38
rs994715211 Chr.11: 10752616-10752616 on Build GRCh38
rs766273456 Chr.11: 10752819-10752819 on Build GRCh38
rs138850547 Chr.11: 10752701-10752701 on Build GRCh38
rs113769029 Chr.11: 10752585-10752585 on Build GRCh38
rs564596204 Chr.11: 10753032-10753032 on Build GRCh38
rs758376573 Chr.11: 10752818-10752818 on Build GRCh38
rs746433028 Chr.11: 10752840-10752840 on Build GRCh38
rs1028107602 Chr.11: 10752862-10752862 on Build GRCh38
rs771372197 Chr.11: 10752798-10752798 on Build GRCh38
rs774896371 Chr.11: 10752786-10752786 on Build GRCh38
rs191895699 Chr.11: 10752834-10752834 on Build GRCh38
rs184141599 Chr.11: 10752614-10752614 on Build GRCh38
rs149264597 Chr.11: 10752839-10752839 on Build GRCh38
rs112818649 Chr.11: 10752902-10752902 on Build GRCh38
rs568557544 Chr.11: 10752586-10752586 on Build GRCh38
rs532242348 Chr.11: 10752781-10752781 on Build GRCh38
rs753458776 Chr.11: 10752623-10752623 on Build GRCh38
rs1036261670 Chr.11: 10752601-10752601 on Build GRCh38
rs577881740 Chr.11: 10752683-10752683 on Build GRCh38
rs998246299 Chr.11: 10752845-10752845 on Build GRCh38
rs888478257 Chr.11: 10752514-10752514 on Build GRCh38
rs965159311 Chr.11: 10752560-10752560 on Build GRCh38
rs770004696 Chr.11: 10752719-10752719 on Build GRCh38
rs74732143 Chr.11: 10752534-10752534 on Build GRCh38
rs979128875 Chr.11: 10752609-10752609 on Build GRCh38
rs1049129184 Chr.11: 10752823-10752823 on Build GRCh38
rs779842828 Chr.11: 10752680-10752680 on Build GRCh38
rs192886164 Chr.11: 10752577-10752577 on Build GRCh38
rs1033347157 Chr.11: 10752884-10752884 on Build GRCh38
rs971119240 Chr.11: 10752971-10752971 on Build GRCh38
rs912576065 Chr.11: 10752579-10752579 on Build GRCh38
rs761465438 Chr.11: 10752807-10752807 on Build GRCh38
rs758289280 Chr.11: 10752643-10752643 on Build GRCh38
rs957588589 Chr.11: 10752492-10752492 on Build GRCh38
rs756711073 Chr.11: 10752626-10752626 on Build GRCh38
rs916948570 Chr.11: 10752942-10752942 on Build GRCh38
rs372687739 Chr.11: 10752941-10752941 on Build GRCh38
rs1037046301 Chr.11: 10752837-10752837 on Build GRCh38
rs553969590 Chr.11: 10752770-10752770 on Build GRCh38
rs557725587 Chr.11: 10752621-10752621 on Build GRCh38

View Full Product Details