Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 67120985-67121342 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KDM2A
Entrez Gene ID
22992
Gene Name
lysine demethylase 2A
Gene Aliases
CXXC8, FBL11, FBL7, FBXL11, JHDM1A, LILINA
UniGene
Hs.124147
Gene Chromosome Location
Chr.11: 67119210-67258082 on Build GRCh38
Transcripts
NM_012308.2 NR_027473.1 XM_011544860.2 XM_017017391.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7157934 c.21G>A p.R7R Chr.11: 67121337-67121337 on Build GRCh38
5627435 c.10G>A p.E4K Chr.11: 67121326-67121326 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1023767583 Chr.11: 67121305-67121305 on Build GRCh38
rs928207207 Chr.11: 67121019-67121019 on Build GRCh38
rs900591867 Chr.11: 67121149-67121149 on Build GRCh38
rs768249114 Chr.11: 67121176-67121176 on Build GRCh38
rs549572174 Chr.11: 67121135-67121136 on Build GRCh38
rs571452924 Chr.11: 67121034-67121034 on Build GRCh38
rs760718472 Chr.11: 67121272-67121272 on Build GRCh38
rs1028538461 Chr.11: 67121142-67121142 on Build GRCh38
rs990957756 Chr.11: 67121260-67121260 on Build GRCh38
rs73493534 Chr.11: 67121078-67121078 on Build GRCh38
rs557030536 Chr.11: 67121219-67121219 on Build GRCh38
rs574677409 Chr.11: 67121042-67121042 on Build GRCh38
rs542058492 Chr.11: 67121043-67121043 on Build GRCh38
rs1006745129 Chr.11: 67121115-67121115 on Build GRCh38
rs990926464 Chr.11: 67121030-67121030 on Build GRCh38
rs960917454 Chr.11: 67121252-67121252 on Build GRCh38
rs61663253 Chr.11: 67121114-67121114 on Build GRCh38
rs200263280 Chr.11: 67121316-67121316 on Build GRCh38
rs866079223 Chr.11: 67121108-67121108 on Build GRCh38
rs999465829 Chr.11: 67121186-67121186 on Build GRCh38
rs553130154 Chr.11: 67121024-67121024 on Build GRCh38
rs370553998 Chr.11: 67121231-67121231 on Build GRCh38
rs143642280 Chr.11: 67121242-67121242 on Build GRCh38
rs752449240 Chr.11: 67121169-67121169 on Build GRCh38
rs1056345292 Chr.11: 67121029-67121029 on Build GRCh38
rs928921573 Chr.11: 67121018-67121018 on Build GRCh38
rs1011031215 Chr.11: 67121154-67121154 on Build GRCh38
rs941088205 Chr.11: 67121050-67121050 on Build GRCh38
rs1051035796 Chr.11: 67121100-67121100 on Build GRCh38
rs749097197 Chr.11: 67121163-67121163 on Build GRCh38
rs966712652 Chr.11: 67121166-67121166 on Build GRCh38
rs553039638 Chr.11: 67121023-67121023 on Build GRCh38
rs1035256147 Chr.11: 67121198-67121198 on Build GRCh38
rs543324502 Chr.11: 67121165-67121165 on Build GRCh38
rs181740725 Chr.11: 67121053-67121053 on Build GRCh38
rs931355414 Chr.11: 67121038-67121038 on Build GRCh38
rs750611586 Chr.11: 67121268-67121268 on Build GRCh38
rs769839540 Chr.11: 67121065-67121065 on Build GRCh38
rs1039511051 Chr.11: 67121132-67121132 on Build GRCh38
rs766414564 Chr.11: 67121289-67121289 on Build GRCh38
rs957928532 Chr.11: 67121148-67121148 on Build GRCh38

View Full Product Details