Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 66565534-66566103 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CTSF
Entrez Gene ID
8722
Gene Name
cathepsin F
Gene Aliases
CATSF, CLN13
UniGene
Hs.11590
Gene Chromosome Location
Chr.11: 66563463-66568576 on Build GRCh38
Transcripts
NM_003793.3 XM_011545328.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5970316 c.962A>G p.Q321R Chr.11: 66565833-66565833 on Build GRCh38
6552310 c.803G>A p.G268D Chr.11: 66566086-66566086 on Build GRCh38
930799 c.1029G>A p.S343S Chr.11: 66565687-66565687 on Build GRCh38
4035752 c.963G>A p.Q321Q Chr.11: 66565832-66565832 on Build GRCh38
4035753 c.901A>T p.T301S Chr.11: 66565894-66565894 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779569569 Chr.11: 66565740-66565740 on Build GRCh38
rs773555427 Chr.11: 66565697-66565697 on Build GRCh38
rs749794934 Chr.11: 66565744-66565744 on Build GRCh38
rs778286278 Chr.11: 66565977-66565977 on Build GRCh38
rs776395150 Chr.11: 66566052-66566052 on Build GRCh38
rs755755081 Chr.11: 66565837-66565837 on Build GRCh38
rs745821135 Chr.11: 66566049-66566050 on Build GRCh38
rs747224695 Chr.11: 66565888-66565888 on Build GRCh38
rs774742010 Chr.11: 66565687-66565687 on Build GRCh38
rs757544408 Chr.11: 66565995-66565995 on Build GRCh38
rs781540749 Chr.11: 66566014-66566014 on Build GRCh38
rs928771076 Chr.11: 66565564-66565564 on Build GRCh38
rs373234919 Chr.11: 66565931-66565931 on Build GRCh38
rs758932306 Chr.11: 66565976-66565976 on Build GRCh38
rs891337822 Chr.11: 66565808-66565808 on Build GRCh38
rs202242498 Chr.11: 66565800-66565800 on Build GRCh38
rs761661216 Chr.11: 66565819-66565819 on Build GRCh38
rs980671668 Chr.11: 66566046-66566046 on Build GRCh38
rs1051519244 Chr.11: 66565752-66565752 on Build GRCh38
rs756862617 Chr.11: 66565627-66565627 on Build GRCh38
rs755473264 Chr.11: 66565765-66565765 on Build GRCh38
rs768927293 Chr.11: 66565664-66565664 on Build GRCh38
rs114727660 Chr.11: 66565856-66565856 on Build GRCh38
rs753334318 Chr.11: 66565844-66565844 on Build GRCh38
rs780071531 Chr.11: 66566016-66566016 on Build GRCh38
rs893705162 Chr.11: 66565832-66565832 on Build GRCh38
rs776587445 Chr.11: 66565897-66565897 on Build GRCh38
rs979422706 Chr.11: 66565940-66565940 on Build GRCh38
rs753084727 Chr.11: 66565841-66565841 on Build GRCh38
rs941653140 Chr.11: 66566076-66566076 on Build GRCh38
rs967357477 Chr.11: 66565895-66565895 on Build GRCh38
rs200009655 Chr.11: 66565822-66565822 on Build GRCh38
rs201489835 Chr.11: 66565945-66565945 on Build GRCh38
rs377331616 Chr.11: 66565766-66565766 on Build GRCh38
rs569797867 Chr.11: 66565723-66565723 on Build GRCh38
rs752001068 Chr.11: 66566078-66566078 on Build GRCh38
rs932976138 Chr.11: 66566065-66566065 on Build GRCh38
rs759062690 Chr.11: 66566055-66566055 on Build GRCh38
rs554536950 Chr.11: 66565944-66565944 on Build GRCh38
rs549988778 Chr.11: 66565718-66565718 on Build GRCh38
rs528090170 Chr.11: 66565572-66565572 on Build GRCh38
rs142805637 Chr.11: 66566022-66566022 on Build GRCh38
rs769277856 Chr.11: 66565640-66565640 on Build GRCh38
rs942306652 Chr.11: 66565602-66565602 on Build GRCh38
rs397514731 Chr.11: 66565833-66565833 on Build GRCh38
rs748086442 Chr.11: 66566029-66566029 on Build GRCh38
rs779160410 Chr.11: 66565748-66565748 on Build GRCh38
rs747166209 Chr.11: 66565942-66565947 on Build GRCh38
rs763792720 Chr.11: 66565683-66565684 on Build GRCh38
rs764755120 Chr.11: 66566073-66566073 on Build GRCh38
rs200646712 Chr.11: 66565688-66565688 on Build GRCh38
rs746813891 Chr.11: 66565637-66565637 on Build GRCh38
rs780490009 Chr.11: 66565648-66565648 on Build GRCh38
rs757342215 Chr.11: 66566005-66566006 on Build GRCh38
rs760906269 Chr.11: 66565702-66565702 on Build GRCh38
rs370003726 Chr.11: 66565614-66565614 on Build GRCh38
rs750069810 Chr.11: 66565728-66565728 on Build GRCh38
rs547731057 Chr.11: 66565595-66565595 on Build GRCh38
rs774308794 Chr.11: 66565930-66565930 on Build GRCh38
rs373433586 Chr.11: 66565639-66565639 on Build GRCh38
rs748464271 Chr.11: 66565600-66565600 on Build GRCh38
rs746013208 Chr.11: 66565907-66565907 on Build GRCh38
rs758884287 Chr.11: 66565853-66565853 on Build GRCh38
rs189862070 Chr.11: 66565710-66565710 on Build GRCh38
rs749783276 Chr.11: 66565655-66565655 on Build GRCh38
rs774875383 Chr.11: 66565973-66565973 on Build GRCh38
rs867463018 Chr.11: 66565703-66565703 on Build GRCh38
rs771117136 Chr.11: 66565890-66565890 on Build GRCh38
rs762134997 Chr.11: 66565690-66565690 on Build GRCh38
rs779630399 Chr.11: 66565850-66565850 on Build GRCh38
rs748637318 Chr.11: 66565852-66565852 on Build GRCh38
rs774076056 Chr.11: 66565712-66565714 on Build GRCh38
rs764869338 Chr.11: 66566056-66566056 on Build GRCh38
rs963662861 Chr.11: 66566001-66566001 on Build GRCh38
rs147398226 Chr.11: 66566079-66566079 on Build GRCh38
rs760262888 Chr.11: 66565947-66565947 on Build GRCh38
rs752624429 Chr.11: 66565707-66565707 on Build GRCh38
rs200423355 Chr.11: 66565961-66565961 on Build GRCh38
rs781196961 Chr.11: 66566027-66566028 on Build GRCh38
rs767333189 Chr.11: 66565943-66565943 on Build GRCh38
rs764030927 Chr.11: 66565813-66565813 on Build GRCh38
rs754420487 Chr.11: 66565758-66565765 on Build GRCh38
rs762896720 Chr.11: 66565708-66565708 on Build GRCh38
rs773124981 Chr.11: 66566037-66566037 on Build GRCh38
rs777967311 Chr.11: 66565923-66565924 on Build GRCh38
rs115077591 Chr.11: 66565978-66565978 on Build GRCh38
rs754434242 Chr.11: 66565849-66565849 on Build GRCh38
rs376173549 Chr.11: 66565949-66565949 on Build GRCh38
rs762803735 Chr.11: 66565811-66565811 on Build GRCh38
rs761415902 Chr.11: 66565716-66565716 on Build GRCh38
rs752186392 Chr.11: 66566058-66566058 on Build GRCh38
rs763567392 Chr.11: 66566059-66566059 on Build GRCh38
rs765816185 Chr.11: 66565841-66565841 on Build GRCh38
rs766453048 Chr.11: 66565705-66565705 on Build GRCh38
rs202050053 Chr.11: 66565803-66565803 on Build GRCh38
rs941706968 Chr.11: 66565755-66565755 on Build GRCh38
rs753429669 Chr.11: 66565960-66565960 on Build GRCh38
rs755617235 Chr.11: 66565733-66565733 on Build GRCh38
rs767265591 Chr.11: 66565824-66565824 on Build GRCh38
rs767081949 Chr.11: 66565723-66565731 on Build GRCh38
rs762549521 Chr.11: 66565647-66565649 on Build GRCh38
rs753277717 Chr.11: 66565964-66565964 on Build GRCh38
rs957016251 Chr.11: 66565948-66565948 on Build GRCh38
rs765562144 Chr.11: 66565808-66565810 on Build GRCh38
rs577007516 Chr.11: 66566015-66566015 on Build GRCh38
rs917810286 Chr.11: 66565619-66565619 on Build GRCh38
rs780681439 Chr.11: 66565635-66565635 on Build GRCh38
rs575373998 Chr.11: 66565756-66565756 on Build GRCh38
rs888913381 Chr.11: 66565764-66565764 on Build GRCh38
rs774255251 Chr.11: 66565818-66565818 on Build GRCh38
rs760584939 Chr.11: 66566040-66566040 on Build GRCh38
rs764820313 Chr.11: 66565963-66565963 on Build GRCh38
rs769143533 Chr.11: 66565745-66565745 on Build GRCh38
rs759461850 Chr.11: 66565911-66565911 on Build GRCh38
rs760955343 Chr.11: 66565790-66565790 on Build GRCh38
rs933049796 Chr.11: 66565739-66565739 on Build GRCh38
rs751192764 Chr.11: 66565626-66565626 on Build GRCh38
rs748582111 Chr.11: 66565751-66565751 on Build GRCh38
rs565888514 Chr.11: 66565835-66565835 on Build GRCh38
rs771943482 Chr.11: 66566033-66566033 on Build GRCh38
rs958903211 Chr.11: 66565599-66565599 on Build GRCh38
rs4630309 Chr.11: 66565601-66565601 on Build GRCh38
rs147269500 Chr.11: 66565711-66565711 on Build GRCh38
rs758605201 Chr.11: 66565843-66565843 on Build GRCh38
rs776587423 Chr.11: 66565820-66565820 on Build GRCh38

View Full Product Details