Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 112175259-112175840 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BCO2
Entrez Gene ID
83875
Gene Name
beta-carotene oxygenase 2
Gene Aliases
B-DIOX-II, BCDO2
UniGene
Hs.728325
Gene Chromosome Location
Chr.11: 112175485-112218929 on Build GRCh38
Transcripts
NM_001256398.1 NM_031938.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4017855 c.79C>T p.R27W Chr.11: 112175680-112175680 on Build GRCh38
6240397 c.16delT p.L7fs*17 Chr.11: 112175617-112175617 on Build GRCh38
5124008 c.3_4insT p.R4fs*75 Chr.11: 112175604-112175605 on Build GRCh38
5682738 c.51A>T p.A17A Chr.11: 112175652-112175652 on Build GRCh38
6857438 c.15C>T p.V5V Chr.11: 112175616-112175616 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs900741094 Chr.11: 112175692-112175692 on Build GRCh38
rs777303682 Chr.11: 112175639-112175639 on Build GRCh38
rs184802086 Chr.11: 112175393-112175393 on Build GRCh38
rs775490373 Chr.11: 112175666-112175666 on Build GRCh38
rs759219872 Chr.11: 112175812-112175812 on Build GRCh38
rs894769402 Chr.11: 112175568-112175568 on Build GRCh38
rs1017834964 Chr.11: 112175441-112175441 on Build GRCh38
rs1053247173 Chr.11: 112175598-112175598 on Build GRCh38
rs1017741909 Chr.11: 112175688-112175688 on Build GRCh38
rs563959898 Chr.11: 112175302-112175302 on Build GRCh38
rs765264497 Chr.11: 112175585-112175585 on Build GRCh38
rs775196003 Chr.11: 112175557-112175557 on Build GRCh38
rs761291940 Chr.11: 112175701-112175702 on Build GRCh38
rs759006668 Chr.11: 112175460-112175460 on Build GRCh38
rs534710109 Chr.11: 112175533-112175533 on Build GRCh38
rs554418402 Chr.11: 112175571-112175571 on Build GRCh38
rs543226302 Chr.11: 112175729-112175729 on Build GRCh38
rs145373157 Chr.11: 112175648-112175648 on Build GRCh38
rs775415073 Chr.11: 112175815-112175815 on Build GRCh38
rs767456599 Chr.11: 112175553-112175553 on Build GRCh38
rs907255841 Chr.11: 112175378-112175378 on Build GRCh38
rs760387441 Chr.11: 112175672-112175672 on Build GRCh38
rs953968612 Chr.11: 112175814-112175814 on Build GRCh38
rs919330605 Chr.11: 112175489-112175489 on Build GRCh38
rs892877520 Chr.11: 112175757-112175757 on Build GRCh38
rs909048914 Chr.11: 112175293-112175293 on Build GRCh38
rs758469836 Chr.11: 112175704-112175704 on Build GRCh38
rs1028071521 Chr.11: 112175807-112175807 on Build GRCh38
rs916531446 Chr.11: 112175523-112175523 on Build GRCh38
rs528610482 Chr.11: 112175505-112175505 on Build GRCh38
rs765425934 Chr.11: 112175689-112175689 on Build GRCh38
rs747400349 Chr.11: 112175622-112175622 on Build GRCh38
rs568281159 Chr.11: 112175728-112175728 on Build GRCh38
rs528570000 Chr.11: 112175362-112175362 on Build GRCh38
rs780599024 Chr.11: 112175611-112175611 on Build GRCh38
rs764836772 Chr.11: 112175712-112175712 on Build GRCh38
rs574249150 Chr.11: 112175657-112175657 on Build GRCh38
rs922959721 Chr.11: 112175481-112175481 on Build GRCh38
rs753094407 Chr.11: 112175654-112175663 on Build GRCh38
rs532594868 Chr.11: 112175373-112175373 on Build GRCh38
rs201320164 Chr.11: 112175592-112175592 on Build GRCh38
rs755303166 Chr.11: 112175636-112175636 on Build GRCh38
rs552173590 Chr.11: 112175291-112175291 on Build GRCh38
rs137871179 Chr.11: 112175423-112175423 on Build GRCh38
rs781579692 Chr.11: 112175741-112175741 on Build GRCh38
rs748899293 Chr.11: 112175643-112175643 on Build GRCh38
rs373466897 Chr.11: 112175665-112175665 on Build GRCh38
rs1055083315 Chr.11: 112175307-112175307 on Build GRCh38
rs781021182 Chr.11: 112175558-112175558 on Build GRCh38
rs752046659 Chr.11: 112175726-112175726 on Build GRCh38
rs563212687 Chr.11: 112175776-112175776 on Build GRCh38
rs571178251 Chr.11: 112175432-112175432 on Build GRCh38
rs766801984 Chr.11: 112175708-112175708 on Build GRCh38
rs998635982 Chr.11: 112175696-112175696 on Build GRCh38
rs769690902 Chr.11: 112175765-112175765 on Build GRCh38
rs949299317 Chr.11: 112175525-112175525 on Build GRCh38
rs184435119 Chr.11: 112175520-112175520 on Build GRCh38
rs538612221 Chr.11: 112175475-112175475 on Build GRCh38
rs957594972 Chr.11: 112175427-112175427 on Build GRCh38
rs757354544 Chr.11: 112175584-112175584 on Build GRCh38
rs1023975565 Chr.11: 112175316-112175316 on Build GRCh38
rs11214111 Chr.11: 112175488-112175488 on Build GRCh38
rs180736678 Chr.11: 112175431-112175431 on Build GRCh38
rs756920894 Chr.11: 112175542-112175542 on Build GRCh38
rs1012946715 Chr.11: 112175366-112175366 on Build GRCh38
rs909033683 Chr.11: 112175504-112175504 on Build GRCh38
rs762065931 Chr.11: 112175685-112175685 on Build GRCh38
rs983606526 Chr.11: 112175817-112175817 on Build GRCh38
rs750561934 Chr.11: 112175703-112175703 on Build GRCh38
rs1031863925 Chr.11: 112175744-112175744 on Build GRCh38
rs989982838 Chr.11: 112175428-112175428 on Build GRCh38
rs764285953 Chr.11: 112175563-112175563 on Build GRCh38
rs941915860 Chr.11: 112175512-112175512 on Build GRCh38
rs758955089 Chr.11: 112175596-112175596 on Build GRCh38
rs45616732 Chr.11: 112175424-112175424 on Build GRCh38
rs983229115 Chr.11: 112175503-112175503 on Build GRCh38
rs770594350 Chr.11: 112175644-112175644 on Build GRCh38
rs776873678 Chr.11: 112175683-112175683 on Build GRCh38
rs149163526 Chr.11: 112175680-112175680 on Build GRCh38

View Full Product Details