Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 125916890-125917477 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DDX25
Entrez Gene ID
29118
Gene Name
DEAD-box helicase 25
Gene Aliases
GRTH
UniGene
Hs.420263
Gene Chromosome Location
Chr.11: 125903275-125923110 on Build GRCh38
Transcripts
NM_013264.4 XM_011542790.2 XM_011542791.2 XM_017017626.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4901380 c.936C>T p.I312I Chr.11: 125917149-125917149 on Build GRCh38
4901381 c.594C>T p.I198I Chr.11: 125917149-125917149 on Build GRCh38
3397525 c.948C>T p.Y316Y Chr.11: 125917161-125917161 on Build GRCh38
260379 c.471C>T p.S157S Chr.11: 125917026-125917026 on Build GRCh38
7304398 c.675T>A p.G225G Chr.11: 125917230-125917230 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs778503027 Chr.11: 125917218-125917218 on Build GRCh38
rs371915891 Chr.11: 125917039-125917039 on Build GRCh38
rs375509873 Chr.11: 125917047-125917047 on Build GRCh38
rs200202882 Chr.11: 125917162-125917162 on Build GRCh38
rs766382483 Chr.11: 125917275-125917275 on Build GRCh38
rs377547552 Chr.11: 125917150-125917150 on Build GRCh38
rs1023680931 Chr.11: 125916937-125916937 on Build GRCh38
rs767615816 Chr.11: 125917313-125917313 on Build GRCh38
rs749542725 Chr.11: 125917149-125917149 on Build GRCh38
rs770456489 Chr.11: 125917121-125917121 on Build GRCh38
rs767427755 Chr.11: 125917144-125917144 on Build GRCh38
rs369309099 Chr.11: 125917103-125917103 on Build GRCh38
rs753128351 Chr.11: 125917192-125917192 on Build GRCh38
rs999713019 Chr.11: 125916918-125916918 on Build GRCh38
rs571765309 Chr.11: 125917308-125917308 on Build GRCh38
rs200801236 Chr.11: 125917196-125917196 on Build GRCh38
rs973316959 Chr.11: 125917301-125917301 on Build GRCh38
rs766440735 Chr.11: 125917173-125917173 on Build GRCh38
rs911342159 Chr.11: 125917400-125917400 on Build GRCh38
rs1012382295 Chr.11: 125917240-125917240 on Build GRCh38
rs891878124 Chr.11: 125917445-125917445 on Build GRCh38
rs778195394 Chr.11: 125917205-125917205 on Build GRCh38
rs756459929 Chr.11: 125917300-125917300 on Build GRCh38
rs201774241 Chr.11: 125917120-125917120 on Build GRCh38
rs557031103 Chr.11: 125917415-125917415 on Build GRCh38
rs751696761 Chr.11: 125917276-125917276 on Build GRCh38
rs962014107 Chr.11: 125917291-125917291 on Build GRCh38
rs191465600 Chr.11: 125917005-125917005 on Build GRCh38
rs768985429 Chr.11: 125916987-125916987 on Build GRCh38
rs748613880 Chr.11: 125917000-125917000 on Build GRCh38
rs137989955 Chr.11: 125916933-125916933 on Build GRCh38
rs758602872 Chr.11: 125916968-125916968 on Build GRCh38
rs773766991 Chr.11: 125917112-125917112 on Build GRCh38
rs770382761 Chr.11: 125917212-125917212 on Build GRCh38
rs759179116 Chr.11: 125916918-125916919 on Build GRCh38
rs752835848 Chr.11: 125917441-125917441 on Build GRCh38
rs768446624 Chr.11: 125917242-125917242 on Build GRCh38
rs748669088 Chr.11: 125917104-125917104 on Build GRCh38
rs907875162 Chr.11: 125916995-125916995 on Build GRCh38
rs770262163 Chr.11: 125917105-125917105 on Build GRCh38
rs760593355 Chr.11: 125917234-125917234 on Build GRCh38
rs375442824 Chr.11: 125917384-125917384 on Build GRCh38
rs920290943 Chr.11: 125917326-125917326 on Build GRCh38
rs891758378 Chr.11: 125917026-125917026 on Build GRCh38
rs962134680 Chr.11: 125917259-125917259 on Build GRCh38
rs371045804 Chr.11: 125917183-125917183 on Build GRCh38
rs539417356 Chr.11: 125917184-125917184 on Build GRCh38
rs766014409 Chr.11: 125917010-125917010 on Build GRCh38
rs372660107 Chr.11: 125917066-125917066 on Build GRCh38
rs549319796 Chr.11: 125917151-125917151 on Build GRCh38
rs762964204 Chr.11: 125917168-125917168 on Build GRCh38
rs755135902 Chr.11: 125917179-125917179 on Build GRCh38
rs528678746 Chr.11: 125916915-125916915 on Build GRCh38
rs902811538 Chr.11: 125916913-125916913 on Build GRCh38
rs767376280 Chr.11: 125917027-125917027 on Build GRCh38
rs749756606 Chr.11: 125917209-125917209 on Build GRCh38
rs374097266 Chr.11: 125917191-125917191 on Build GRCh38
rs767768323 Chr.11: 125917284-125917284 on Build GRCh38
rs759229312 Chr.11: 125917138-125917138 on Build GRCh38
rs985695616 Chr.11: 125917371-125917371 on Build GRCh38
rs552995568 Chr.11: 125917008-125917008 on Build GRCh38
rs780186824 Chr.11: 125917073-125917073 on Build GRCh38
rs752896974 Chr.11: 125917296-125917296 on Build GRCh38
rs920169725 Chr.11: 125916948-125916948 on Build GRCh38
rs972923708 Chr.11: 125916932-125916932 on Build GRCh38
rs747369986 Chr.11: 125916975-125916975 on Build GRCh38
rs971146192 Chr.11: 125916980-125916980 on Build GRCh38
rs771914420 Chr.11: 125917224-125917224 on Build GRCh38
rs780316800 Chr.11: 125916972-125916972 on Build GRCh38
rs764959974 Chr.11: 125916925-125916925 on Build GRCh38
rs78506115 Chr.11: 125917254-125917254 on Build GRCh38
rs755366877 Chr.11: 125917087-125917087 on Build GRCh38
rs774027403 Chr.11: 125917013-125917013 on Build GRCh38
rs548080420 Chr.11: 125917193-125917193 on Build GRCh38
rs751784540 Chr.11: 125917178-125917178 on Build GRCh38
rs376342805 Chr.11: 125917449-125917449 on Build GRCh38
rs777145507 Chr.11: 125916990-125916990 on Build GRCh38
rs370049057 Chr.11: 125917065-125917065 on Build GRCh38
rs375741829 Chr.11: 125917278-125917278 on Build GRCh38
rs34015034 Chr.11: 125917017-125917017 on Build GRCh38
rs756471581 Chr.11: 125917203-125917203 on Build GRCh38
rs775249786 Chr.11: 125917231-125917231 on Build GRCh38
rs1003612496 Chr.11: 125917269-125917269 on Build GRCh38
rs970879148 Chr.11: 125917267-125917267 on Build GRCh38
rs534793318 Chr.11: 125917262-125917262 on Build GRCh38
rs545307523 Chr.11: 125917007-125917007 on Build GRCh38
rs931537040 Chr.11: 125917341-125917341 on Build GRCh38
rs375780207 Chr.11: 125917088-125917088 on Build GRCh38
rs745354339 Chr.11: 125917221-125917221 on Build GRCh38
rs756053356 Chr.11: 125917053-125917053 on Build GRCh38
rs747182884 Chr.11: 125917085-125917085 on Build GRCh38
rs986019901 Chr.11: 125917440-125917440 on Build GRCh38
rs188061977 Chr.11: 125917312-125917312 on Build GRCh38
rs920487551 Chr.11: 125917325-125917325 on Build GRCh38
rs907508203 Chr.11: 125916911-125916911 on Build GRCh38
rs763976592 Chr.11: 125917157-125917157 on Build GRCh38

View Full Product Details