Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 105043042-105043640 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CARD16
Entrez Gene ID
114769
Gene Name
caspase recruitment domain family member 16
Gene Aliases
COP, COP1, PSEUDO-ICE
UniGene
Hs.348365
Gene Chromosome Location
Chr.11: 105040218-105045416 on Build GRCh38
Transcripts
NM_001017534.1 NM_052889.2 XM_011542583.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
922455 c.288A>C p.G96G Chr.11: 105043532-105043532 on Build GRCh38
6620125 c.278C>T p.P93L Chr.11: 105043542-105043542 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs528959228 Chr.11: 105043090-105043090 on Build GRCh38
rs955835070 Chr.11: 105043330-105043330 on Build GRCh38
rs370234571 Chr.11: 105043487-105043487 on Build GRCh38
rs1139551 Chr.11: 105043517-105043517 on Build GRCh38
rs760639891 Chr.11: 105043591-105043591 on Build GRCh38
rs548738815 Chr.11: 105043129-105043129 on Build GRCh38
rs989372062 Chr.11: 105043310-105043310 on Build GRCh38
rs895706096 Chr.11: 105043209-105043209 on Build GRCh38
rs113908181 Chr.11: 105043575-105043575 on Build GRCh38
rs1035367382 Chr.11: 105043262-105043262 on Build GRCh38
rs778167915 Chr.11: 105043572-105043572 on Build GRCh38
rs757656586 Chr.11: 105043581-105043583 on Build GRCh38
rs779668838 Chr.11: 105043087-105043087 on Build GRCh38
rs745718311 Chr.11: 105043582-105043582 on Build GRCh38
rs759257539 Chr.11: 105043501-105043501 on Build GRCh38
rs370535181 Chr.11: 105043536-105043536 on Build GRCh38
rs534292201 Chr.11: 105043188-105043188 on Build GRCh38
rs536006729 Chr.11: 105043429-105043429 on Build GRCh38
rs747517423 Chr.11: 105043553-105043553 on Build GRCh38
rs924647387 Chr.11: 105043349-105043349 on Build GRCh38
rs187217579 Chr.11: 105043570-105043570 on Build GRCh38
rs775847222 Chr.11: 105043596-105043596 on Build GRCh38
rs778478685 Chr.11: 105043578-105043578 on Build GRCh38
rs1018480389 Chr.11: 105043618-105043618 on Build GRCh38
rs933301690 Chr.11: 105043378-105043378 on Build GRCh38
rs1139550 Chr.11: 105043541-105043541 on Build GRCh38
rs1126877 Chr.11: 105043526-105043526 on Build GRCh38
rs493426 Chr.11: 105043138-105043138 on Build GRCh38
rs542322676 Chr.11: 105043299-105043299 on Build GRCh38
rs757524229 Chr.11: 105043529-105043529 on Build GRCh38
rs569697863 Chr.11: 105043064-105043064 on Build GRCh38
rs186522272 Chr.11: 105043288-105043288 on Build GRCh38
rs911232225 Chr.11: 105043360-105043361 on Build GRCh38
rs199574838 Chr.11: 105043516-105043516 on Build GRCh38
rs375964015 Chr.11: 105043511-105043511 on Build GRCh38
rs899039353 Chr.11: 105043156-105043156 on Build GRCh38
rs746590335 Chr.11: 105043106-105043106 on Build GRCh38
rs78543105 Chr.11: 105043213-105043213 on Build GRCh38
rs985712205 Chr.11: 105043589-105043589 on Build GRCh38
rs563482189 Chr.11: 105043510-105043510 on Build GRCh38
rs528085832 Chr.11: 105043583-105043583 on Build GRCh38
rs192150849 Chr.11: 105043588-105043588 on Build GRCh38
rs376068204 Chr.11: 105043379-105043379 on Build GRCh38
rs910614795 Chr.11: 105043422-105043422 on Build GRCh38
rs941358687 Chr.11: 105043382-105043382 on Build GRCh38
rs539688565 Chr.11: 105043231-105043231 on Build GRCh38
rs1002611213 Chr.11: 105043246-105043246 on Build GRCh38
rs541393899 Chr.11: 105043401-105043401 on Build GRCh38
rs755624938 Chr.11: 105043563-105043563 on Build GRCh38
rs1139549 Chr.11: 105043542-105043542 on Build GRCh38
rs754347807 Chr.11: 105043581-105043581 on Build GRCh38
rs772810058 Chr.11: 105043484-105043484 on Build GRCh38
rs554047247 Chr.11: 105043201-105043201 on Build GRCh38
rs71037294 Chr.11: 105043361-105043361 on Build GRCh38
rs564411950 Chr.11: 105043404-105043404 on Build GRCh38
rs1014589804 Chr.11: 105043459-105043459 on Build GRCh38
rs1038454864 Chr.11: 105043395-105043395 on Build GRCh38
rs993733722 Chr.11: 105043180-105043180 on Build GRCh38
rs368391656 Chr.11: 105043544-105043544 on Build GRCh38
rs181358366 Chr.11: 105043268-105043268 on Build GRCh38
rs1022655763 Chr.11: 105043260-105043260 on Build GRCh38
rs774231233 Chr.11: 105043499-105043499 on Build GRCh38
rs373188276 Chr.11: 105043507-105043507 on Build GRCh38
rs190937535 Chr.11: 105043302-105043302 on Build GRCh38
rs776805464 Chr.11: 105043597-105043597 on Build GRCh38
rs999953736 Chr.11: 105043279-105043279 on Build GRCh38
rs376861355 Chr.11: 105043441-105043441 on Build GRCh38
rs770437660 Chr.11: 105043495-105043495 on Build GRCh38
rs775391773 Chr.11: 105043509-105043509 on Build GRCh38
rs1013068465 Chr.11: 105043555-105043555 on Build GRCh38
rs747676450 Chr.11: 105043480-105043480 on Build GRCh38
rs969027608 Chr.11: 105043319-105043319 on Build GRCh38
rs1006695929 Chr.11: 105043080-105043080 on Build GRCh38
rs1039565315 Chr.11: 105043432-105043432 on Build GRCh38
rs78361407 Chr.11: 105043203-105043203 on Build GRCh38
rs999846216 Chr.11: 105043512-105043512 on Build GRCh38
rs370953871 Chr.11: 105043566-105043566 on Build GRCh38
rs914822122 Chr.11: 105043309-105043309 on Build GRCh38
rs577377185 Chr.11: 105043241-105043241 on Build GRCh38
rs543280371 Chr.11: 105043466-105043466 on Build GRCh38
rs995814736 Chr.11: 105043172-105043172 on Build GRCh38
rs978060986 Chr.11: 105043329-105043329 on Build GRCh38
rs78242046 Chr.11: 105043230-105043230 on Build GRCh38
rs533259743 Chr.11: 105043443-105043443 on Build GRCh38
rs765338562 Chr.11: 105043534-105043534 on Build GRCh38

View Full Product Details