Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 108645698-108646296 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CORO1C
Entrez Gene ID
23603
Gene Name
coronin 1C
Gene Aliases
HCRNN4
UniGene
Hs.330384
Gene Chromosome Location
Chr.12: 108645109-108731550 on Build GRCh38
Transcripts
NM_001105237.2 NM_001276471.1 NM_014325.3 XM_011538125.1 XM_017019121.1 XM_017019122.1 XM_017019123.1

SNP Detail

SNP ID Chromosome Location
rs527606176 Chr.12: 108646212-108646212 on Build GRCh38
rs756386649 Chr.12: 108646263-108646264 on Build GRCh38
rs1048487657 Chr.12: 108645926-108645926 on Build GRCh38
rs542782687 Chr.12: 108646019-108646019 on Build GRCh38
rs906555376 Chr.12: 108646181-108646181 on Build GRCh38
rs989926558 Chr.12: 108645787-108645787 on Build GRCh38
rs1052841102 Chr.12: 108646240-108646240 on Build GRCh38
rs1024938724 Chr.12: 108645867-108645867 on Build GRCh38
rs189519591 Chr.12: 108646048-108646048 on Build GRCh38
rs755152346 Chr.12: 108646062-108646062 on Build GRCh38
rs2111212 Chr.12: 108646018-108646018 on Build GRCh38
rs2111211 Chr.12: 108646093-108646093 on Build GRCh38
rs961288647 Chr.12: 108646218-108646218 on Build GRCh38
rs759210917 Chr.12: 108645807-108645807 on Build GRCh38
rs906496313 Chr.12: 108645979-108645979 on Build GRCh38
rs767708215 Chr.12: 108646058-108646059 on Build GRCh38
rs999514186 Chr.12: 108646187-108646187 on Build GRCh38
rs979400474 Chr.12: 108646056-108646056 on Build GRCh38
rs1037285019 Chr.12: 108645896-108645896 on Build GRCh38
rs925690786 Chr.12: 108646022-108646022 on Build GRCh38
rs976726537 Chr.12: 108646219-108646219 on Build GRCh38
rs530163017 Chr.12: 108645915-108645915 on Build GRCh38
rs1033756651 Chr.12: 108646148-108646148 on Build GRCh38
rs367933341 Chr.12: 108646225-108646225 on Build GRCh38
rs1005343695 Chr.12: 108646185-108646185 on Build GRCh38
rs143509755 Chr.12: 108646065-108646065 on Build GRCh38
rs985185732 Chr.12: 108646044-108646044 on Build GRCh38
rs767889142 Chr.12: 108646010-108646010 on Build GRCh38
rs895252574 Chr.12: 108645929-108645929 on Build GRCh38
rs914305960 Chr.12: 108645795-108645795 on Build GRCh38
rs965067810 Chr.12: 108646049-108646049 on Build GRCh38
rs1034160162 Chr.12: 108645903-108645903 on Build GRCh38
rs184008121 Chr.12: 108645936-108645936 on Build GRCh38
rs1040678521 Chr.12: 108645794-108645794 on Build GRCh38
rs554744074 Chr.12: 108646036-108646036 on Build GRCh38
rs138436436 Chr.12: 108645828-108645828 on Build GRCh38
rs377732766 Chr.12: 108646006-108646006 on Build GRCh38
rs961239052 Chr.12: 108645996-108645996 on Build GRCh38
rs532971688 Chr.12: 108646150-108646150 on Build GRCh38
rs955848935 Chr.12: 108645755-108645755 on Build GRCh38
rs527515941 Chr.12: 108645822-108645822 on Build GRCh38
rs111829744 Chr.12: 108645804-108645804 on Build GRCh38
rs897351609 Chr.12: 108645899-108645899 on Build GRCh38
rs918823456 Chr.12: 108646055-108646055 on Build GRCh38
rs538340244 Chr.12: 108645831-108645831 on Build GRCh38
rs1051047038 Chr.12: 108645806-108645806 on Build GRCh38
rs545306596 Chr.12: 108646162-108646162 on Build GRCh38
rs1021437341 Chr.12: 108646025-108646025 on Build GRCh38
rs148857954 Chr.12: 108646005-108646005 on Build GRCh38
rs953777688 Chr.12: 108646266-108646266 on Build GRCh38
rs560452161 Chr.12: 108646204-108646204 on Build GRCh38
rs948139673 Chr.12: 108646118-108646118 on Build GRCh38
rs1025405060 Chr.12: 108645974-108645974 on Build GRCh38
rs1046474073 Chr.12: 108646171-108646171 on Build GRCh38
rs116426326 Chr.12: 108645825-108645825 on Build GRCh38
rs780122698 Chr.12: 108646040-108646040 on Build GRCh38
rs906556576 Chr.12: 108646115-108646115 on Build GRCh38
rs571658664 Chr.12: 108645901-108645901 on Build GRCh38
rs973425840 Chr.12: 108646050-108646050 on Build GRCh38
rs556244041 Chr.12: 108645832-108645832 on Build GRCh38

View Full Product Details