Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 159887869-159888457 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CFAP45
Entrez Gene ID
25790
Gene Name
cilia and flagella associated protein 45
Gene Aliases
CCDC19, NESG1
UniGene
Hs.647705
Gene Chromosome Location
Chr.1: 159872364-159900116 on Build GRCh38
Transcripts
NM_012337.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6261689 c.559C>A p.Q187K Chr.1: 159887870-159887870 on Build GRCh38
898087 c.367G>T p.E123* Chr.1: 159888402-159888402 on Build GRCh38
1688921 c.434G>A p.R145Q Chr.1: 159887995-159887995 on Build GRCh38
6266961 c.315C>A p.S105R Chr.1: 159888454-159888454 on Build GRCh38
898088 c.364G>T p.E122* Chr.1: 159888405-159888405 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs552434069 Chr.1: 159888235-159888235 on Build GRCh38
rs966876018 Chr.1: 159888005-159888005 on Build GRCh38
rs900419248 Chr.1: 159888245-159888245 on Build GRCh38
rs34262601 Chr.1: 159887911-159887912 on Build GRCh38
rs757774771 Chr.1: 159887975-159887975 on Build GRCh38
rs768700634 Chr.1: 159888369-159888369 on Build GRCh38
rs761608256 Chr.1: 159888019-159888019 on Build GRCh38
rs944510708 Chr.1: 159888218-159888218 on Build GRCh38
rs772282119 Chr.1: 159888322-159888323 on Build GRCh38
rs370180211 Chr.1: 159888018-159888018 on Build GRCh38
rs767207218 Chr.1: 159888059-159888059 on Build GRCh38
rs201284760 Chr.1: 159888061-159888061 on Build GRCh38
rs770971917 Chr.1: 159888399-159888399 on Build GRCh38
rs768988314 Chr.1: 159888009-159888009 on Build GRCh38
rs760450248 Chr.1: 159888431-159888431 on Build GRCh38
rs143740745 Chr.1: 159887892-159887892 on Build GRCh38
rs16842792 Chr.1: 159888239-159888239 on Build GRCh38
rs749875277 Chr.1: 159887924-159887924 on Build GRCh38
rs900330018 Chr.1: 159888198-159888198 on Build GRCh38
rs538012571 Chr.1: 159888327-159888327 on Build GRCh38
rs769547313 Chr.1: 159888384-159888384 on Build GRCh38
rs566853701 Chr.1: 159888191-159888191 on Build GRCh38
rs907743453 Chr.1: 159888090-159888090 on Build GRCh38
rs145313899 Chr.1: 159888380-159888380 on Build GRCh38
rs773041219 Chr.1: 159888394-159888394 on Build GRCh38
rs990944379 Chr.1: 159888370-159888370 on Build GRCh38
rs980095259 Chr.1: 159888069-159888069 on Build GRCh38
rs776543202 Chr.1: 159887914-159887914 on Build GRCh38
rs979557173 Chr.1: 159888036-159888036 on Build GRCh38
rs767031751 Chr.1: 159888422-159888422 on Build GRCh38
rs371697957 Chr.1: 159888053-159888053 on Build GRCh38
rs891101540 Chr.1: 159887956-159887956 on Build GRCh38
rs779579032 Chr.1: 159887977-159887977 on Build GRCh38
rs758998332 Chr.1: 159888335-159888335 on Build GRCh38
rs201664440 Chr.1: 159888437-159888437 on Build GRCh38
rs747115757 Chr.1: 159888361-159888361 on Build GRCh38
rs765162327 Chr.1: 159887922-159887922 on Build GRCh38
rs376594060 Chr.1: 159888054-159888054 on Build GRCh38
rs372597365 Chr.1: 159888419-159888419 on Build GRCh38
rs149228386 Chr.1: 159888398-159888398 on Build GRCh38
rs983392857 Chr.1: 159888086-159888086 on Build GRCh38
rs902743819 Chr.1: 159888287-159888287 on Build GRCh38
rs527619884 Chr.1: 159888221-159888221 on Build GRCh38
rs934333684 Chr.1: 159888093-159888093 on Build GRCh38
rs140567807 Chr.1: 159887995-159887995 on Build GRCh38
rs756776331 Chr.1: 159888212-159888212 on Build GRCh38
rs1008612248 Chr.1: 159888250-159888250 on Build GRCh38
rs777990509 Chr.1: 159887965-159887965 on Build GRCh38
rs757725619 Chr.1: 159888309-159888309 on Build GRCh38
rs891161208 Chr.1: 159888248-159888248 on Build GRCh38
rs751220693 Chr.1: 159888188-159888188 on Build GRCh38
rs949071956 Chr.1: 159888076-159888076 on Build GRCh38
rs866603642 Chr.1: 159887923-159887923 on Build GRCh38
rs201730535 Chr.1: 159888056-159888056 on Build GRCh38
rs754324447 Chr.1: 159887973-159887973 on Build GRCh38
rs775249902 Chr.1: 159888428-159888428 on Build GRCh38
rs941885257 Chr.1: 159888182-159888182 on Build GRCh38
rs925606839 Chr.1: 159888070-159888070 on Build GRCh38
rs188816002 Chr.1: 159888136-159888136 on Build GRCh38
rs761733171 Chr.1: 159887915-159887915 on Build GRCh38
rs200733249 Chr.1: 159887996-159887996 on Build GRCh38
rs754709147 Chr.1: 159887954-159887954 on Build GRCh38
rs375316215 Chr.1: 159888333-159888333 on Build GRCh38
rs1021210324 Chr.1: 159888275-159888275 on Build GRCh38
rs745899805 Chr.1: 159887992-159887992 on Build GRCh38
rs960809910 Chr.1: 159888359-159888359 on Build GRCh38
rs780237970 Chr.1: 159888324-159888324 on Build GRCh38
rs762475175 Chr.1: 159887928-159887928 on Build GRCh38
rs1052850540 Chr.1: 159888165-159888165 on Build GRCh38
rs750940499 Chr.1: 159888331-159888331 on Build GRCh38
rs751141465 Chr.1: 159887950-159887950 on Build GRCh38
rs763774044 Chr.1: 159887899-159887899 on Build GRCh38
rs371163694 Chr.1: 159888040-159888040 on Build GRCh38
rs766016903 Chr.1: 159887945-159887945 on Build GRCh38
rs779993381 Chr.1: 159888343-159888343 on Build GRCh38
rs532705028 Chr.1: 159888025-159888025 on Build GRCh38
rs746084264 Chr.1: 159887950-159887952 on Build GRCh38
rs996814177 Chr.1: 159887908-159887908 on Build GRCh38

View Full Product Details