Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 187345395-187345818 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CALCRL
Entrez Gene ID
10203
Gene Name
calcitonin receptor like receptor
Gene Aliases
CGRPR, CRLR
UniGene
Hs.470882
Gene Chromosome Location
Chr.2: 187341963-187448307 on Build GRCh38
Transcripts
NM_001271751.1 NM_005795.5 XM_005246231.3 XM_005246232.3 XM_005246234.3 XM_017003151.1

SNP Detail

SNP ID Chromosome Location
rs989571560 Chr.2: 187345526-187345526 on Build GRCh38
rs911336184 Chr.2: 187345782-187345782 on Build GRCh38
rs567109697 Chr.2: 187345777-187345777 on Build GRCh38
rs916666229 Chr.2: 187345534-187345534 on Build GRCh38
rs772339230 Chr.2: 187345508-187345508 on Build GRCh38
rs935166231 Chr.2: 187345640-187345640 on Build GRCh38
rs1033457668 Chr.2: 187345618-187345618 on Build GRCh38
rs958243311 Chr.2: 187345788-187345788 on Build GRCh38
rs1048236275 Chr.2: 187345683-187345683 on Build GRCh38
rs900831894 Chr.2: 187345708-187345708 on Build GRCh38
rs575365822 Chr.2: 187345689-187345690 on Build GRCh38
rs185098690 Chr.2: 187345479-187345479 on Build GRCh38
rs1031219517 Chr.2: 187345753-187345753 on Build GRCh38
rs540966266 Chr.2: 187345669-187345669 on Build GRCh38
rs1019210175 Chr.2: 187345422-187345422 on Build GRCh38
rs114131506 Chr.2: 187345676-187345676 on Build GRCh38
rs747138354 Chr.2: 187345593-187345593 on Build GRCh38
rs919927161 Chr.2: 187345699-187345699 on Build GRCh38
rs1011047208 Chr.2: 187345792-187345792 on Build GRCh38
rs777804093 Chr.2: 187345566-187345566 on Build GRCh38
rs113345773 Chr.2: 187345601-187345601 on Build GRCh38
rs948114135 Chr.2: 187345582-187345582 on Build GRCh38
rs696092 Chr.2: 187345487-187345487 on Build GRCh38
rs562793352 Chr.2: 187345529-187345529 on Build GRCh38
rs938636842 Chr.2: 187345674-187345674 on Build GRCh38
rs1056599 Chr.2: 187345791-187345791 on Build GRCh38
rs1041131101 Chr.2: 187345700-187345700 on Build GRCh38
rs992316250 Chr.2: 187345670-187345670 on Build GRCh38
rs966044748 Chr.2: 187345444-187345444 on Build GRCh38
rs533027963 Chr.2: 187345530-187345530 on Build GRCh38
rs555431820 Chr.2: 187345689-187345689 on Build GRCh38
rs777166013 Chr.2: 187345784-187345784 on Build GRCh38
rs145758812 Chr.2: 187345478-187345478 on Build GRCh38
rs927969152 Chr.2: 187345666-187345666 on Build GRCh38
rs537837641 Chr.2: 187345790-187345790 on Build GRCh38
rs5837031 Chr.2: 187345688-187345689 on Build GRCh38
rs16828912 Chr.2: 187345679-187345679 on Build GRCh38
rs527352935 Chr.2: 187345677-187345677 on Build GRCh38
rs1049789830 Chr.2: 187345759-187345759 on Build GRCh38
rs376576168 Chr.2: 187345711-187345711 on Build GRCh38

View Full Product Details