Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 98717325-98717857 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C14orf177
Entrez Gene ID
283598
Gene Name
chromosome 14 open reading frame 177
Gene Aliases
-
UniGene
Hs.448754
Gene Chromosome Location
Chr.14: 98711613-98717766 on Build GRCh38
Transcripts
NM_182560.2

SNP Detail

SNP ID Chromosome Location
rs541760327 Chr.14: 98717386-98717386 on Build GRCh38
rs907127006 Chr.14: 98717670-98717670 on Build GRCh38
rs933826059 Chr.14: 98717363-98717363 on Build GRCh38
rs553515152 Chr.14: 98717457-98717457 on Build GRCh38
rs550761886 Chr.14: 98717762-98717762 on Build GRCh38
rs1001705936 Chr.14: 98717460-98717460 on Build GRCh38
rs955324817 Chr.14: 98717751-98717752 on Build GRCh38
rs936082058 Chr.14: 98717618-98717618 on Build GRCh38
rs922049201 Chr.14: 98717358-98717358 on Build GRCh38
rs114525320 Chr.14: 98717566-98717566 on Build GRCh38
rs985982775 Chr.14: 98717375-98717375 on Build GRCh38
rs201500806 Chr.14: 98717627-98717627 on Build GRCh38
rs1029702855 Chr.14: 98717735-98717735 on Build GRCh38
rs540930088 Chr.14: 98717568-98717568 on Build GRCh38
rs568926032 Chr.14: 98717822-98717822 on Build GRCh38
rs1011310194 Chr.14: 98717536-98717536 on Build GRCh38
rs560965840 Chr.14: 98717626-98717626 on Build GRCh38
rs1036567899 Chr.14: 98717633-98717633 on Build GRCh38
rs532248817 Chr.14: 98717698-98717698 on Build GRCh38
rs768094939 Chr.14: 98717489-98717489 on Build GRCh38
rs921935773 Chr.14: 98717590-98717590 on Build GRCh38
rs1055976022 Chr.14: 98717673-98717673 on Build GRCh38
rs536374720 Chr.14: 98717834-98717834 on Build GRCh38
rs191538482 Chr.14: 98717679-98717679 on Build GRCh38
rs966660957 Chr.14: 98717576-98717576 on Build GRCh38
rs907092300 Chr.14: 98717442-98717442 on Build GRCh38
rs993845938 Chr.14: 98717718-98717718 on Build GRCh38
rs946296352 Chr.14: 98717415-98717415 on Build GRCh38
rs981470053 Chr.14: 98717835-98717835 on Build GRCh38
rs894267415 Chr.14: 98717527-98717527 on Build GRCh38
rs760935559 Chr.14: 98717636-98717636 on Build GRCh38
rs907450478 Chr.14: 98717665-98717665 on Build GRCh38
rs967824829 Chr.14: 98717833-98717833 on Build GRCh38
rs74626923 Chr.14: 98717569-98717569 on Build GRCh38
rs913314189 Chr.14: 98717806-98717806 on Build GRCh38
rs370057485 Chr.14: 98717540-98717540 on Build GRCh38
rs146991324 Chr.14: 98717380-98717380 on Build GRCh38
rs1038067765 Chr.14: 98717702-98717702 on Build GRCh38
rs937552360 Chr.14: 98717671-98717671 on Build GRCh38
rs891771597 Chr.14: 98717414-98717414 on Build GRCh38
rs896860482 Chr.14: 98717709-98717709 on Build GRCh38
rs867170346 Chr.14: 98717567-98717567 on Build GRCh38
rs957171057 Chr.14: 98717786-98717786 on Build GRCh38
rs911022285 Chr.14: 98717607-98717607 on Build GRCh38
rs1055487191 Chr.14: 98717787-98717787 on Build GRCh38
rs1045962591 Chr.14: 98717419-98717420 on Build GRCh38
rs542586427 Chr.14: 98717608-98717608 on Build GRCh38
rs1002546923 Chr.14: 98717462-98717462 on Build GRCh38
rs573531445 Chr.14: 98717554-98717554 on Build GRCh38
rs976144137 Chr.14: 98717580-98717580 on Build GRCh38
rs1004788510 Chr.14: 98717406-98717406 on Build GRCh38
rs764286354 Chr.14: 98717838-98717838 on Build GRCh38
rs1046440329 Chr.14: 98717634-98717634 on Build GRCh38
rs546608012 Chr.14: 98717629-98717629 on Build GRCh38
rs188316147 Chr.14: 98717369-98717369 on Build GRCh38
rs972647834 Chr.14: 98717358-98717358 on Build GRCh38
rs1005276777 Chr.14: 98717693-98717693 on Build GRCh38
rs1036297177 Chr.14: 98717433-98717433 on Build GRCh38
rs932091941 Chr.14: 98717591-98717591 on Build GRCh38
rs1031254268 Chr.14: 98717573-98717573 on Build GRCh38
rs554608145 Chr.14: 98717372-98717372 on Build GRCh38
rs932060846 Chr.14: 98717364-98717364 on Build GRCh38
rs545479596 Chr.14: 98717546-98717546 on Build GRCh38
rs1018138269 Chr.14: 98717781-98717781 on Build GRCh38
rs1034421838 Chr.14: 98717491-98717491 on Build GRCh38

View Full Product Details