Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 12663787-12664386 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CPPED1
Entrez Gene ID
55313
Gene Name
calcineurin like phosphoesterase domain containing 1
Gene Aliases
CSTP1
UniGene
Hs.460002
Gene Chromosome Location
Chr.16: 12659799-12803887 on Build GRCh38
Transcripts
NM_001099455.1 NM_018340.2

SNP Detail

SNP ID Chromosome Location
rs550710295 Chr.16: 12664172-12664172 on Build GRCh38
rs903970862 Chr.16: 12664197-12664197 on Build GRCh38
rs746889677 Chr.16: 12663901-12663901 on Build GRCh38
rs951811601 Chr.16: 12664353-12664353 on Build GRCh38
rs776742795 Chr.16: 12664159-12664159 on Build GRCh38
rs190176690 Chr.16: 12664104-12664104 on Build GRCh38
rs959676841 Chr.16: 12663987-12663987 on Build GRCh38
rs968411780 Chr.16: 12664247-12664247 on Build GRCh38
rs560403693 Chr.16: 12663952-12663952 on Build GRCh38
rs1042469444 Chr.16: 12664189-12664189 on Build GRCh38
rs529760253 Chr.16: 12663942-12663942 on Build GRCh38
rs977150783 Chr.16: 12664253-12664253 on Build GRCh38
rs959851608 Chr.16: 12664258-12664258 on Build GRCh38
rs949749365 Chr.16: 12664040-12664040 on Build GRCh38
rs1021291495 Chr.16: 12664220-12664220 on Build GRCh38
rs916925384 Chr.16: 12664039-12664039 on Build GRCh38
rs754829259 Chr.16: 12663921-12663921 on Build GRCh38
rs187750846 Chr.16: 12664219-12664219 on Build GRCh38
rs921446429 Chr.16: 12664303-12664303 on Build GRCh38
rs373594166 Chr.16: 12663931-12663931 on Build GRCh38
rs1044961 Chr.16: 12664144-12664144 on Build GRCh38
rs866183161 Chr.16: 12663966-12663966 on Build GRCh38
rs185850405 Chr.16: 12663962-12663962 on Build GRCh38
rs1014728549 Chr.16: 12663854-12663854 on Build GRCh38
rs1035545376 Chr.16: 12664168-12664168 on Build GRCh38
rs544357281 Chr.16: 12663881-12663881 on Build GRCh38
rs745992344 Chr.16: 12664244-12664244 on Build GRCh38
rs971072708 Chr.16: 12664321-12664321 on Build GRCh38
rs568747729 Chr.16: 12664090-12664090 on Build GRCh38
rs373088044 Chr.16: 12664249-12664249 on Build GRCh38
rs1003089169 Chr.16: 12664150-12664150 on Build GRCh38
rs560942390 Chr.16: 12663895-12663895 on Build GRCh38
rs192012939 Chr.16: 12664311-12664311 on Build GRCh38
rs141322829 Chr.16: 12664276-12664276 on Build GRCh38
rs774406472 Chr.16: 12664143-12664143 on Build GRCh38
rs894355946 Chr.16: 12664187-12664187 on Build GRCh38
rs369435999 Chr.16: 12664349-12664349 on Build GRCh38
rs1021302714 Chr.16: 12663939-12663939 on Build GRCh38
rs1004258191 Chr.16: 12663853-12663853 on Build GRCh38
rs899813619 Chr.16: 12663820-12663820 on Build GRCh38
rs977203167 Chr.16: 12663948-12663948 on Build GRCh38
rs769522497 Chr.16: 12663916-12663917 on Build GRCh38
rs771235336 Chr.16: 12664106-12664106 on Build GRCh38
rs1031867320 Chr.16: 12664267-12664267 on Build GRCh38
rs1034328571 Chr.16: 12663967-12663967 on Build GRCh38
rs991128401 Chr.16: 12663902-12663902 on Build GRCh38
rs962221829 Chr.16: 12663863-12663863 on Build GRCh38
rs796727163 Chr.16: 12664230-12664230 on Build GRCh38
rs889022374 Chr.16: 12664137-12664137 on Build GRCh38
rs148251425 Chr.16: 12663988-12663988 on Build GRCh38
rs771997384 Chr.16: 12664348-12664348 on Build GRCh38
rs1002772800 Chr.16: 12663879-12663879 on Build GRCh38
rs1048395894 Chr.16: 12663832-12663832 on Build GRCh38
rs901856629 Chr.16: 12664169-12664169 on Build GRCh38
rs552366997 Chr.16: 12664013-12664013 on Build GRCh38
rs182670678 Chr.16: 12664186-12664186 on Build GRCh38
rs770839558 Chr.16: 12663852-12663852 on Build GRCh38
rs59305865 Chr.16: 12664289-12664289 on Build GRCh38
rs375668646 Chr.16: 12664045-12664045 on Build GRCh38
rs996832199 Chr.16: 12663822-12663822 on Build GRCh38
rs924801270 Chr.16: 12663969-12663969 on Build GRCh38
rs546266714 Chr.16: 12663944-12663944 on Build GRCh38
rs944228075 Chr.16: 12663981-12663981 on Build GRCh38
rs55815325 Chr.16: 12664093-12664093 on Build GRCh38
rs932923895 Chr.16: 12664060-12664060 on Build GRCh38
rs1048916713 Chr.16: 12664116-12664116 on Build GRCh38

View Full Product Details