Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 15581189-15581766 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C16orf45
Entrez Gene ID
89927
Gene Name
chromosome 16 open reading frame 45
Gene Aliases
MINP
UniGene
Hs.401798
Gene Chromosome Location
Chr.16: 15434468-15594521 on Build GRCh38
Transcripts
NM_001142469.1 NM_033201.2 XM_011522731.2 XM_017023824.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4853413 c.357G>T p.K119N Chr.16: 15581269-15581269 on Build GRCh38
5953484 c.319G>A p.V107M Chr.16: 15581282-15581282 on Build GRCh38
2126765 c.310G>A p.E104K Chr.16: 15581222-15581222 on Build GRCh38
5913736 c.254-8C>T p.? Chr.16: 15581209-15581209 on Build GRCh38
4256306 c.259G>A p.E87K Chr.16: 15581222-15581222 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs202099414 Chr.16: 15581372-15581372 on Build GRCh38
rs200139620 Chr.16: 15581312-15581312 on Build GRCh38
rs757365966 Chr.16: 15581229-15581229 on Build GRCh38
rs756055074 Chr.16: 15581345-15581345 on Build GRCh38
rs887043375 Chr.16: 15581692-15581692 on Build GRCh38
rs555711229 Chr.16: 15581667-15581667 on Build GRCh38
rs984375635 Chr.16: 15581410-15581410 on Build GRCh38
rs753437093 Chr.16: 15581299-15581299 on Build GRCh38
rs1015329885 Chr.16: 15581437-15581437 on Build GRCh38
rs567648444 Chr.16: 15581311-15581311 on Build GRCh38
rs978082441 Chr.16: 15581604-15581604 on Build GRCh38
rs761331776 Chr.16: 15581484-15581484 on Build GRCh38
rs536426347 Chr.16: 15581567-15581567 on Build GRCh38
rs751708436 Chr.16: 15581344-15581344 on Build GRCh38
rs201702585 Chr.16: 15581222-15581222 on Build GRCh38
rs777659710 Chr.16: 15581276-15581276 on Build GRCh38
rs868544146 Chr.16: 15581299-15581299 on Build GRCh38
rs769145064 Chr.16: 15581374-15581374 on Build GRCh38
rs951298057 Chr.16: 15581366-15581366 on Build GRCh38
rs922297553 Chr.16: 15581442-15581442 on Build GRCh38
rs917789846 Chr.16: 15581580-15581580 on Build GRCh38
rs763726131 Chr.16: 15581222-15581222 on Build GRCh38
rs778401935 Chr.16: 15581358-15581358 on Build GRCh38
rs573646330 Chr.16: 15581618-15581618 on Build GRCh38
rs1016744938 Chr.16: 15581435-15581435 on Build GRCh38
rs1005449288 Chr.16: 15581700-15581700 on Build GRCh38
rs1012311993 Chr.16: 15581340-15581340 on Build GRCh38
rs550156788 Chr.16: 15581320-15581320 on Build GRCh38
rs184963421 Chr.16: 15581362-15581362 on Build GRCh38
rs774925174 Chr.16: 15581363-15581363 on Build GRCh38
rs543366351 Chr.16: 15581263-15581263 on Build GRCh38
rs139423361 Chr.16: 15581515-15581515 on Build GRCh38
rs779198476 Chr.16: 15581242-15581242 on Build GRCh38
rs922130107 Chr.16: 15581651-15581651 on Build GRCh38
rs935506446 Chr.16: 15581293-15581293 on Build GRCh38
rs571669961 Chr.16: 15581324-15581324 on Build GRCh38
rs932101392 Chr.16: 15581666-15581666 on Build GRCh38
rs370345498 Chr.16: 15581375-15581375 on Build GRCh38
rs1017209342 Chr.16: 15581710-15581710 on Build GRCh38
rs936755246 Chr.16: 15581623-15581623 on Build GRCh38
rs367722389 Chr.16: 15581215-15581215 on Build GRCh38
rs549188506 Chr.16: 15581256-15581256 on Build GRCh38
rs975148099 Chr.16: 15581438-15581438 on Build GRCh38
rs906589087 Chr.16: 15581665-15581665 on Build GRCh38
rs757403045 Chr.16: 15581490-15581490 on Build GRCh38
rs371464387 Chr.16: 15581313-15581313 on Build GRCh38
rs756446865 Chr.16: 15581576-15581576 on Build GRCh38
rs750705222 Chr.16: 15581326-15581326 on Build GRCh38
rs761001019 Chr.16: 15581217-15581217 on Build GRCh38
rs758950219 Chr.16: 15581308-15581308 on Build GRCh38
rs534762493 Chr.16: 15581570-15581570 on Build GRCh38
rs753255088 Chr.16: 15581216-15581216 on Build GRCh38
rs370851979 Chr.16: 15581342-15581342 on Build GRCh38
rs749025474 Chr.16: 15581352-15581352 on Build GRCh38
rs200068450 Chr.16: 15581261-15581261 on Build GRCh38
rs145141186 Chr.16: 15581314-15581314 on Build GRCh38
rs36049896 Chr.16: 15581484-15581485 on Build GRCh38
rs774113668 Chr.16: 15581302-15581302 on Build GRCh38
rs750463940 Chr.16: 15581251-15581251 on Build GRCh38
rs577234139 Chr.16: 15581670-15581670 on Build GRCh38
rs867863719 Chr.16: 15581297-15581298 on Build GRCh38
rs900642638 Chr.16: 15581333-15581333 on Build GRCh38
rs781252753 Chr.16: 15581270-15581270 on Build GRCh38
rs748132650 Chr.16: 15581272-15581272 on Build GRCh38
rs8049274 Chr.16: 15581648-15581648 on Build GRCh38
rs769879938 Chr.16: 15581274-15581274 on Build GRCh38
rs990761491 Chr.16: 15581572-15581572 on Build GRCh38
rs1052062586 Chr.16: 15581701-15581701 on Build GRCh38
rs890693630 Chr.16: 15581714-15581714 on Build GRCh38
rs544638947 Chr.16: 15581733-15581733 on Build GRCh38
rs970794273 Chr.16: 15581601-15581601 on Build GRCh38
rs192461391 Chr.16: 15581228-15581228 on Build GRCh38
rs758521242 Chr.16: 15581339-15581339 on Build GRCh38
rs541826325 Chr.16: 15581361-15581361 on Build GRCh38
rs201187962 Chr.16: 15581284-15581284 on Build GRCh38
rs777767775 Chr.16: 15581349-15581349 on Build GRCh38
rs1047024140 Chr.16: 15581672-15581672 on Build GRCh38
rs749119469 Chr.16: 15581277-15581277 on Build GRCh38
rs1007846264 Chr.16: 15581378-15581378 on Build GRCh38
rs566078362 Chr.16: 15581506-15581506 on Build GRCh38
rs749305611 Chr.16: 15581450-15581450 on Build GRCh38
rs150722366 Chr.16: 15581353-15581353 on Build GRCh38
rs893959299 Chr.16: 15581597-15581597 on Build GRCh38

View Full Product Details