Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 3636871-3637356 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CTNS
Entrez Gene ID
1497
Gene Name
cystinosin, lysosomal cystine transporter
Gene Aliases
CTNS-LSB, PQLC4
UniGene
Hs.187667
Gene Chromosome Location
Chr.17: 3636391-3663103 on Build GRCh38
Transcripts
NM_001031681.2 NM_004937.2 XM_005256485.2 XM_006721463.2 XM_006721464.2 XM_011523691.2 XM_011523692.2 XM_017024254.1 XM_017024255.1 XM_017024256.1 XM_017024257.1 XM_017024258.1

SNP Detail

SNP ID Chromosome Location
rs867822900 Chr.17: 3637245-3637245 on Build GRCh38
rs956975285 Chr.17: 3637022-3637022 on Build GRCh38
rs774549023 Chr.17: 3637185-3637185 on Build GRCh38
rs550436044 Chr.17: 3637313-3637314 on Build GRCh38
rs771868957 Chr.17: 3637280-3637280 on Build GRCh38
rs956882439 Chr.17: 3637254-3637254 on Build GRCh38
rs749550322 Chr.17: 3637087-3637087 on Build GRCh38
rs780032861 Chr.17: 3637329-3637329 on Build GRCh38
rs886052863 Chr.17: 3637200-3637200 on Build GRCh38
rs989658111 Chr.17: 3637261-3637261 on Build GRCh38
rs886052867 Chr.17: 3637315-3637316 on Build GRCh38
rs948057294 Chr.17: 3637090-3637090 on Build GRCh38
rs534881680 Chr.17: 3637184-3637184 on Build GRCh38
rs1049455294 Chr.17: 3637189-3637189 on Build GRCh38
rs886052865 Chr.17: 3637289-3637289 on Build GRCh38
rs780257774 Chr.17: 3636982-3636982 on Build GRCh38
rs896088954 Chr.17: 3637264-3637265 on Build GRCh38
rs1052975634 Chr.17: 3637135-3637135 on Build GRCh38
rs1025799797 Chr.17: 3637284-3637284 on Build GRCh38
rs980840947 Chr.17: 3637121-3637121 on Build GRCh38
rs952875122 Chr.17: 3637312-3637312 on Build GRCh38
rs915217536 Chr.17: 3637075-3637075 on Build GRCh38
rs745696012 Chr.17: 3637203-3637203 on Build GRCh38
rs989708909 Chr.17: 3637038-3637038 on Build GRCh38
rs933203798 Chr.17: 3637131-3637131 on Build GRCh38
rs998290387 Chr.17: 3637242-3637242 on Build GRCh38
rs778786655 Chr.17: 3637146-3637146 on Build GRCh38
rs536787781 Chr.17: 3636978-3636978 on Build GRCh38
rs976921752 Chr.17: 3636985-3636985 on Build GRCh38
rs960536214 Chr.17: 3636911-3636911 on Build GRCh38
rs1027349375 Chr.17: 3636921-3636921 on Build GRCh38
rs558096619 Chr.17: 3637324-3637324 on Build GRCh38
rs888747156 Chr.17: 3637190-3637190 on Build GRCh38
rs905565378 Chr.17: 3637228-3637228 on Build GRCh38
rs1019110034 Chr.17: 3636979-3636979 on Build GRCh38
rs939161326 Chr.17: 3637152-3637152 on Build GRCh38
rs541943702 Chr.17: 3637031-3637038 on Build GRCh38
rs1036227713 Chr.17: 3637158-3637158 on Build GRCh38
rs566880414 Chr.17: 3637101-3637101 on Build GRCh38
rs886052864 Chr.17: 3637262-3637262 on Build GRCh38
rs897729691 Chr.17: 3637170-3637170 on Build GRCh38
rs1018687737 Chr.17: 3637220-3637220 on Build GRCh38
rs886052866 Chr.17: 3637312-3637312 on Build GRCh38
rs930526453 Chr.17: 3637177-3637177 on Build GRCh38
rs984157539 Chr.17: 3637030-3637030 on Build GRCh38
rs537567463 Chr.17: 3637123-3637123 on Build GRCh38
rs901628964 Chr.17: 3637232-3637232 on Build GRCh38
rs372649305 Chr.17: 3637021-3637021 on Build GRCh38
rs1007274196 Chr.17: 3636971-3636971 on Build GRCh38
rs888790193 Chr.17: 3636961-3636961 on Build GRCh38
rs964012856 Chr.17: 3637037-3637037 on Build GRCh38

View Full Product Details