Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 34575222-34575757 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C17orf102
Entrez Gene ID
400591
Gene Name
chromosome 17 open reading frame 102
Gene Aliases
-
UniGene
Hs.514090
Gene Chromosome Location
Chr.17: 34574123-34579369 on Build GRCh38
Transcripts
NM_207454.2

SNP Detail

SNP ID Chromosome Location
rs931651500 Chr.17: 34575522-34575522 on Build GRCh38
rs892418579 Chr.17: 34575340-34575340 on Build GRCh38
rs966146681 Chr.17: 34575377-34575377 on Build GRCh38
rs962981726 Chr.17: 34575466-34575466 on Build GRCh38
rs979088875 Chr.17: 34575413-34575413 on Build GRCh38
rs559230642 Chr.17: 34575543-34575543 on Build GRCh38
rs201390810 Chr.17: 34575678-34575678 on Build GRCh38
rs1002182204 Chr.17: 34575412-34575412 on Build GRCh38
rs1014250 Chr.17: 34575608-34575608 on Build GRCh38
rs368929872 Chr.17: 34575299-34575299 on Build GRCh38
rs1037403952 Chr.17: 34575258-34575260 on Build GRCh38
rs775884377 Chr.17: 34575635-34575635 on Build GRCh38
rs967673445 Chr.17: 34575409-34575409 on Build GRCh38
rs538370685 Chr.17: 34575453-34575453 on Build GRCh38
rs1010672563 Chr.17: 34575624-34575624 on Build GRCh38
rs890180244 Chr.17: 34575285-34575285 on Build GRCh38
rs913687232 Chr.17: 34575405-34575405 on Build GRCh38
rs995542391 Chr.17: 34575298-34575298 on Build GRCh38
rs550184722 Chr.17: 34575557-34575557 on Build GRCh38
rs185322984 Chr.17: 34575275-34575275 on Build GRCh38
rs765179405 Chr.17: 34575421-34575421 on Build GRCh38
rs1049777303 Chr.17: 34575577-34575577 on Build GRCh38
rs761656530 Chr.17: 34575399-34575399 on Build GRCh38
rs185266959 Chr.17: 34575245-34575245 on Build GRCh38
rs565114355 Chr.17: 34575602-34575602 on Build GRCh38
rs146448910 Chr.17: 34575734-34575734 on Build GRCh38
rs1008688947 Chr.17: 34575302-34575302 on Build GRCh38
rs746408535 Chr.17: 34575574-34575574 on Build GRCh38
rs538955419 Chr.17: 34575324-34575324 on Build GRCh38
rs1020640097 Chr.17: 34575704-34575704 on Build GRCh38
rs1056669465 Chr.17: 34575468-34575468 on Build GRCh38
rs547323222 Chr.17: 34575279-34575279 on Build GRCh38
rs974709771 Chr.17: 34575469-34575469 on Build GRCh38
rs189798370 Chr.17: 34575248-34575248 on Build GRCh38
rs554609144 Chr.17: 34575458-34575458 on Build GRCh38
rs908702317 Chr.17: 34575436-34575436 on Build GRCh38
rs190179666 Chr.17: 34575687-34575687 on Build GRCh38
rs570964942 Chr.17: 34575375-34575375 on Build GRCh38
rs1035024897 Chr.17: 34575728-34575728 on Build GRCh38
rs1008789099 Chr.17: 34575353-34575353 on Build GRCh38
rs181998982 Chr.17: 34575266-34575266 on Build GRCh38
rs574513308 Chr.17: 34575510-34575510 on Build GRCh38
rs141760122 Chr.17: 34575435-34575436 on Build GRCh38
rs890007286 Chr.17: 34575592-34575592 on Build GRCh38
rs544970532 Chr.17: 34575569-34575569 on Build GRCh38
rs905074969 Chr.17: 34575677-34575677 on Build GRCh38
rs550719670 Chr.17: 34575669-34575669 on Build GRCh38
rs751708836 Chr.17: 34575548-34575548 on Build GRCh38
rs573130041 Chr.17: 34575553-34575553 on Build GRCh38
rs559153147 Chr.17: 34575403-34575403 on Build GRCh38
rs1036996642 Chr.17: 34575288-34575288 on Build GRCh38
rs898810622 Chr.17: 34575274-34575274 on Build GRCh38
rs549901027 Chr.17: 34575429-34575429 on Build GRCh38
rs941590731 Chr.17: 34575254-34575254 on Build GRCh38
rs567114974 Chr.17: 34575322-34575322 on Build GRCh38
rs969814198 Chr.17: 34575397-34575397 on Build GRCh38
rs1037112695 Chr.17: 34575544-34575544 on Build GRCh38
rs540187646 Chr.17: 34575539-34575539 on Build GRCh38
rs1056316387 Chr.17: 34575264-34575264 on Build GRCh38
rs898498496 Chr.17: 34575296-34575296 on Build GRCh38
rs956224863 Chr.17: 34575684-34575684 on Build GRCh38

View Full Product Details