Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 80188142-80188642 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CARD14
Entrez Gene ID
79092
Gene Name
caspase recruitment domain family member 14
Gene Aliases
BIMP2, CARMA2, PRP, PSORS2, PSS1
UniGene
Hs.675480
Gene Chromosome Location
Chr.17: 80169992-80209331 on Build GRCh38
Transcripts
NM_001257970.1 NM_024110.4 NM_052819.2 NR_047566.1 XM_011525212.1 XM_011525213.1 XM_011525215.1 XM_011525216.1 XM_011525217.1 XM_011525218.2 XM_017025057.1 XM_017025058.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4070402 c.710A>G p.N237S Chr.17: 80188411-80188411 on Build GRCh38
6620101 c.66C>T p.S22S Chr.17: 80188478-80188478 on Build GRCh38
3523313 c.732G>A p.L244L Chr.17: 80188433-80188433 on Build GRCh38
6082617 c.753G>C p.L251L Chr.17: 80188454-80188454 on Build GRCh38
5853722 c.97G>A p.E33K Chr.17: 80188509-80188509 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs888843015 Chr.17: 80188243-80188243 on Build GRCh38
rs774074787 Chr.17: 80188493-80188493 on Build GRCh38
rs768758745 Chr.17: 80188463-80188463 on Build GRCh38
rs747175055 Chr.17: 80188369-80188369 on Build GRCh38
rs746999591 Chr.17: 80188460-80188460 on Build GRCh38
rs372114482 Chr.17: 80188367-80188367 on Build GRCh38
rs765773292 Chr.17: 80188345-80188345 on Build GRCh38
rs971020345 Chr.17: 80188416-80188416 on Build GRCh38
rs781587949 Chr.17: 80188339-80188339 on Build GRCh38
rs770433643 Chr.17: 80188490-80188490 on Build GRCh38
rs754420302 Chr.17: 80188407-80188407 on Build GRCh38
rs757727838 Chr.17: 80188361-80188361 on Build GRCh38
rs200596848 Chr.17: 80188373-80188373 on Build GRCh38
rs750856042 Chr.17: 80188443-80188443 on Build GRCh38
rs200837077 Chr.17: 80188395-80188395 on Build GRCh38
rs1017397504 Chr.17: 80188581-80188581 on Build GRCh38
rs142246283 Chr.17: 80188384-80188384 on Build GRCh38
rs140720686 Chr.17: 80188421-80188421 on Build GRCh38
rs765237261 Chr.17: 80188504-80188506 on Build GRCh38
rs940850652 Chr.17: 80188505-80188505 on Build GRCh38
rs138616955 Chr.17: 80188497-80188497 on Build GRCh38
rs748988466 Chr.17: 80188561-80188561 on Build GRCh38
rs778437466 Chr.17: 80188359-80188359 on Build GRCh38
rs1031993269 Chr.17: 80188266-80188266 on Build GRCh38
rs764656786 Chr.17: 80188404-80188404 on Build GRCh38
rs752056613 Chr.17: 80188199-80188199 on Build GRCh38
rs758115256 Chr.17: 80188313-80188313 on Build GRCh38
rs377340180 Chr.17: 80188555-80188555 on Build GRCh38
rs1051608633 Chr.17: 80188232-80188232 on Build GRCh38
rs569395465 Chr.17: 80188585-80188585 on Build GRCh38
rs750958441 Chr.17: 80188590-80188590 on Build GRCh38
rs762506583 Chr.17: 80188394-80188394 on Build GRCh38
rs760262523 Chr.17: 80188588-80188588 on Build GRCh38
rs765778353 Chr.17: 80188589-80188589 on Build GRCh38
rs772988369 Chr.17: 80188479-80188479 on Build GRCh38
rs1007743886 Chr.17: 80188258-80188258 on Build GRCh38
rs750094571 Chr.17: 80188308-80188308 on Build GRCh38
rs752302933 Chr.17: 80188524-80188524 on Build GRCh38
rs751778974 Chr.17: 80188534-80188534 on Build GRCh38
rs28674001 Chr.17: 80188371-80188371 on Build GRCh38
rs373638168 Chr.17: 80188572-80188572 on Build GRCh38
rs140965756 Chr.17: 80188280-80188280 on Build GRCh38
rs150424747 Chr.17: 80188529-80188529 on Build GRCh38
rs759203476 Chr.17: 80188573-80188573 on Build GRCh38
rs369755459 Chr.17: 80188382-80188382 on Build GRCh38
rs201478315 Chr.17: 80188360-80188360 on Build GRCh38
rs149318654 Chr.17: 80188528-80188528 on Build GRCh38
rs961495025 Chr.17: 80188597-80188597 on Build GRCh38
rs1049220201 Chr.17: 80188255-80188255 on Build GRCh38
rs746067298 Chr.17: 80188366-80188366 on Build GRCh38
rs748295383 Chr.17: 80188553-80188553 on Build GRCh38
rs367767898 Chr.17: 80188554-80188554 on Build GRCh38
rs117360605 Chr.17: 80188525-80188525 on Build GRCh38
rs1005579004 Chr.17: 80188283-80188283 on Build GRCh38
rs933242599 Chr.17: 80188431-80188431 on Build GRCh38
rs199561010 Chr.17: 80188347-80188347 on Build GRCh38
rs756564250 Chr.17: 80188348-80188348 on Build GRCh38
rs770790125 Chr.17: 80188563-80188563 on Build GRCh38
rs775431566 Chr.17: 80188368-80188368 on Build GRCh38
rs865784001 Chr.17: 80188420-80188420 on Build GRCh38
rs780324929 Chr.17: 80188459-80188459 on Build GRCh38
rs937390955 Chr.17: 80188223-80188223 on Build GRCh38
rs761804037 Chr.17: 80188436-80188437 on Build GRCh38
rs781292154 Chr.17: 80188466-80188466 on Build GRCh38
rs150138223 Chr.17: 80188449-80188449 on Build GRCh38
rs781238013 Chr.17: 80188543-80188543 on Build GRCh38
rs767104415 Chr.17: 80188498-80188498 on Build GRCh38
rs927392856 Chr.17: 80188218-80188218 on Build GRCh38
rs959431928 Chr.17: 80188368-80188369 on Build GRCh38
rs563102415 Chr.17: 80188583-80188583 on Build GRCh38
rs1009613472 Chr.17: 80188508-80188508 on Build GRCh38
rs143622685 Chr.17: 80188560-80188560 on Build GRCh38
rs776733187 Chr.17: 80188433-80188433 on Build GRCh38
rs555281512 Chr.17: 80188447-80188447 on Build GRCh38
rs773193211 Chr.17: 80188557-80188557 on Build GRCh38
rs755246184 Chr.17: 80188541-80188541 on Build GRCh38
rs1032218958 Chr.17: 80188376-80188376 on Build GRCh38
rs768424345 Chr.17: 80188370-80188370 on Build GRCh38
rs1009423742 Chr.17: 80188612-80188612 on Build GRCh38
rs1045819106 Chr.17: 80188176-80188176 on Build GRCh38
rs114218658 Chr.17: 80188410-80188410 on Build GRCh38
rs901529037 Chr.17: 80188343-80188343 on Build GRCh38
rs769783308 Chr.17: 80188478-80188478 on Build GRCh38
rs530546975 Chr.17: 80188584-80188584 on Build GRCh38
rs557035385 Chr.17: 80188400-80188400 on Build GRCh38
rs369763813 Chr.17: 80188362-80188362 on Build GRCh38
rs1036603152 Chr.17: 80188301-80188301 on Build GRCh38
rs762812663 Chr.17: 80188483-80188483 on Build GRCh38
rs144570537 Chr.17: 80188467-80188467 on Build GRCh38
rs750975292 Chr.17: 80188399-80188399 on Build GRCh38

View Full Product Details