Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 42789500-42790056 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM216B
Entrez Gene ID
144809
Gene Name
family with sequence similarity 216 member B
Gene Aliases
C13orf30
UniGene
Hs.16970
Gene Chromosome Location
Chr.13: 42781550-42791549 on Build GRCh38
Transcripts
NM_001318932.1 NM_182508.2

SNP Detail

SNP ID Chromosome Location
rs67999777 Chr.13: 42789637-42789644 on Build GRCh38
rs757595586 Chr.13: 42789557-42789557 on Build GRCh38
rs61051465 Chr.13: 42789662-42789669 on Build GRCh38
rs948336606 Chr.13: 42790011-42790011 on Build GRCh38
rs753688634 Chr.13: 42789873-42789873 on Build GRCh38
rs751907755 Chr.13: 42789536-42789536 on Build GRCh38
rs1008044332 Chr.13: 42789711-42789711 on Build GRCh38
rs1004292493 Chr.13: 42789771-42789771 on Build GRCh38
rs193064175 Chr.13: 42789999-42789999 on Build GRCh38
rs992380137 Chr.13: 42789981-42789981 on Build GRCh38
rs879083200 Chr.13: 42789637-42789642 on Build GRCh38
rs34058453 Chr.13: 42789671-42789671 on Build GRCh38
rs537792433 Chr.13: 42789587-42789587 on Build GRCh38
rs559243389 Chr.13: 42790010-42790010 on Build GRCh38
rs554429586 Chr.13: 42789657-42789657 on Build GRCh38
rs1038075133 Chr.13: 42789582-42789582 on Build GRCh38
rs112183910 Chr.13: 42789716-42789716 on Build GRCh38
rs944364327 Chr.13: 42789693-42789693 on Build GRCh38
rs1022298486 Chr.13: 42789858-42789858 on Build GRCh38
rs1049393901 Chr.13: 42789740-42789740 on Build GRCh38
rs548536432 Chr.13: 42789943-42789943 on Build GRCh38
rs750659315 Chr.13: 42789585-42789585 on Build GRCh38
rs77800748 Chr.13: 42789707-42789707 on Build GRCh38
rs916729601 Chr.13: 42790001-42790001 on Build GRCh38
rs553684401 Chr.13: 42789958-42789958 on Build GRCh38
rs77982642 Chr.13: 42789745-42789745 on Build GRCh38
rs576700755 Chr.13: 42789891-42789891 on Build GRCh38
rs900438367 Chr.13: 42789699-42789699 on Build GRCh38
rs977804802 Chr.13: 42789966-42789966 on Build GRCh38
rs539269220 Chr.13: 42789901-42789901 on Build GRCh38
rs7329140 Chr.13: 42789959-42789959 on Build GRCh38
rs780175245 Chr.13: 42789824-42789824 on Build GRCh38
rs199533129 Chr.13: 42789646-42789646 on Build GRCh38
rs59003170 Chr.13: 42789669-42789670 on Build GRCh38
rs140175847 Chr.13: 42789637-42789646 on Build GRCh38
rs76442258 Chr.13: 42789667-42789667 on Build GRCh38
rs769862998 Chr.13: 42789639-42789646 on Build GRCh38
rs978185544 Chr.13: 42789860-42789860 on Build GRCh38
rs3223409 Chr.13: 42789667-42789668 on Build GRCh38
rs993879279 Chr.13: 42789619-42789619 on Build GRCh38
rs754990450 Chr.13: 42789933-42789933 on Build GRCh38
rs767662262 Chr.13: 42789577-42789577 on Build GRCh38
rs1038914443 Chr.13: 42789697-42789697 on Build GRCh38
rs74056880 Chr.13: 42789555-42789555 on Build GRCh38
rs149932932 Chr.13: 42789790-42789790 on Build GRCh38
rs898119253 Chr.13: 42789601-42789601 on Build GRCh38
rs1003955774 Chr.13: 42789859-42789859 on Build GRCh38
rs888510378 Chr.13: 42789741-42789741 on Build GRCh38
rs556628562 Chr.13: 42789859-42789860 on Build GRCh38

View Full Product Details